Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 38316663-38317162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG10B
Entrez Gene ID
144245
Gene Name
ALG10B, alpha-1,2-glucosyltransferase
Gene Aliases
ALG10, KCR1
UniGene
Hs.259305
Gene Chromosome Location
Chr.12: 38316679-38329726 on Build GRCh38
Transcripts
NM_001013620.3 NM_001308340.1 XM_005268665.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4931176 c.14A>G p.E5G Chr.12: 38316907-38316907 on Build GRCh38
69617 c.42C>A p.S14R Chr.12: 38316935-38316935 on Build GRCh38
7133687 c.7C>G p.Q3E Chr.12: 38316900-38316900 on Build GRCh38
5985857 c.30G>T p.S10S Chr.12: 38316923-38316923 on Build GRCh38
6598366 c.49delT p.L18fs*1 Chr.12: 38316942-38316942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930132389 Chr.12: 38316828-38316828 on Build GRCh38
rs752798471 Chr.12: 38317104-38317104 on Build GRCh38
rs200705857 Chr.12: 38316956-38316956 on Build GRCh38
rs769555561 Chr.12: 38317068-38317068 on Build GRCh38
rs373377379 Chr.12: 38316869-38316869 on Build GRCh38
rs553770263 Chr.12: 38317065-38317065 on Build GRCh38
rs1057385356 Chr.12: 38316797-38316797 on Build GRCh38
rs888155477 Chr.12: 38316689-38316689 on Build GRCh38
rs776686327 Chr.12: 38316998-38316998 on Build GRCh38
rs866239763 Chr.12: 38316912-38316912 on Build GRCh38
rs1005701481 Chr.12: 38316851-38316851 on Build GRCh38
rs149150389 Chr.12: 38316898-38316898 on Build GRCh38
rs200429393 Chr.12: 38316999-38316999 on Build GRCh38
rs7296576 Chr.12: 38316781-38316781 on Build GRCh38
rs12304754 Chr.12: 38316771-38316771 on Build GRCh38
rs758754153 Chr.12: 38316950-38316950 on Build GRCh38
rs775066889 Chr.12: 38317072-38317072 on Build GRCh38
rs757571673 Chr.12: 38317027-38317027 on Build GRCh38
rs1018330450 Chr.12: 38316706-38316706 on Build GRCh38
rs769896715 Chr.12: 38316976-38316976 on Build GRCh38
rs760677128 Chr.12: 38316909-38316909 on Build GRCh38
rs763672847 Chr.12: 38317015-38317015 on Build GRCh38
rs779228443 Chr.12: 38317045-38317045 on Build GRCh38
rs550478757 Chr.12: 38317133-38317133 on Build GRCh38
rs774190193 Chr.12: 38316914-38316914 on Build GRCh38
rs780169637 Chr.12: 38317033-38317033 on Build GRCh38
rs574255128 Chr.12: 38316931-38316931 on Build GRCh38
rs1005239303 Chr.12: 38316690-38316690 on Build GRCh38
rs756216725 Chr.12: 38317032-38317032 on Build GRCh38
rs554731852 Chr.12: 38317126-38317126 on Build GRCh38
rs927265640 Chr.12: 38316795-38316795 on Build GRCh38
rs1047241159 Chr.12: 38316830-38316830 on Build GRCh38
rs752360964 Chr.12: 38316940-38316940 on Build GRCh38
rs561475549 Chr.12: 38317052-38317052 on Build GRCh38
rs763239713 Chr.12: 38316832-38316832 on Build GRCh38
rs111665234 Chr.12: 38316805-38316805 on Build GRCh38
rs769944478 Chr.12: 38316895-38316895 on Build GRCh38
rs979249885 Chr.12: 38317054-38317054 on Build GRCh38
rs752346329 Chr.12: 38317000-38317000 on Build GRCh38
rs768953246 Chr.12: 38317036-38317036 on Build GRCh38
rs749522473 Chr.12: 38316984-38316984 on Build GRCh38
rs767671919 Chr.12: 38317004-38317004 on Build GRCh38
rs571178968 Chr.12: 38316767-38316767 on Build GRCh38
rs141825041 Chr.12: 38316997-38316997 on Build GRCh38
rs767865160 Chr.12: 38317116-38317116 on Build GRCh38
rs370286437 Chr.12: 38316978-38316978 on Build GRCh38
rs188043534 Chr.12: 38316992-38316992 on Build GRCh38
rs757770173 Chr.12: 38316959-38316959 on Build GRCh38
rs376126647 Chr.12: 38316935-38316935 on Build GRCh38
rs879004334 Chr.12: 38316842-38316842 on Build GRCh38
rs769210696 Chr.12: 38316779-38316779 on Build GRCh38
rs746446010 Chr.12: 38316775-38316775 on Build GRCh38
rs778792747 Chr.12: 38317134-38317134 on Build GRCh38
rs749270475 Chr.12: 38317034-38317034 on Build GRCh38
rs386762265 Chr.12: 38316780-38316781 on Build GRCh38
rs776656733 Chr.12: 38316692-38316692 on Build GRCh38
rs751868373 Chr.12: 38316957-38316957 on Build GRCh38
rs937646733 Chr.12: 38316796-38316796 on Build GRCh38
rs776773958 Chr.12: 38317056-38317056 on Build GRCh38
rs548024560 Chr.12: 38316724-38316724 on Build GRCh38
rs762334708 Chr.12: 38317083-38317083 on Build GRCh38
rs199628866 Chr.12: 38316960-38316960 on Build GRCh38
rs534768937 Chr.12: 38316808-38316808 on Build GRCh38
rs148038148 Chr.12: 38317028-38317028 on Build GRCh38
rs888217982 Chr.12: 38316841-38316841 on Build GRCh38
rs368700098 Chr.12: 38317013-38317013 on Build GRCh38
rs559045503 Chr.12: 38316788-38316788 on Build GRCh38
rs751193700 Chr.12: 38317110-38317110 on Build GRCh38
rs868484225 Chr.12: 38316814-38316814 on Build GRCh38
rs7315028 Chr.12: 38316721-38316721 on Build GRCh38
rs550165710 Chr.12: 38316773-38316773 on Build GRCh38
rs750911656 Chr.12: 38317030-38317030 on Build GRCh38
rs138776092 Chr.12: 38316969-38316969 on Build GRCh38
rs746847185 Chr.12: 38317049-38317049 on Build GRCh38
rs554349592 Chr.12: 38316865-38316865 on Build GRCh38
rs776709774 Chr.12: 38316883-38316883 on Build GRCh38
rs775735667 Chr.12: 38316896-38316896 on Build GRCh38
rs773351003 Chr.12: 38316901-38316901 on Build GRCh38
rs74618076 Chr.12: 38317017-38317017 on Build GRCh38
rs745885600 Chr.12: 38316887-38316887 on Build GRCh38
rs753430478 Chr.12: 38316926-38316926 on Build GRCh38
rs766352413 Chr.12: 38316920-38316920 on Build GRCh38
rs754460276 Chr.12: 38316927-38316927 on Build GRCh38
rs762207475 Chr.12: 38317113-38317113 on Build GRCh38
rs771821564 Chr.12: 38316990-38316990 on Build GRCh38
rs567831127 Chr.12: 38316764-38316764 on Build GRCh38
rs530313950 Chr.12: 38317128-38317128 on Build GRCh38
rs570080512 Chr.12: 38316780-38316780 on Build GRCh38
rs759748370 Chr.12: 38316956-38316958 on Build GRCh38
rs761926915 Chr.12: 38316712-38316712 on Build GRCh38
rs759501920 Chr.12: 38317057-38317057 on Build GRCh38
rs761564225 Chr.12: 38316897-38316897 on Build GRCh38
rs370840298 Chr.12: 38317092-38317092 on Build GRCh38
rs747506315 Chr.12: 38316880-38316880 on Build GRCh38

View Full Product Details