Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 38320749-38321281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG10B
Entrez Gene ID
144245
Gene Name
ALG10B, alpha-1,2-glucosyltransferase
Gene Aliases
ALG10, KCR1
UniGene
Hs.259305
Gene Chromosome Location
Chr.12: 38316679-38329726 on Build GRCh38
Transcripts
NM_001013620.3 NM_001308340.1 XM_005268665.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6501006 c.1177A>T p.I393F Chr.12: 38320968-38320968 on Build GRCh38
6537653 c.1130A>G p.Y377C Chr.12: 38320921-38320921 on Build GRCh38
1361299 c.1039T>C p.L347L Chr.12: 38320830-38320830 on Build GRCh38
3460376 c.1246C>T p.R416C Chr.12: 38321037-38321037 on Build GRCh38
3460377 c.1416G>A p.M472I Chr.12: 38321207-38321207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368402159 Chr.12: 38320992-38320992 on Build GRCh38
rs61730283 Chr.12: 38321127-38321127 on Build GRCh38
rs905309907 Chr.12: 38321058-38321058 on Build GRCh38
rs746759268 Chr.12: 38320891-38320891 on Build GRCh38
rs763373115 Chr.12: 38320837-38320837 on Build GRCh38
rs192771324 Chr.12: 38320947-38320947 on Build GRCh38
rs965807201 Chr.12: 38321140-38321140 on Build GRCh38
rs993865780 Chr.12: 38321112-38321112 on Build GRCh38
rs758006139 Chr.12: 38321205-38321205 on Build GRCh38
rs765898488 Chr.12: 38321246-38321246 on Build GRCh38
rs773338826 Chr.12: 38320786-38320786 on Build GRCh38
rs540925724 Chr.12: 38321086-38321086 on Build GRCh38
rs770705820 Chr.12: 38320797-38320797 on Build GRCh38
rs777334687 Chr.12: 38321229-38321229 on Build GRCh38
rs772320377 Chr.12: 38321029-38321029 on Build GRCh38
rs143372601 Chr.12: 38321051-38321051 on Build GRCh38
rs754025912 Chr.12: 38321016-38321016 on Build GRCh38
rs767876437 Chr.12: 38320844-38320844 on Build GRCh38
rs750210623 Chr.12: 38321175-38321175 on Build GRCh38
rs761050680 Chr.12: 38321119-38321119 on Build GRCh38
rs746955343 Chr.12: 38320789-38320789 on Build GRCh38
rs375350214 Chr.12: 38321037-38321037 on Build GRCh38
rs531634792 Chr.12: 38320901-38320901 on Build GRCh38
rs774733129 Chr.12: 38320921-38320921 on Build GRCh38
rs762278761 Chr.12: 38321013-38321013 on Build GRCh38
rs757790239 Chr.12: 38320843-38320843 on Build GRCh38
rs558917050 Chr.12: 38321184-38321187 on Build GRCh38
rs562512926 Chr.12: 38320880-38320880 on Build GRCh38
rs776236525 Chr.12: 38320805-38320805 on Build GRCh38
rs751866711 Chr.12: 38320842-38320842 on Build GRCh38
rs766344970 Chr.12: 38321185-38321185 on Build GRCh38
rs762609458 Chr.12: 38320917-38320917 on Build GRCh38
rs1028273507 Chr.12: 38320798-38320798 on Build GRCh38
rs375293238 Chr.12: 38321046-38321046 on Build GRCh38
rs35518352 Chr.12: 38321198-38321198 on Build GRCh38
rs955736476 Chr.12: 38321181-38321181 on Build GRCh38
rs750168940 Chr.12: 38321073-38321073 on Build GRCh38
rs142760513 Chr.12: 38320849-38320849 on Build GRCh38
rs1054385340 Chr.12: 38320953-38320953 on Build GRCh38
rs781253829 Chr.12: 38320975-38320975 on Build GRCh38
rs779228432 Chr.12: 38320954-38320954 on Build GRCh38
rs772570711 Chr.12: 38321075-38321075 on Build GRCh38
rs758917010 Chr.12: 38320813-38320813 on Build GRCh38
rs757131129 Chr.12: 38321059-38321059 on Build GRCh38
rs771614193 Chr.12: 38321125-38321125 on Build GRCh38
rs764985683 Chr.12: 38320814-38320814 on Build GRCh38
rs868523220 Chr.12: 38321053-38321053 on Build GRCh38
rs371578262 Chr.12: 38321005-38321005 on Build GRCh38
rs777713108 Chr.12: 38320889-38320889 on Build GRCh38
rs373473655 Chr.12: 38321191-38321191 on Build GRCh38
rs200428663 Chr.12: 38320937-38320937 on Build GRCh38
rs375328893 Chr.12: 38320866-38320866 on Build GRCh38
rs754544107 Chr.12: 38321192-38321192 on Build GRCh38
rs764340429 Chr.12: 38321173-38321173 on Build GRCh38
rs751415026 Chr.12: 38320957-38320957 on Build GRCh38
rs567006133 Chr.12: 38321047-38321047 on Build GRCh38
rs371189234 Chr.12: 38320836-38320836 on Build GRCh38
rs768323028 Chr.12: 38321006-38321006 on Build GRCh38
rs775178812 Chr.12: 38320911-38320911 on Build GRCh38
rs777456744 Chr.12: 38321207-38321207 on Build GRCh38
rs759172569 Chr.12: 38320923-38320923 on Build GRCh38
rs770922821 Chr.12: 38320895-38320895 on Build GRCh38
rs150044231 Chr.12: 38321143-38321143 on Build GRCh38
rs753993282 Chr.12: 38321042-38321042 on Build GRCh38
rs764273464 Chr.12: 38321244-38321244 on Build GRCh38
rs897819934 Chr.12: 38321085-38321085 on Build GRCh38
rs773970056 Chr.12: 38321011-38321011 on Build GRCh38
rs746389211 Chr.12: 38321233-38321233 on Build GRCh38
rs760738986 Chr.12: 38320985-38320986 on Build GRCh38
rs372347662 Chr.12: 38321038-38321038 on Build GRCh38
rs61730285 Chr.12: 38321135-38321135 on Build GRCh38
rs764090946 Chr.12: 38320986-38320986 on Build GRCh38
rs182557966 Chr.12: 38321079-38321079 on Build GRCh38
rs952215654 Chr.12: 38320821-38320821 on Build GRCh38
rs57963306 Chr.12: 38320939-38320939 on Build GRCh38
rs376132127 Chr.12: 38321242-38321242 on Build GRCh38
rs774601086 Chr.12: 38321247-38321247 on Build GRCh38
rs201414593 Chr.12: 38321076-38321076 on Build GRCh38
rs768119321 Chr.12: 38320932-38320932 on Build GRCh38
rs760534989 Chr.12: 38321249-38321249 on Build GRCh38
rs771417808 Chr.12: 38321228-38321228 on Build GRCh38
rs555931622 Chr.12: 38321090-38321090 on Build GRCh38
rs762480868 Chr.12: 38321123-38321123 on Build GRCh38
rs776406232 Chr.12: 38320897-38320897 on Build GRCh38
rs147370346 Chr.12: 38320886-38320886 on Build GRCh38
rs763672694 Chr.12: 38320919-38320919 on Build GRCh38
rs184416396 Chr.12: 38321178-38321178 on Build GRCh38
rs371467443 Chr.12: 38321070-38321070 on Build GRCh38
rs778301838 Chr.12: 38321196-38321196 on Build GRCh38
rs753902115 Chr.12: 38320944-38320944 on Build GRCh38
rs551343568 Chr.12: 38320903-38320903 on Build GRCh38
rs750864980 Chr.12: 38320933-38320933 on Build GRCh38
rs778694676 Chr.12: 38321093-38321093 on Build GRCh38
rs775464689 Chr.12: 38321236-38321236 on Build GRCh38
rs756126762 Chr.12: 38320865-38320865 on Build GRCh38
rs752761844 Chr.12: 38321153-38321153 on Build GRCh38
rs773478474 Chr.12: 38321117-38321117 on Build GRCh38
rs529138537 Chr.12: 38320839-38320839 on Build GRCh38
rs780465104 Chr.12: 38320877-38320877 on Build GRCh38
rs745777753 Chr.12: 38320989-38320989 on Build GRCh38
rs747745077 Chr.12: 38321095-38321095 on Build GRCh38
rs763092524 Chr.12: 38321163-38321163 on Build GRCh38
rs188303169 Chr.12: 38321102-38321102 on Build GRCh38
rs753774176 Chr.12: 38321187-38321187 on Build GRCh38
rs771686989 Chr.12: 38321114-38321114 on Build GRCh38
rs367596603 Chr.12: 38320965-38320965 on Build GRCh38
rs747413390 Chr.12: 38321222-38321222 on Build GRCh38

View Full Product Details