Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 112823253-112823746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP11A
Entrez Gene ID
23250
Gene Name
ATPase phospholipid transporting 11A
Gene Aliases
ATPIH, ATPIS
UniGene
Hs.29189
Gene Chromosome Location
Chr.13: 112690034-112887168 on Build GRCh38
Transcripts
NM_015205.2 NM_032189.3 XM_005268299.4 XM_005268300.4 XM_005268301.4 XM_005268303.4 XM_005268305.4 XM_005268306.4 XM_011537480.2 XM_017020490.1 XM_017020491.1 XM_017020492.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5016437 c.780_781insA p.I262fs*60 Chr.13: 112823399-112823400 on Build GRCh38
5016438 c.780_781insA p.I262fs*60 Chr.13: 112823399-112823400 on Build GRCh38
1365532 c.787delT p.F263fs*15 Chr.13: 112823406-112823406 on Build GRCh38
5832160 c.735_736insTC p.E247fs*11 Chr.13: 112823354-112823355 on Build GRCh38
301726 c.738G>A p.S246S Chr.13: 112823357-112823357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1052960752 Chr.13: 112823368-112823368 on Build GRCh38
rs375961305 Chr.13: 112823280-112823280 on Build GRCh38
rs947221021 Chr.13: 112823606-112823606 on Build GRCh38
rs772523895 Chr.13: 112823435-112823435 on Build GRCh38
rs538180289 Chr.13: 112823284-112823284 on Build GRCh38
rs776903712 Chr.13: 112823484-112823484 on Build GRCh38
rs562245842 Chr.13: 112823547-112823547 on Build GRCh38
rs541263319 Chr.13: 112823579-112823579 on Build GRCh38
rs911326139 Chr.13: 112823535-112823535 on Build GRCh38
rs961418240 Chr.13: 112823272-112823272 on Build GRCh38
rs547966799 Chr.13: 112823711-112823711 on Build GRCh38
rs1035398662 Chr.13: 112823640-112823640 on Build GRCh38
rs557013301 Chr.13: 112823381-112823381 on Build GRCh38
rs185765447 Chr.13: 112823503-112823503 on Build GRCh38
rs530816164 Chr.13: 112823659-112823659 on Build GRCh38
rs749725054 Chr.13: 112823321-112823321 on Build GRCh38
rs535490283 Chr.13: 112823372-112823372 on Build GRCh38
rs1044137830 Chr.13: 112823607-112823607 on Build GRCh38
rs969688553 Chr.13: 112823478-112823478 on Build GRCh38
rs1011359701 Chr.13: 112823394-112823394 on Build GRCh38
rs781082921 Chr.13: 112823309-112823309 on Build GRCh38
rs751537379 Chr.13: 112823348-112823348 on Build GRCh38
rs867818726 Chr.13: 112823497-112823497 on Build GRCh38
rs777729193 Chr.13: 112823304-112823304 on Build GRCh38
rs574689150 Chr.13: 112823534-112823534 on Build GRCh38
rs372760786 Chr.13: 112823330-112823330 on Build GRCh38
rs376191916 Chr.13: 112823315-112823315 on Build GRCh38
rs985607492 Chr.13: 112823710-112823710 on Build GRCh38
rs892883834 Chr.13: 112823369-112823369 on Build GRCh38
rs572204344 Chr.13: 112823479-112823479 on Build GRCh38
rs533298728 Chr.13: 112823613-112823613 on Build GRCh38
rs571890934 Chr.13: 112823656-112823656 on Build GRCh38
rs769228938 Chr.13: 112823443-112823443 on Build GRCh38
rs79761235 Chr.13: 112823672-112823672 on Build GRCh38
rs756246398 Chr.13: 112823384-112823384 on Build GRCh38
rs758193486 Chr.13: 112823334-112823334 on Build GRCh38
rs764928404 Chr.13: 112823333-112823333 on Build GRCh38
rs544992259 Chr.13: 112823614-112823614 on Build GRCh38
rs776054852 Chr.13: 112823439-112823439 on Build GRCh38
rs146632559 Chr.13: 112823308-112823308 on Build GRCh38
rs976788398 Chr.13: 112823537-112823537 on Build GRCh38
rs369624424 Chr.13: 112823329-112823329 on Build GRCh38
rs778555743 Chr.13: 112823660-112823660 on Build GRCh38
rs766123805 Chr.13: 112823338-112823338 on Build GRCh38
rs973306457 Chr.13: 112823490-112823490 on Build GRCh38
rs990691537 Chr.13: 112823290-112823290 on Build GRCh38
rs1026932195 Chr.13: 112823283-112823284 on Build GRCh38
rs559400341 Chr.13: 112823598-112823598 on Build GRCh38
rs772883042 Chr.13: 112823449-112823449 on Build GRCh38
rs1053349142 Chr.13: 112823570-112823570 on Build GRCh38
rs771450533 Chr.13: 112823409-112823409 on Build GRCh38
rs999060847 Chr.13: 112823367-112823367 on Build GRCh38
rs773561782 Chr.13: 112823316-112823316 on Build GRCh38
rs771360900 Chr.13: 112823322-112823322 on Build GRCh38
rs1040673048 Chr.13: 112823319-112823319 on Build GRCh38
rs576652353 Chr.13: 112823523-112823523 on Build GRCh38
rs575580526 Chr.13: 112823515-112823516 on Build GRCh38
rs779197620 Chr.13: 112823427-112823427 on Build GRCh38
rs752785696 Chr.13: 112823366-112823366 on Build GRCh38
rs1037131468 Chr.13: 112823496-112823496 on Build GRCh38
rs754920317 Chr.13: 112823356-112823356 on Build GRCh38
rs914862489 Chr.13: 112823410-112823410 on Build GRCh38
rs373731087 Chr.13: 112823432-112823432 on Build GRCh38
rs373565552 Chr.13: 112823314-112823314 on Build GRCh38
rs553870684 Chr.13: 112823476-112823476 on Build GRCh38
rs375717586 Chr.13: 112823295-112823295 on Build GRCh38
rs758800977 Chr.13: 112823647-112823647 on Build GRCh38
rs978301626 Chr.13: 112823569-112823569 on Build GRCh38
rs146522718 Chr.13: 112823407-112823407 on Build GRCh38
rs897283860 Chr.13: 112823677-112823677 on Build GRCh38
rs574147499 Chr.13: 112823552-112823552 on Build GRCh38
rs3928842 Chr.13: 112823430-112823430 on Build GRCh38
rs1022618341 Chr.13: 112823411-112823411 on Build GRCh38
rs778153151 Chr.13: 112823402-112823402 on Build GRCh38
rs895152223 Chr.13: 112823720-112823720 on Build GRCh38
rs905617543 Chr.13: 112823610-112823610 on Build GRCh38
rs762544966 Chr.13: 112823453-112823453 on Build GRCh38
rs756708779 Chr.13: 112823510-112823510 on Build GRCh38
rs921609780 Chr.13: 112823622-112823622 on Build GRCh38
rs761384079 Chr.13: 112823332-112823332 on Build GRCh38
rs961650992 Chr.13: 112823485-112823485 on Build GRCh38
rs892997459 Chr.13: 112823600-112823600 on Build GRCh38
rs746217736 Chr.13: 112823434-112823434 on Build GRCh38
rs776049128 Chr.13: 112823331-112823331 on Build GRCh38
rs761245227 Chr.13: 112823441-112823441 on Build GRCh38
rs368991136 Chr.13: 112823289-112823289 on Build GRCh38
rs765565842 Chr.13: 112823536-112823536 on Build GRCh38
rs535671823 Chr.13: 112823357-112823357 on Build GRCh38
rs746854753 Chr.13: 112823298-112823298 on Build GRCh38
rs375124079 Chr.13: 112823483-112823483 on Build GRCh38

View Full Product Details