Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 114324910-114325417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHAMP1
Entrez Gene ID
283489
Gene Name
chromosome alignment maintaining phosphoprotein 1
Gene Aliases
C13orf8, CAMP, CHAMP, MRD40, ZNF828
UniGene
Hs.7542
Gene Chromosome Location
Chr.13: 114314490-114327328 on Build GRCh38
Transcripts
NM_001164144.1 NM_001164145.1 NM_032436.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6074507 c.1253A>C p.E418A Chr.13: 114325095-114325095 on Build GRCh38
2264760 c.1098T>G p.S366S Chr.13: 114324940-114324940 on Build GRCh38
1234675 c.1258C>T p.R420C Chr.13: 114325100-114325100 on Build GRCh38
3793160 c.1328C>T p.S443L Chr.13: 114325170-114325170 on Build GRCh38
2264763 c.1413T>C p.S471S Chr.13: 114325255-114325255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782132271 Chr.13: 114325259-114325259 on Build GRCh38
rs782051660 Chr.13: 114324933-114324933 on Build GRCh38
rs529321452 Chr.13: 114325026-114325026 on Build GRCh38
rs142049023 Chr.13: 114325321-114325321 on Build GRCh38
rs374095161 Chr.13: 114325209-114325209 on Build GRCh38
rs548630533 Chr.13: 114325260-114325260 on Build GRCh38
rs138325089 Chr.13: 114325001-114325001 on Build GRCh38
rs782623150 Chr.13: 114325328-114325328 on Build GRCh38
rs781985494 Chr.13: 114325056-114325056 on Build GRCh38
rs782157659 Chr.13: 114325196-114325197 on Build GRCh38
rs371126156 Chr.13: 114325153-114325153 on Build GRCh38
rs1023708288 Chr.13: 114325057-114325057 on Build GRCh38
rs782397980 Chr.13: 114325331-114325331 on Build GRCh38
rs201826600 Chr.13: 114325159-114325159 on Build GRCh38
rs782248613 Chr.13: 114325330-114325330 on Build GRCh38
rs782714399 Chr.13: 114324934-114324934 on Build GRCh38
rs139821586 Chr.13: 114325317-114325317 on Build GRCh38
rs782647633 Chr.13: 114325215-114325215 on Build GRCh38
rs782283006 Chr.13: 114325108-114325108 on Build GRCh38
rs375854088 Chr.13: 114325368-114325368 on Build GRCh38
rs935216320 Chr.13: 114325362-114325362 on Build GRCh38
rs782358795 Chr.13: 114325347-114325347 on Build GRCh38
rs782085677 Chr.13: 114325345-114325345 on Build GRCh38
rs920062428 Chr.13: 114325207-114325207 on Build GRCh38
rs782052857 Chr.13: 114325262-114325262 on Build GRCh38
rs150434475 Chr.13: 114324961-114324961 on Build GRCh38
rs782448533 Chr.13: 114325199-114325199 on Build GRCh38
rs893582610 Chr.13: 114325365-114325365 on Build GRCh38
rs199550269 Chr.13: 114325298-114325298 on Build GRCh38
rs781847798 Chr.13: 114325101-114325101 on Build GRCh38
rs782564405 Chr.13: 114325109-114325109 on Build GRCh38
rs150753986 Chr.13: 114325363-114325363 on Build GRCh38
rs543967281 Chr.13: 114324995-114324995 on Build GRCh38
rs781895659 Chr.13: 114325086-114325086 on Build GRCh38
rs782386969 Chr.13: 114325144-114325144 on Build GRCh38
rs142250564 Chr.13: 114325266-114325266 on Build GRCh38
rs144334556 Chr.13: 114325120-114325120 on Build GRCh38
rs370765566 Chr.13: 114325200-114325200 on Build GRCh38
rs781990427 Chr.13: 114325244-114325244 on Build GRCh38
rs782662682 Chr.13: 114325337-114325337 on Build GRCh38
rs782758094 Chr.13: 114325179-114325179 on Build GRCh38
rs797044962 Chr.13: 114325034-114325034 on Build GRCh38
rs547434000 Chr.13: 114325134-114325134 on Build GRCh38
rs374334158 Chr.13: 114325146-114325146 on Build GRCh38
rs985509991 Chr.13: 114325228-114325228 on Build GRCh38
rs782128382 Chr.13: 114325065-114325065 on Build GRCh38
rs782275339 Chr.13: 114325216-114325216 on Build GRCh38
rs782561524 Chr.13: 114325224-114325224 on Build GRCh38
rs782361734 Chr.13: 114325241-114325241 on Build GRCh38
rs782752774 Chr.13: 114325100-114325100 on Build GRCh38
rs367949169 Chr.13: 114325187-114325187 on Build GRCh38
rs782224574 Chr.13: 114325208-114325208 on Build GRCh38
rs146382660 Chr.13: 114325314-114325314 on Build GRCh38
rs35076210 Chr.13: 114325108-114325109 on Build GRCh38
rs782814233 Chr.13: 114325189-114325189 on Build GRCh38
rs782187552 Chr.13: 114325237-114325237 on Build GRCh38
rs149624011 Chr.13: 114325051-114325051 on Build GRCh38
rs781825292 Chr.13: 114325295-114325295 on Build GRCh38
rs782609105 Chr.13: 114325201-114325201 on Build GRCh38
rs782338415 Chr.13: 114325133-114325133 on Build GRCh38
rs781934532 Chr.13: 114325344-114325344 on Build GRCh38
rs1040913799 Chr.13: 114324931-114324931 on Build GRCh38
rs552043197 Chr.13: 114325313-114325313 on Build GRCh38
rs781819969 Chr.13: 114324950-114324950 on Build GRCh38
rs367684543 Chr.13: 114325352-114325352 on Build GRCh38
rs782548665 Chr.13: 114324969-114324969 on Build GRCh38
rs781792138 Chr.13: 114325196-114325196 on Build GRCh38
rs573576960 Chr.13: 114325166-114325166 on Build GRCh38
rs201708961 Chr.13: 114325151-114325151 on Build GRCh38
rs782418326 Chr.13: 114325221-114325221 on Build GRCh38
rs138201298 Chr.13: 114324990-114324990 on Build GRCh38
rs782510090 Chr.13: 114325107-114325107 on Build GRCh38
rs782678181 Chr.13: 114325318-114325318 on Build GRCh38
rs781865278 Chr.13: 114324963-114324963 on Build GRCh38
rs782782357 Chr.13: 114325074-114325074 on Build GRCh38
rs782021774 Chr.13: 114325021-114325021 on Build GRCh38
rs782393001 Chr.13: 114325009-114325009 on Build GRCh38
rs782454954 Chr.13: 114325096-114325096 on Build GRCh38
rs376497995 Chr.13: 114325035-114325035 on Build GRCh38
rs373905564 Chr.13: 114324967-114324967 on Build GRCh38
rs781880614 Chr.13: 114325184-114325184 on Build GRCh38
rs782251855 Chr.13: 114325002-114325002 on Build GRCh38
rs782292119 Chr.13: 114325339-114325339 on Build GRCh38
rs372468779 Chr.13: 114325168-114325168 on Build GRCh38
rs863225075 Chr.13: 114325386-114325386 on Build GRCh38
rs370703699 Chr.13: 114325043-114325043 on Build GRCh38
rs782313622 Chr.13: 114325342-114325342 on Build GRCh38
rs782192484 Chr.13: 114325130-114325130 on Build GRCh38
rs782242916 Chr.13: 114325327-114325327 on Build GRCh38

View Full Product Details