Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15461591-15462131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELA2A
Entrez Gene ID
63036
Gene Name
chymotrypsin like elastase family member 2A
Gene Aliases
ELA2A, PE-1
UniGene
Hs.631866
Gene Chromosome Location
Chr.1: 15456728-15472091 on Build GRCh38
Transcripts
NM_033440.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6370578 c.173G>A p.C58Y Chr.1: 15461604-15461604 on Build GRCh38
676231 c.206T>A p.L69Q Chr.1: 15461637-15461637 on Build GRCh38
897607 c.210G>A p.T70T Chr.1: 15461641-15461641 on Build GRCh38
5077547 c.227+1_227+2insTAAC p.? Chr.1: 15461659-15461660 on Build GRCh38
1985569 c.221G>A p.C74Y Chr.1: 15461652-15461652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766553806 Chr.1: 15461778-15461778 on Build GRCh38
rs761214770 Chr.1: 15461835-15461835 on Build GRCh38
rs773598391 Chr.1: 15461769-15461769 on Build GRCh38
rs544183163 Chr.1: 15461891-15461891 on Build GRCh38
rs567782707 Chr.1: 15461967-15461967 on Build GRCh38
rs777430213 Chr.1: 15461699-15461699 on Build GRCh38
rs778765956 Chr.1: 15461693-15461693 on Build GRCh38
rs746539560 Chr.1: 15461653-15461653 on Build GRCh38
rs3737696 Chr.1: 15461774-15461774 on Build GRCh38
rs772288206 Chr.1: 15461764-15461764 on Build GRCh38
rs756956727 Chr.1: 15461804-15461804 on Build GRCh38
rs780152288 Chr.1: 15461621-15461621 on Build GRCh38
rs747823974 Chr.1: 15461642-15461642 on Build GRCh38
rs960091225 Chr.1: 15461790-15461790 on Build GRCh38
rs759452813 Chr.1: 15461786-15461786 on Build GRCh38
rs767618149 Chr.1: 15461610-15461610 on Build GRCh38
rs1021166870 Chr.1: 15461942-15461942 on Build GRCh38
rs1003787979 Chr.1: 15461981-15461981 on Build GRCh38
rs573139696 Chr.1: 15461751-15461751 on Build GRCh38
rs774632258 Chr.1: 15461671-15461671 on Build GRCh38
rs755454047 Chr.1: 15462095-15462095 on Build GRCh38
rs764744472 Chr.1: 15461838-15461838 on Build GRCh38
rs180861476 Chr.1: 15461678-15461678 on Build GRCh38
rs766607053 Chr.1: 15461682-15461682 on Build GRCh38
rs771690549 Chr.1: 15461649-15461649 on Build GRCh38
rs1048323325 Chr.1: 15461612-15461612 on Build GRCh38
rs754040262 Chr.1: 15461780-15461780 on Build GRCh38
rs908195126 Chr.1: 15462099-15462099 on Build GRCh38
rs769039006 Chr.1: 15461668-15461668 on Build GRCh38
rs370563283 Chr.1: 15461691-15461691 on Build GRCh38
rs549387423 Chr.1: 15461954-15461954 on Build GRCh38
rs374563517 Chr.1: 15461698-15461698 on Build GRCh38
rs1010841141 Chr.1: 15461941-15461941 on Build GRCh38
rs749042017 Chr.1: 15461629-15461629 on Build GRCh38
rs901003403 Chr.1: 15461916-15461916 on Build GRCh38
rs772715037 Chr.1: 15461650-15461650 on Build GRCh38
rs529103064 Chr.1: 15462059-15462059 on Build GRCh38
rs867193439 Chr.1: 15461684-15461684 on Build GRCh38
rs115788608 Chr.1: 15462007-15462007 on Build GRCh38
rs983748122 Chr.1: 15462083-15462083 on Build GRCh38
rs930952977 Chr.1: 15461843-15461843 on Build GRCh38
rs879016266 Chr.1: 15462069-15462069 on Build GRCh38
rs377663645 Chr.1: 15461641-15461641 on Build GRCh38
rs1049789637 Chr.1: 15461939-15461939 on Build GRCh38
rs770239262 Chr.1: 15461657-15461657 on Build GRCh38
rs769107134 Chr.1: 15461755-15461755 on Build GRCh38
rs1041310519 Chr.1: 15461912-15461912 on Build GRCh38
rs776011341 Chr.1: 15461665-15461665 on Build GRCh38
rs531164738 Chr.1: 15461940-15461940 on Build GRCh38
rs750597578 Chr.1: 15461614-15461614 on Build GRCh38
rs763489011 Chr.1: 15461666-15461666 on Build GRCh38
rs746486230 Chr.1: 15461702-15461702 on Build GRCh38
rs765103364 Chr.1: 15461787-15461787 on Build GRCh38
rs915601433 Chr.1: 15461842-15461842 on Build GRCh38
rs991264440 Chr.1: 15462031-15462031 on Build GRCh38
rs577810519 Chr.1: 15461968-15461968 on Build GRCh38
rs575099298 Chr.1: 15461848-15461848 on Build GRCh38
rs939872211 Chr.1: 15461907-15461907 on Build GRCh38
rs770394136 Chr.1: 15461748-15461748 on Build GRCh38
rs756049001 Chr.1: 15461619-15461619 on Build GRCh38
rs758163415 Chr.1: 15461795-15461795 on Build GRCh38
rs751187383 Chr.1: 15461797-15461797 on Build GRCh38
rs967329026 Chr.1: 15461972-15461972 on Build GRCh38
rs997364386 Chr.1: 15461918-15461918 on Build GRCh38
rs777588660 Chr.1: 15461796-15461796 on Build GRCh38
rs745373650 Chr.1: 15461861-15461861 on Build GRCh38
rs377324941 Chr.1: 15461685-15461685 on Build GRCh38
rs952375491 Chr.1: 15462082-15462082 on Build GRCh38
rs752490042 Chr.1: 15461794-15461794 on Build GRCh38
rs3737697 Chr.1: 15461996-15461996 on Build GRCh38
rs372947070 Chr.1: 15461640-15461640 on Build GRCh38
rs762088783 Chr.1: 15461674-15461674 on Build GRCh38
rs966924758 Chr.1: 15461773-15461773 on Build GRCh38
rs915634144 Chr.1: 15462077-15462077 on Build GRCh38
rs908431274 Chr.1: 15461903-15461903 on Build GRCh38
rs149138464 Chr.1: 15461754-15461754 on Build GRCh38
rs373809860 Chr.1: 15461679-15461679 on Build GRCh38

View Full Product Details