Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20806444-20806983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4G3
Entrez Gene ID
8672
Gene Name
eukaryotic translation initiation factor 4 gamma 3
Gene Aliases
eIF-4G 3, eIF4G 3, eIF4GII
UniGene
Hs.467084
Gene Chromosome Location
Chr.1: 20806292-21177320 on Build GRCh38
Transcripts
NM_001198801.1 NM_001198802.1 NM_003760.4 XM_011542356.2 XM_011542361.2 XM_011542370.1 XM_011542371.1 XM_011542374.1 XM_011542375.2 XM_011542381.2 XM_017002672.1 XM_017002673.1 XM_017002674.1 XM_017002675.1 XM_017002676.1 XM_017002677.1 XM_017002678.1 XM_017002679.1 XM_017002680.1 XM_017002681.1 XM_017002682.1 XM_017002683.1 XM_017002684.1 XM_017002685.1 XM_017002686.1 XM_017002687.1 XM_017002688.1 XM_017002689.1 XM_017002690.1 XM_017002691.1 XM_017002692.1 XM_017002693.1 XM_017002694.1 XM_017002695.1 XM_017002696.1 XM_017002697.1 XM_017002698.1 XM_017002699.1 XM_017002700.1 XM_017002701.1 XM_017002702.1 XM_017002703.1 XM_017002704.1 XM_017002705.1 XM_017002706.1 XM_017002707.1 XM_017002708.1 XM_017002709.1

SNP Detail

SNP ID Chromosome Location
rs376512912 Chr.1: 20806568-20806569 on Build GRCh38
rs749122429 Chr.1: 20806794-20806794 on Build GRCh38
rs1040267838 Chr.1: 20806880-20806880 on Build GRCh38
rs886239508 Chr.1: 20806808-20806808 on Build GRCh38
rs961773739 Chr.1: 20806853-20806853 on Build GRCh38
rs532703823 Chr.1: 20806652-20806653 on Build GRCh38
rs953164612 Chr.1: 20806924-20806924 on Build GRCh38
rs74522845 Chr.1: 20806722-20806722 on Build GRCh38
rs957122860 Chr.1: 20806938-20806938 on Build GRCh38
rs928647366 Chr.1: 20806581-20806581 on Build GRCh38
rs995833547 Chr.1: 20806890-20806890 on Build GRCh38
rs937380111 Chr.1: 20806594-20806594 on Build GRCh38
rs1056369624 Chr.1: 20806765-20806765 on Build GRCh38
rs549082543 Chr.1: 20806628-20806628 on Build GRCh38
rs748803491 Chr.1: 20806793-20806794 on Build GRCh38
rs952035662 Chr.1: 20806817-20806817 on Build GRCh38
rs946872202 Chr.1: 20806702-20806702 on Build GRCh38
rs968044211 Chr.1: 20806484-20806484 on Build GRCh38
rs1014795309 Chr.1: 20806848-20806848 on Build GRCh38
rs530545337 Chr.1: 20806576-20806576 on Build GRCh38
rs1012133672 Chr.1: 20806806-20806806 on Build GRCh38
rs981690825 Chr.1: 20806568-20806568 on Build GRCh38
rs1048927183 Chr.1: 20806794-20806794 on Build GRCh38
rs922927402 Chr.1: 20806920-20806920 on Build GRCh38
rs535341005 Chr.1: 20806830-20806830 on Build GRCh38
rs1055788941 Chr.1: 20806720-20806720 on Build GRCh38
rs901744811 Chr.1: 20806883-20806883 on Build GRCh38
rs931382059 Chr.1: 20806747-20806747 on Build GRCh38
rs1028614544 Chr.1: 20806937-20806937 on Build GRCh38
rs1043904490 Chr.1: 20806712-20806712 on Build GRCh38
rs922954131 Chr.1: 20806538-20806538 on Build GRCh38
rs975733785 Chr.1: 20806491-20806491 on Build GRCh38
rs934230146 Chr.1: 20806554-20806555 on Build GRCh38
rs953419188 Chr.1: 20806478-20806478 on Build GRCh38

View Full Product Details