Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23868086-23868605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUCA1
Entrez Gene ID
2517
Gene Name
fucosidase, alpha-L- 1, tissue
Gene Aliases
FUCA
UniGene
Hs.370858
Gene Chromosome Location
Chr.1: 23845077-23868369 on Build GRCh38
Transcripts
NM_000147.4 XM_005245821.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5735328 c.151G>A p.A51T Chr.1: 23868136-23868136 on Build GRCh38
6367683 c.25C>T p.R9W Chr.1: 23868262-23868262 on Build GRCh38
6247538 c.82G>A p.V28M Chr.1: 23868205-23868205 on Build GRCh38
5705354 c.1A>G p.M1V Chr.1: 23868286-23868286 on Build GRCh38
7167184 c.139C>T p.R47W Chr.1: 23868148-23868148 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982769570 Chr.1: 23868473-23868473 on Build GRCh38
rs1011763920 Chr.1: 23868316-23868316 on Build GRCh38
rs960392301 Chr.1: 23868268-23868268 on Build GRCh38
rs1043240453 Chr.1: 23868527-23868527 on Build GRCh38
rs951497998 Chr.1: 23868438-23868438 on Build GRCh38
rs886046326 Chr.1: 23868257-23868257 on Build GRCh38
rs746017062 Chr.1: 23868272-23868272 on Build GRCh38
rs551560114 Chr.1: 23868220-23868220 on Build GRCh38
rs367641377 Chr.1: 23868150-23868150 on Build GRCh38
rs761603943 Chr.1: 23868242-23868242 on Build GRCh38
rs1021897274 Chr.1: 23868257-23868257 on Build GRCh38
rs749870910 Chr.1: 23868158-23868158 on Build GRCh38
rs2070956 Chr.1: 23868258-23868258 on Build GRCh38
rs763773855 Chr.1: 23868116-23868116 on Build GRCh38
rs747205141 Chr.1: 23868314-23868314 on Build GRCh38
rs534639612 Chr.1: 23868262-23868262 on Build GRCh38
rs2070953 Chr.1: 23868372-23868372 on Build GRCh38
rs750382845 Chr.1: 23868251-23868251 on Build GRCh38
rs887432872 Chr.1: 23868120-23868120 on Build GRCh38
rs754703239 Chr.1: 23868176-23868176 on Build GRCh38
rs780157255 Chr.1: 23868311-23868311 on Build GRCh38
rs111783498 Chr.1: 23868321-23868321 on Build GRCh38
rs899736238 Chr.1: 23868226-23868227 on Build GRCh38
rs778425491 Chr.1: 23868190-23868190 on Build GRCh38
rs1013453954 Chr.1: 23868379-23868379 on Build GRCh38
rs1025741959 Chr.1: 23868380-23868380 on Build GRCh38
rs748584281 Chr.1: 23868183-23868183 on Build GRCh38
rs901607680 Chr.1: 23868172-23868172 on Build GRCh38
rs550501788 Chr.1: 23868264-23868264 on Build GRCh38
rs567536324 Chr.1: 23868261-23868261 on Build GRCh38
rs756742678 Chr.1: 23868227-23868228 on Build GRCh38
rs993064338 Chr.1: 23868294-23868294 on Build GRCh38
rs972881768 Chr.1: 23868269-23868269 on Build GRCh38
rs933045040 Chr.1: 23868306-23868306 on Build GRCh38
rs750020102 Chr.1: 23868238-23868239 on Build GRCh38
rs909992905 Chr.1: 23868474-23868474 on Build GRCh38
rs558338123 Chr.1: 23868286-23868286 on Build GRCh38
rs780907796 Chr.1: 23868255-23868255 on Build GRCh38
rs950584296 Chr.1: 23868392-23868392 on Build GRCh38
rs561207660 Chr.1: 23868530-23868530 on Build GRCh38
rs776233426 Chr.1: 23868232-23868232 on Build GRCh38
rs373007532 Chr.1: 23868259-23868259 on Build GRCh38
rs79319279 Chr.1: 23868295-23868295 on Build GRCh38
rs764731226 Chr.1: 23868235-23868236 on Build GRCh38
rs753706036 Chr.1: 23868117-23868117 on Build GRCh38
rs573987543 Chr.1: 23868340-23868340 on Build GRCh38
rs115766287 Chr.1: 23868368-23868368 on Build GRCh38
rs1047146664 Chr.1: 23868115-23868115 on Build GRCh38
rs770365153 Chr.1: 23868185-23868185 on Build GRCh38
rs749337153 Chr.1: 23868192-23868192 on Build GRCh38
rs201209052 Chr.1: 23868181-23868181 on Build GRCh38
rs2070952 Chr.1: 23868408-23868408 on Build GRCh38
rs371134787 Chr.1: 23868171-23868171 on Build GRCh38
rs2070954 Chr.1: 23868298-23868298 on Build GRCh38
rs747889861 Chr.1: 23868276-23868276 on Build GRCh38
rs530854024 Chr.1: 23868253-23868253 on Build GRCh38
rs530646066 Chr.1: 23868482-23868482 on Build GRCh38
rs761732955 Chr.1: 23868136-23868136 on Build GRCh38
rs964594372 Chr.1: 23868409-23868409 on Build GRCh38
rs888280638 Chr.1: 23868141-23868141 on Build GRCh38
rs370638102 Chr.1: 23868492-23868492 on Build GRCh38
rs769555043 Chr.1: 23868279-23868279 on Build GRCh38
rs947502372 Chr.1: 23868498-23868498 on Build GRCh38
rs61996282 Chr.1: 23868280-23868280 on Build GRCh38
rs775815377 Chr.1: 23868275-23868275 on Build GRCh38
rs886046325 Chr.1: 23868120-23868120 on Build GRCh38
rs982906713 Chr.1: 23868299-23868299 on Build GRCh38
rs758528364 Chr.1: 23868309-23868309 on Build GRCh38
rs1014921634 Chr.1: 23868428-23868428 on Build GRCh38
rs139832548 Chr.1: 23868330-23868330 on Build GRCh38
rs765174598 Chr.1: 23868138-23868138 on Build GRCh38
rs745898413 Chr.1: 23868307-23868307 on Build GRCh38
rs549145692 Chr.1: 23868483-23868483 on Build GRCh38
rs1038737730 Chr.1: 23868167-23868167 on Build GRCh38
rs906070802 Chr.1: 23868575-23868575 on Build GRCh38
rs769914190 Chr.1: 23868414-23868414 on Build GRCh38
rs751151042 Chr.1: 23868254-23868254 on Build GRCh38
rs569701574 Chr.1: 23868250-23868250 on Build GRCh38
rs781211392 Chr.1: 23868301-23868301 on Build GRCh38
rs749125399 Chr.1: 23868324-23868324 on Build GRCh38
rs903593217 Chr.1: 23868387-23868388 on Build GRCh38
rs752415554 Chr.1: 23868290-23868290 on Build GRCh38
rs1035690676 Chr.1: 23868353-23868353 on Build GRCh38
rs891929720 Chr.1: 23868313-23868313 on Build GRCh38
rs767034274 Chr.1: 23868287-23868287 on Build GRCh38
rs143545485 Chr.1: 23868175-23868175 on Build GRCh38
rs774464015 Chr.1: 23868109-23868109 on Build GRCh38
rs768938319 Chr.1: 23868318-23868318 on Build GRCh38
rs760470041 Chr.1: 23868113-23868113 on Build GRCh38
rs2070955 Chr.1: 23868283-23868283 on Build GRCh38
rs368369988 Chr.1: 23868187-23868187 on Build GRCh38
rs912982550 Chr.1: 23868484-23868484 on Build GRCh38
rs921105568 Chr.1: 23868489-23868489 on Build GRCh38
rs771153989 Chr.1: 23868194-23868194 on Build GRCh38
rs749698038 Chr.1: 23868303-23868303 on Build GRCh38
rs533240590 Chr.1: 23868217-23868217 on Build GRCh38
rs772980237 Chr.1: 23868322-23868322 on Build GRCh38
rs563520700 Chr.1: 23868430-23868430 on Build GRCh38
rs185764829 Chr.1: 23868400-23868400 on Build GRCh38
rs371987465 Chr.1: 23868300-23868300 on Build GRCh38
rs1044401721 Chr.1: 23868391-23868391 on Build GRCh38
rs774539460 Chr.1: 23868196-23868196 on Build GRCh38
rs1001821759 Chr.1: 23868337-23868337 on Build GRCh38
rs770703662 Chr.1: 23868329-23868329 on Build GRCh38
rs933801267 Chr.1: 23868503-23868504 on Build GRCh38

View Full Product Details