Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24120466-24120966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL22RA1
Entrez Gene ID
58985
Gene Name
interleukin 22 receptor subunit alpha 1
Gene Aliases
CRF2-9, IL22R, IL22R1
UniGene
Hs.110915
Gene Chromosome Location
Chr.1: 24119771-24143285 on Build GRCh38
Transcripts
NM_021258.3 XM_011541882.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3486371 c.1685T>A p.L562H Chr.1: 24120845-24120845 on Build GRCh38
3735352 c.1568C>T p.S523L Chr.1: 24120962-24120962 on Build GRCh38
7134018 c.1682C>G p.S561C Chr.1: 24120848-24120848 on Build GRCh38
906652 c.1670C>T p.T557I Chr.1: 24120860-24120860 on Build GRCh38
311977 c.1637delC p.P546fs*23 Chr.1: 24120893-24120893 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1033371175 Chr.1: 24120701-24120701 on Build GRCh38
rs892294356 Chr.1: 24120598-24120598 on Build GRCh38
rs899716714 Chr.1: 24120556-24120556 on Build GRCh38
rs560313967 Chr.1: 24120495-24120495 on Build GRCh38
rs776887352 Chr.1: 24120871-24120871 on Build GRCh38
rs1004079065 Chr.1: 24120562-24120562 on Build GRCh38
rs1007929032 Chr.1: 24120849-24120849 on Build GRCh38
rs755634348 Chr.1: 24120804-24120804 on Build GRCh38
rs111645604 Chr.1: 24120718-24120718 on Build GRCh38
rs780628736 Chr.1: 24120621-24120621 on Build GRCh38
rs772391534 Chr.1: 24120846-24120846 on Build GRCh38
rs763566860 Chr.1: 24120938-24120938 on Build GRCh38
rs765555847 Chr.1: 24120873-24120873 on Build GRCh38
rs781675220 Chr.1: 24120500-24120500 on Build GRCh38
rs183784131 Chr.1: 24120755-24120755 on Build GRCh38
rs781261671 Chr.1: 24120779-24120779 on Build GRCh38
rs1009396983 Chr.1: 24120618-24120618 on Build GRCh38
rs761916023 Chr.1: 24120872-24120872 on Build GRCh38
rs367753377 Chr.1: 24120865-24120865 on Build GRCh38
rs3795302 Chr.1: 24120626-24120626 on Build GRCh38
rs754861443 Chr.1: 24120776-24120776 on Build GRCh38
rs996784090 Chr.1: 24120782-24120782 on Build GRCh38
rs561037617 Chr.1: 24120710-24120710 on Build GRCh38
rs886560233 Chr.1: 24120712-24120712 on Build GRCh38
rs573703892 Chr.1: 24120753-24120753 on Build GRCh38
rs766053633 Chr.1: 24120763-24120763 on Build GRCh38
rs146841813 Chr.1: 24120900-24120900 on Build GRCh38
rs564402425 Chr.1: 24120709-24120709 on Build GRCh38
rs776153907 Chr.1: 24120847-24120847 on Build GRCh38
rs371277731 Chr.1: 24120941-24120941 on Build GRCh38
rs189208294 Chr.1: 24120503-24120503 on Build GRCh38
rs746309819 Chr.1: 24120845-24120845 on Build GRCh38
rs779131004 Chr.1: 24120842-24120842 on Build GRCh38
rs191254729 Chr.1: 24120695-24120695 on Build GRCh38
rs565870752 Chr.1: 24120693-24120693 on Build GRCh38
rs746389103 Chr.1: 24120585-24120585 on Build GRCh38
rs867579021 Chr.1: 24120769-24120769 on Build GRCh38
rs148823055 Chr.1: 24120501-24120501 on Build GRCh38
rs571079976 Chr.1: 24120519-24120519 on Build GRCh38
rs955260999 Chr.1: 24120831-24120831 on Build GRCh38
rs535928842 Chr.1: 24120629-24120629 on Build GRCh38
rs759383447 Chr.1: 24120892-24120892 on Build GRCh38
rs768884900 Chr.1: 24120867-24120867 on Build GRCh38
rs909932565 Chr.1: 24120813-24120813 on Build GRCh38
rs3795301 Chr.1: 24120630-24120630 on Build GRCh38
rs531628197 Chr.1: 24120565-24120565 on Build GRCh38
rs938271519 Chr.1: 24120680-24120680 on Build GRCh38
rs148843028 Chr.1: 24120826-24120826 on Build GRCh38
rs199519515 Chr.1: 24120898-24120898 on Build GRCh38
rs755584982 Chr.1: 24120934-24120934 on Build GRCh38
rs3795300 Chr.1: 24120724-24120724 on Build GRCh38
rs996728394 Chr.1: 24120597-24120597 on Build GRCh38
rs917936067 Chr.1: 24120694-24120694 on Build GRCh38
rs986446961 Chr.1: 24120616-24120616 on Build GRCh38
rs374269399 Chr.1: 24120818-24120818 on Build GRCh38
rs575411350 Chr.1: 24120599-24120599 on Build GRCh38
rs774015789 Chr.1: 24120882-24120882 on Build GRCh38
rs145274429 Chr.1: 24120738-24120738 on Build GRCh38
rs751421169 Chr.1: 24120772-24120772 on Build GRCh38
rs550113235 Chr.1: 24120606-24120606 on Build GRCh38
rs951523665 Chr.1: 24120773-24120773 on Build GRCh38
rs1051483888 Chr.1: 24120568-24120568 on Build GRCh38
rs748971813 Chr.1: 24120823-24120823 on Build GRCh38
rs752657948 Chr.1: 24120913-24120913 on Build GRCh38
rs752140778 Chr.1: 24120791-24120791 on Build GRCh38
rs753398266 Chr.1: 24120943-24120943 on Build GRCh38
rs1047781986 Chr.1: 24120512-24120512 on Build GRCh38

View Full Product Details