Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 73683949-73684450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD276
Entrez Gene ID
80381
Gene Name
CD276 molecule
Gene Aliases
4Ig-B7-H3, B7-H3, B7H3, B7RP-2
UniGene
Hs.744915
Gene Chromosome Location
Chr.15: 73683966-73714518 on Build GRCh38
Transcripts
NM_001024736.1 NM_025240.2 XM_017022638.1

SNP Detail

SNP ID Chromosome Location
rs527672813 Chr.15: 73684280-73684280 on Build GRCh38
rs947473973 Chr.15: 73684383-73684383 on Build GRCh38
rs889788826 Chr.15: 73684409-73684409 on Build GRCh38
rs886598867 Chr.15: 73684329-73684329 on Build GRCh38
rs924909472 Chr.15: 73684196-73684196 on Build GRCh38
rs1025825043 Chr.15: 73684250-73684250 on Build GRCh38
rs988630165 Chr.15: 73684403-73684403 on Build GRCh38
rs969077442 Chr.15: 73684336-73684336 on Build GRCh38
rs200819633 Chr.15: 73683999-73684001 on Build GRCh38
rs1043114119 Chr.15: 73684400-73684400 on Build GRCh38
rs926933841 Chr.15: 73684418-73684418 on Build GRCh38
rs935246280 Chr.15: 73684367-73684367 on Build GRCh38
rs192679813 Chr.15: 73684093-73684093 on Build GRCh38
rs994709264 Chr.15: 73684236-73684236 on Build GRCh38
rs144414408 Chr.15: 73684000-73684003 on Build GRCh38
rs1028931951 Chr.15: 73684360-73684360 on Build GRCh38
rs892784318 Chr.15: 73684092-73684092 on Build GRCh38
rs576066682 Chr.15: 73684094-73684094 on Build GRCh38
rs912413529 Chr.15: 73684077-73684077 on Build GRCh38
rs952502207 Chr.15: 73683977-73683977 on Build GRCh38
rs1041017768 Chr.15: 73684419-73684419 on Build GRCh38
rs1023709755 Chr.15: 73684299-73684299 on Build GRCh38
rs1046376238 Chr.15: 73684088-73684088 on Build GRCh38
rs899384292 Chr.15: 73684344-73684344 on Build GRCh38
rs891842747 Chr.15: 73684100-73684100 on Build GRCh38
rs1016851794 Chr.15: 73684337-73684338 on Build GRCh38
rs927828454 Chr.15: 73684365-73684365 on Build GRCh38
rs1016550361 Chr.15: 73684131-73684131 on Build GRCh38
rs905547347 Chr.15: 73684300-73684300 on Build GRCh38
rs927775914 Chr.15: 73684091-73684091 on Build GRCh38
rs916266892 Chr.15: 73683993-73683993 on Build GRCh38
rs934543998 Chr.15: 73684242-73684242 on Build GRCh38
rs901434539 Chr.15: 73684233-73684233 on Build GRCh38
rs983911151 Chr.15: 73684295-73684295 on Build GRCh38
rs982024228 Chr.15: 73684343-73684343 on Build GRCh38
rs921948906 Chr.15: 73684206-73684206 on Build GRCh38
rs370997437 Chr.15: 73684151-73684151 on Build GRCh38
rs938362510 Chr.15: 73684430-73684430 on Build GRCh38
rs901462307 Chr.15: 73684428-73684428 on Build GRCh38
rs905579952 Chr.15: 73684104-73684104 on Build GRCh38
rs1002638334 Chr.15: 73684108-73684108 on Build GRCh38
rs984164340 Chr.15: 73683988-73683988 on Build GRCh38
rs1040692733 Chr.15: 73684207-73684207 on Build GRCh38
rs1019861374 Chr.15: 73684220-73684220 on Build GRCh38
rs988354075 Chr.15: 73684369-73684369 on Build GRCh38
rs543448774 Chr.15: 73684149-73684149 on Build GRCh38
rs934552878 Chr.15: 73684077-73684077 on Build GRCh38
rs1050854946 Chr.15: 73684040-73684040 on Build GRCh38
rs368183381 Chr.15: 73683990-73683990 on Build GRCh38
rs1042387517 Chr.15: 73684191-73684191 on Build GRCh38
rs943671112 Chr.15: 73684201-73684201 on Build GRCh38
rs996402312 Chr.15: 73684349-73684349 on Build GRCh38
rs947302491 Chr.15: 73684282-73684282 on Build GRCh38
rs915059005 Chr.15: 73684371-73684371 on Build GRCh38
rs947934664 Chr.15: 73684078-73684078 on Build GRCh38
rs889811659 Chr.15: 73684130-73684130 on Build GRCh38
rs914405266 Chr.15: 73684407-73684407 on Build GRCh38
rs1031123646 Chr.15: 73684379-73684379 on Build GRCh38
rs1043430518 Chr.15: 73684122-73684122 on Build GRCh38
rs942744963 Chr.15: 73684410-73684410 on Build GRCh38
rs1011677117 Chr.15: 73684120-73684120 on Build GRCh38
rs1056932329 Chr.15: 73684320-73684320 on Build GRCh38
rs933415575 Chr.15: 73684018-73684018 on Build GRCh38
rs1040320351 Chr.15: 73684186-73684201 on Build GRCh38
rs564776799 Chr.15: 73684209-73684209 on Build GRCh38
rs1002605624 Chr.15: 73684308-73684308 on Build GRCh38

View Full Product Details