Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 73708255-73708755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD276
Entrez Gene ID
80381
Gene Name
CD276 molecule
Gene Aliases
4Ig-B7-H3, B7-H3, B7H3, B7RP-2
UniGene
Hs.744915
Gene Chromosome Location
Chr.15: 73683966-73714518 on Build GRCh38
Transcripts
NM_001024736.1 NM_025240.2 XM_005254700.4 XM_011522095.2 XM_011522096.2 XM_017022638.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
138066 c.1409C>T p.T470I Chr.15: 73708378-73708378 on Build GRCh38
4805866 c.1416G>C p.G472G Chr.15: 73708385-73708385 on Build GRCh38
5677166 c.1395G>T p.E465D Chr.15: 73708364-73708364 on Build GRCh38
6555067 c.1459G>T p.V487L Chr.15: 73708428-73708428 on Build GRCh38
6354118 c.1419G>C p.L473L Chr.15: 73708388-73708388 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112297497 Chr.15: 73708340-73708340 on Build GRCh38
rs758852090 Chr.15: 73708288-73708288 on Build GRCh38
rs751929224 Chr.15: 73708551-73708552 on Build GRCh38
rs1049455419 Chr.15: 73708723-73708723 on Build GRCh38
rs559363212 Chr.15: 73708298-73708298 on Build GRCh38
rs751223215 Chr.15: 73708478-73708483 on Build GRCh38
rs200206394 Chr.15: 73708374-73708374 on Build GRCh38
rs775920988 Chr.15: 73708386-73708389 on Build GRCh38
rs939695831 Chr.15: 73708635-73708638 on Build GRCh38
rs992102658 Chr.15: 73708619-73708619 on Build GRCh38
rs117028762 Chr.15: 73708563-73708563 on Build GRCh38
rs754631520 Chr.15: 73708483-73708488 on Build GRCh38
rs145286416 Chr.15: 73708478-73708481 on Build GRCh38
rs556281946 Chr.15: 73708458-73708458 on Build GRCh38
rs1043543524 Chr.15: 73708489-73708490 on Build GRCh38
rs747485367 Chr.15: 73708293-73708293 on Build GRCh38
rs746846091 Chr.15: 73708423-73708423 on Build GRCh38
rs765864478 Chr.15: 73708478-73708479 on Build GRCh38
rs779701778 Chr.15: 73708627-73708627 on Build GRCh38
rs769843380 Chr.15: 73708460-73708460 on Build GRCh38
rs748470997 Chr.15: 73708630-73708630 on Build GRCh38
rs368212813 Chr.15: 73708385-73708385 on Build GRCh38
rs1017600653 Chr.15: 73708621-73708621 on Build GRCh38
rs908963741 Chr.15: 73708652-73708652 on Build GRCh38
rs766351492 Chr.15: 73708360-73708360 on Build GRCh38
rs577696420 Chr.15: 73708477-73708477 on Build GRCh38
rs762246981 Chr.15: 73708445-73708448 on Build GRCh38
rs912597069 Chr.15: 73708433-73708433 on Build GRCh38
rs370168964 Chr.15: 73708491-73708491 on Build GRCh38
rs747884113 Chr.15: 73708339-73708339 on Build GRCh38
rs984574171 Chr.15: 73708402-73708402 on Build GRCh38
rs148146340 Chr.15: 73708502-73708502 on Build GRCh38
rs1008461727 Chr.15: 73708572-73708572 on Build GRCh38
rs375564059 Chr.15: 73708427-73708427 on Build GRCh38
rs950924317 Chr.15: 73708638-73708638 on Build GRCh38
rs1037208582 Chr.15: 73708512-73708512 on Build GRCh38
rs763087764 Chr.15: 73708376-73708376 on Build GRCh38
rs566637289 Chr.15: 73708364-73708364 on Build GRCh38
rs757986559 Chr.15: 73708578-73708578 on Build GRCh38
rs750359383 Chr.15: 73708391-73708392 on Build GRCh38
rs968789514 Chr.15: 73708596-73708596 on Build GRCh38
rs749936663 Chr.15: 73708569-73708569 on Build GRCh38
rs1019845845 Chr.15: 73708577-73708578 on Build GRCh38
rs903903143 Chr.15: 73708576-73708576 on Build GRCh38
rs931105210 Chr.15: 73708697-73708697 on Build GRCh38
rs371789053 Chr.15: 73708365-73708365 on Build GRCh38
rs995639564 Chr.15: 73708549-73708550 on Build GRCh38
rs762987344 Chr.15: 73708520-73708520 on Build GRCh38
rs752489974 Chr.15: 73708501-73708504 on Build GRCh38
rs2291010 Chr.15: 73708631-73708631 on Build GRCh38
rs781775734 Chr.15: 73708387-73708387 on Build GRCh38
rs1007519512 Chr.15: 73708611-73708611 on Build GRCh38
rs2291012 Chr.15: 73708388-73708388 on Build GRCh38
rs937815133 Chr.15: 73708613-73708613 on Build GRCh38
rs928313172 Chr.15: 73708696-73708696 on Build GRCh38
rs770846226 Chr.15: 73708430-73708430 on Build GRCh38
rs765671852 Chr.15: 73708450-73708451 on Build GRCh38
rs772709455 Chr.15: 73708458-73708460 on Build GRCh38
rs375502531 Chr.15: 73708497-73708497 on Build GRCh38
rs756695894 Chr.15: 73708489-73708489 on Build GRCh38
rs926479408 Chr.15: 73708610-73708613 on Build GRCh38
rs776603714 Chr.15: 73708434-73708434 on Build GRCh38
rs1015331914 Chr.15: 73708361-73708361 on Build GRCh38
rs201611942 Chr.15: 73708610-73708611 on Build GRCh38
rs369418465 Chr.15: 73708428-73708428 on Build GRCh38
rs777601806 Chr.15: 73708419-73708419 on Build GRCh38
rs950927815 Chr.15: 73708392-73708392 on Build GRCh38
rs149679033 Chr.15: 73708406-73708406 on Build GRCh38
rs756469060 Chr.15: 73708510-73708510 on Build GRCh38
rs752197016 Chr.15: 73708495-73708495 on Build GRCh38
rs764275656 Chr.15: 73708468-73708468 on Build GRCh38
rs775466357 Chr.15: 73708464-73708464 on Build GRCh38
rs745763153 Chr.15: 73708503-73708503 on Build GRCh38
rs568647279 Chr.15: 73708385-73708386 on Build GRCh38
rs769645020 Chr.15: 73708505-73708505 on Build GRCh38
rs748860461 Chr.15: 73708335-73708335 on Build GRCh38
rs765428112 Chr.15: 73708369-73708369 on Build GRCh38
rs979687145 Chr.15: 73708287-73708287 on Build GRCh38
rs778301688 Chr.15: 73708401-73708401 on Build GRCh38
rs775621253 Chr.15: 73708515-73708515 on Build GRCh38
rs979840035 Chr.15: 73708688-73708688 on Build GRCh38
rs772909584 Chr.15: 73708343-73708343 on Build GRCh38
rs760555580 Chr.15: 73708348-73708348 on Build GRCh38
rs767250503 Chr.15: 73708485-73708488 on Build GRCh38
rs954203126 Chr.15: 73708555-73708555 on Build GRCh38
rs371940763 Chr.15: 73708410-73708410 on Build GRCh38
rs774121373 Chr.15: 73708338-73708338 on Build GRCh38
rs532673793 Chr.15: 73708651-73708651 on Build GRCh38
rs751945352 Chr.15: 73708476-73708476 on Build GRCh38
rs966012163 Chr.15: 73708623-73708623 on Build GRCh38
rs768922776 Chr.15: 73708523-73708523 on Build GRCh38
rs757873058 Chr.15: 73708297-73708297 on Build GRCh38
rs200433977 Chr.15: 73708579-73708580 on Build GRCh38
rs2291011 Chr.15: 73708498-73708498 on Build GRCh38
rs545258004 Chr.15: 73708480-73708480 on Build GRCh38
rs779665599 Chr.15: 73708306-73708306 on Build GRCh38
rs531589982 Chr.15: 73708543-73708543 on Build GRCh38
rs566680275 Chr.15: 73708699-73708699 on Build GRCh38
rs368475211 Chr.15: 73708336-73708336 on Build GRCh38
rs908925461 Chr.15: 73708403-73708403 on Build GRCh38
rs768941439 Chr.15: 73708387-73708388 on Build GRCh38
rs763188455 Chr.15: 73708465-73708465 on Build GRCh38
rs746788918 Chr.15: 73708499-73708499 on Build GRCh38
rs772426782 Chr.15: 73708360-73708361 on Build GRCh38
rs372288103 Chr.15: 73708488-73708488 on Build GRCh38
rs551253248 Chr.15: 73708658-73708658 on Build GRCh38
rs754594435 Chr.15: 73708389-73708389 on Build GRCh38
rs542917505 Chr.15: 73708501-73708501 on Build GRCh38
rs150498569 Chr.15: 73708380-73708380 on Build GRCh38
rs546317569 Chr.15: 73708486-73708486 on Build GRCh38
rs779154867 Chr.15: 73708342-73708342 on Build GRCh38
rs1022133740 Chr.15: 73708585-73708585 on Build GRCh38
rs374313192 Chr.15: 73708379-73708379 on Build GRCh38
rs745883489 Chr.15: 73708354-73708355 on Build GRCh38

View Full Product Details