Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 73709451-73709954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD276
Entrez Gene ID
80381
Gene Name
CD276 molecule
Gene Aliases
4Ig-B7-H3, B7-H3, B7H3, B7RP-2
UniGene
Hs.744915
Gene Chromosome Location
Chr.15: 73683966-73714518 on Build GRCh38
Transcripts
NM_001024736.1 NM_025240.2 XM_005254700.4 XM_011522095.2 XM_011522096.2 XM_017022638.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5849735 c.1513G>A p.D505N Chr.15: 73709656-73709656 on Build GRCh38
6350631 c.1522G>A p.G508R Chr.15: 73709665-73709665 on Build GRCh38
6350451 c.1509T>C p.A503A Chr.15: 73709652-73709652 on Build GRCh38
6464064 c.1507G>A p.A503T Chr.15: 73709650-73709650 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752246825 Chr.15: 73709658-73709658 on Build GRCh38
rs1037976902 Chr.15: 73709695-73709695 on Build GRCh38
rs545704567 Chr.15: 73709653-73709653 on Build GRCh38
rs942541527 Chr.15: 73709528-73709528 on Build GRCh38
rs536427912 Chr.15: 73709557-73709557 on Build GRCh38
rs766833906 Chr.15: 73709656-73709656 on Build GRCh38
rs777786710 Chr.15: 73709623-73709625 on Build GRCh38
rs751089402 Chr.15: 73709726-73709726 on Build GRCh38
rs768238926 Chr.15: 73709710-73709710 on Build GRCh38
rs577178317 Chr.15: 73709689-73709689 on Build GRCh38
rs965402668 Chr.15: 73709568-73709568 on Build GRCh38
rs555106059 Chr.15: 73709783-73709783 on Build GRCh38
rs1018127144 Chr.15: 73709917-73709917 on Build GRCh38
rs796736296 Chr.15: 73709550-73709550 on Build GRCh38
rs757933401 Chr.15: 73709752-73709752 on Build GRCh38
rs773678010 Chr.15: 73709701-73709701 on Build GRCh38
rs779923784 Chr.15: 73709678-73709678 on Build GRCh38
rs968800374 Chr.15: 73709935-73709935 on Build GRCh38
rs144368803 Chr.15: 73709671-73709671 on Build GRCh38
rs767198242 Chr.15: 73709733-73709733 on Build GRCh38
rs867297222 Chr.15: 73709681-73709681 on Build GRCh38
rs765097230 Chr.15: 73709716-73709716 on Build GRCh38
rs774771850 Chr.15: 73709598-73709598 on Build GRCh38
rs199731294 Chr.15: 73709662-73709662 on Build GRCh38
rs377107054 Chr.15: 73709666-73709666 on Build GRCh38
rs774579719 Chr.15: 73709691-73709691 on Build GRCh38
rs1026812882 Chr.15: 73709851-73709851 on Build GRCh38
rs558629661 Chr.15: 73709670-73709670 on Build GRCh38
rs371900951 Chr.15: 73709611-73709611 on Build GRCh38
rs766892381 Chr.15: 73709705-73709705 on Build GRCh38
rs11552770 Chr.15: 73709652-73709652 on Build GRCh38
rs911116444 Chr.15: 73709511-73709511 on Build GRCh38
rs994017690 Chr.15: 73709770-73709770 on Build GRCh38
rs549343866 Chr.15: 73709510-73709510 on Build GRCh38
rs748343221 Chr.15: 73709693-73709693 on Build GRCh38
rs976553112 Chr.15: 73709577-73709577 on Build GRCh38
rs767928742 Chr.15: 73709643-73709643 on Build GRCh38
rs530243951 Chr.15: 73709810-73709810 on Build GRCh38
rs542810750 Chr.15: 73709811-73709811 on Build GRCh38
rs910920019 Chr.15: 73709564-73709564 on Build GRCh38
rs906074232 Chr.15: 73709713-73709713 on Build GRCh38
rs3825859 Chr.15: 73709665-73709665 on Build GRCh38
rs897870523 Chr.15: 73709769-73709769 on Build GRCh38
rs1020017751 Chr.15: 73709896-73709896 on Build GRCh38
rs538315026 Chr.15: 73709498-73709498 on Build GRCh38
rs888269931 Chr.15: 73709878-73709878 on Build GRCh38
rs561088750 Chr.15: 73709816-73709816 on Build GRCh38
rs772469627 Chr.15: 73709698-73709698 on Build GRCh38
rs952589696 Chr.15: 73709543-73709543 on Build GRCh38
rs749377613 Chr.15: 73709687-73709687 on Build GRCh38
rs561091827 Chr.15: 73709799-73709799 on Build GRCh38
rs201294636 Chr.15: 73709737-73709737 on Build GRCh38
rs776945716 Chr.15: 73709711-73709711 on Build GRCh38
rs895982265 Chr.15: 73709861-73709861 on Build GRCh38
rs112368076 Chr.15: 73709778-73709778 on Build GRCh38
rs761008337 Chr.15: 73709704-73709704 on Build GRCh38
rs756135849 Chr.15: 73709622-73709622 on Build GRCh38
rs572093268 Chr.15: 73709514-73709514 on Build GRCh38
rs749883476 Chr.15: 73709723-73709723 on Build GRCh38
rs780918414 Chr.15: 73709669-73709669 on Build GRCh38
rs760092128 Chr.15: 73709714-73709714 on Build GRCh38
rs976522996 Chr.15: 73709569-73709569 on Build GRCh38
rs1010424128 Chr.15: 73709881-73709881 on Build GRCh38

View Full Product Details