Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74619921-74620429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLK3
Entrez Gene ID
1198
Gene Name
CDC like kinase 3
Gene Aliases
PHCLK3, PHCLK3/152
UniGene
Hs.511790
Gene Chromosome Location
Chr.15: 74598476-74645418 on Build GRCh38
Transcripts
NM_001130028.1 NM_003992.4 XM_005254151.3 XM_011521205.2 XM_011521206.2 XM_011521209.2 XM_017021905.1 XM_017021906.1 XM_017021907.1 XM_017021908.1 XM_017021909.1 XM_017021910.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1189064 c.269G>C p.R90P Chr.15: 74620125-74620125 on Build GRCh38
6630494 c.807C>A p.A269A Chr.15: 74620219-74620219 on Build GRCh38
416816 c.735G>A p.E245E Chr.15: 74620147-74620147 on Build GRCh38
6630493 c.369G>A p.S123S Chr.15: 74620225-74620225 on Build GRCh38
4844086 c.248A>G p.Y83C Chr.15: 74620104-74620104 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778392975 Chr.15: 74620193-74620193 on Build GRCh38
rs778298698 Chr.15: 74620156-74620156 on Build GRCh38
rs116177097 Chr.15: 74620344-74620344 on Build GRCh38
rs141991982 Chr.15: 74620062-74620062 on Build GRCh38
rs775615815 Chr.15: 74620229-74620230 on Build GRCh38
rs1013836659 Chr.15: 74620281-74620281 on Build GRCh38
rs146248770 Chr.15: 74620213-74620213 on Build GRCh38
rs765392463 Chr.15: 74620144-74620144 on Build GRCh38
rs11543197 Chr.15: 74620198-74620198 on Build GRCh38
rs748355287 Chr.15: 74620073-74620073 on Build GRCh38
rs765409260 Chr.15: 74620229-74620229 on Build GRCh38
rs756011168 Chr.15: 74620068-74620068 on Build GRCh38
rs1018098172 Chr.15: 74620360-74620360 on Build GRCh38
rs936846480 Chr.15: 74620237-74620237 on Build GRCh38
rs895364263 Chr.15: 74620107-74620107 on Build GRCh38
rs923292759 Chr.15: 74620395-74620395 on Build GRCh38
rs952400565 Chr.15: 74620334-74620334 on Build GRCh38
rs753905373 Chr.15: 74620071-74620071 on Build GRCh38
rs371347609 Chr.15: 74620009-74620009 on Build GRCh38
rs762665220 Chr.15: 74619996-74619996 on Build GRCh38
rs371419854 Chr.15: 74620026-74620026 on Build GRCh38
rs759672533 Chr.15: 74620101-74620103 on Build GRCh38
rs919745120 Chr.15: 74620313-74620313 on Build GRCh38
rs141693165 Chr.15: 74620194-74620194 on Build GRCh38
rs752348186 Chr.15: 74619995-74619995 on Build GRCh38
rs755185690 Chr.15: 74620072-74620072 on Build GRCh38
rs760558107 Chr.15: 74620209-74620209 on Build GRCh38
rs202095468 Chr.15: 74620155-74620155 on Build GRCh38
rs374366533 Chr.15: 74620233-74620233 on Build GRCh38
rs943934447 Chr.15: 74620177-74620177 on Build GRCh38
rs777526433 Chr.15: 74620304-74620304 on Build GRCh38
rs377701238 Chr.15: 74620268-74620268 on Build GRCh38
rs760914890 Chr.15: 74620284-74620285 on Build GRCh38
rs369940204 Chr.15: 74620055-74620055 on Build GRCh38
rs763294328 Chr.15: 74620253-74620253 on Build GRCh38
rs773131274 Chr.15: 74619966-74619967 on Build GRCh38
rs183557935 Chr.15: 74620174-74620174 on Build GRCh38
rs546267843 Chr.15: 74620180-74620180 on Build GRCh38
rs768214567 Chr.15: 74620182-74620182 on Build GRCh38
rs976675886 Chr.15: 74620390-74620390 on Build GRCh38
rs774040682 Chr.15: 74620118-74620118 on Build GRCh38
rs955519680 Chr.15: 74620224-74620224 on Build GRCh38
rs768865287 Chr.15: 74620040-74620040 on Build GRCh38
rs771661811 Chr.15: 74620196-74620196 on Build GRCh38
rs764392396 Chr.15: 74620287-74620287 on Build GRCh38
rs773020998 Chr.15: 74620197-74620197 on Build GRCh38
rs139627456 Chr.15: 74620143-74620143 on Build GRCh38
rs377532088 Chr.15: 74620115-74620115 on Build GRCh38
rs367609434 Chr.15: 74620045-74620045 on Build GRCh38
rs776219419 Chr.15: 74620309-74620309 on Build GRCh38
rs780907715 Chr.15: 74620017-74620017 on Build GRCh38
rs747353105 Chr.15: 74620160-74620160 on Build GRCh38
rs760551120 Chr.15: 74620133-74620133 on Build GRCh38
rs768019957 Chr.15: 74620110-74620110 on Build GRCh38
rs748795519 Chr.15: 74620103-74620103 on Build GRCh38
rs759152978 Chr.15: 74619986-74619986 on Build GRCh38
rs896155561 Chr.15: 74620364-74620364 on Build GRCh38
rs975358822 Chr.15: 74620034-74620034 on Build GRCh38
rs751007122 Chr.15: 74620279-74620279 on Build GRCh38
rs986745385 Chr.15: 74620247-74620247 on Build GRCh38
rs139058373 Chr.15: 74620324-74620324 on Build GRCh38
rs767369515 Chr.15: 74620124-74620124 on Build GRCh38
rs775001314 Chr.15: 74620049-74620049 on Build GRCh38
rs776684802 Chr.15: 74620216-74620216 on Build GRCh38
rs146399462 Chr.15: 74620047-74620047 on Build GRCh38
rs191789873 Chr.15: 74620388-74620388 on Build GRCh38
rs1033374096 Chr.15: 74620106-74620106 on Build GRCh38
rs772169718 Chr.15: 74620126-74620128 on Build GRCh38
rs757740424 Chr.15: 74620166-74620166 on Build GRCh38
rs761116304 Chr.15: 74620287-74620287 on Build GRCh38
rs747525547 Chr.15: 74620083-74620083 on Build GRCh38
rs764374883 Chr.15: 74620260-74620260 on Build GRCh38
rs779166711 Chr.15: 74620041-74620041 on Build GRCh38
rs752769035 Chr.15: 74620250-74620250 on Build GRCh38
rs371659907 Chr.15: 74620046-74620046 on Build GRCh38
rs769258296 Chr.15: 74620048-74620048 on Build GRCh38
rs911145093 Chr.15: 74620169-74620169 on Build GRCh38
rs766299793 Chr.15: 74620134-74620134 on Build GRCh38
rs937777336 Chr.15: 74620340-74620340 on Build GRCh38
rs750421202 Chr.15: 74620067-74620067 on Build GRCh38
rs1042570674 Chr.15: 74620088-74620088 on Build GRCh38
rs770822743 Chr.15: 74620308-74620308 on Build GRCh38
rs761636972 Chr.15: 74620061-74620061 on Build GRCh38
rs886951541 Chr.15: 74620051-74620051 on Build GRCh38
rs781398684 Chr.15: 74620277-74620277 on Build GRCh38
rs753032664 Chr.15: 74620146-74620146 on Build GRCh38
rs749474059 Chr.15: 74620030-74620030 on Build GRCh38
rs753682825 Chr.15: 74620142-74620142 on Build GRCh38
rs757212136 Chr.15: 74620016-74620016 on Build GRCh38
rs751301893 Chr.15: 74620012-74620012 on Build GRCh38
rs11543198 Chr.15: 74619987-74619987 on Build GRCh38
rs781743086 Chr.15: 74620173-74620173 on Build GRCh38
rs772738675 Chr.15: 74620125-74620125 on Build GRCh38
rs148465298 Chr.15: 74620217-74620217 on Build GRCh38
rs146329671 Chr.15: 74620080-74620080 on Build GRCh38
rs796316616 Chr.15: 74620161-74620161 on Build GRCh38
rs998948391 Chr.15: 74620201-74620201 on Build GRCh38
rs147060953 Chr.15: 74620005-74620006 on Build GRCh38
rs373663118 Chr.15: 74620027-74620027 on Build GRCh38
rs1024938106 Chr.15: 74620179-74620179 on Build GRCh38
rs759383924 Chr.15: 74620222-74620222 on Build GRCh38
rs573934140 Chr.15: 74619964-74619964 on Build GRCh38
rs754199835 Chr.15: 74620304-74620304 on Build GRCh38
rs188194375 Chr.15: 74620057-74620057 on Build GRCh38
rs763804384 Chr.15: 74619999-74619999 on Build GRCh38
rs939816913 Chr.15: 74620058-74620058 on Build GRCh38
rs772657206 Chr.15: 74619961-74619961 on Build GRCh38
rs201552338 Chr.15: 74620131-74620131 on Build GRCh38
rs371973085 Chr.15: 74620079-74620079 on Build GRCh38
rs761442417 Chr.15: 74620119-74620119 on Build GRCh38
rs771539104 Chr.15: 74620296-74620296 on Build GRCh38
rs376812584 Chr.15: 74620097-74620097 on Build GRCh38
rs527476353 Chr.15: 74620306-74620306 on Build GRCh38
rs766382295 Chr.15: 74620069-74620069 on Build GRCh38
rs530100240 Chr.15: 74619952-74619952 on Build GRCh38
rs375125545 Chr.15: 74620050-74620050 on Build GRCh38
rs762423353 Chr.15: 74619960-74619960 on Build GRCh38
rs756548398 Chr.15: 74620285-74620285 on Build GRCh38
rs1033073473 Chr.15: 74620205-74620205 on Build GRCh38
rs1013633913 Chr.15: 74620374-74620374 on Build GRCh38

View Full Product Details