Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74622409-74622913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLK3
Entrez Gene ID
1198
Gene Name
CDC like kinase 3
Gene Aliases
PHCLK3, PHCLK3/152
UniGene
Hs.511790
Gene Chromosome Location
Chr.15: 74598476-74645418 on Build GRCh38
Transcripts
NM_001130028.1 NM_003992.4 XM_005254151.3 XM_011521205.2 XM_011521206.2 XM_011521209.2 XM_017021905.1 XM_017021906.1 XM_017021907.1 XM_017021908.1 XM_017021909.1 XM_017021910.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4568130 c.488_489GT>AA p.G163E Chr.15: 74622515-74622516 on Build GRCh38
4568129 c.932_933GT>AA p.G311E Chr.15: 74622515-74622516 on Build GRCh38
3706731 c.911-4C>T p.? Chr.15: 74622490-74622490 on Build GRCh38
4450152 c.467-1G>A p.? Chr.15: 74622493-74622493 on Build GRCh38
434295 c.913G>A p.E305K Chr.15: 74622496-74622496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772109926 Chr.15: 74622558-74622558 on Build GRCh38
rs754600889 Chr.15: 74622448-74622448 on Build GRCh38
rs1022620508 Chr.15: 74622674-74622674 on Build GRCh38
rs932756155 Chr.15: 74622807-74622807 on Build GRCh38
rs566723035 Chr.15: 74622815-74622819 on Build GRCh38
rs770174422 Chr.15: 74622576-74622576 on Build GRCh38
rs927990343 Chr.15: 74622623-74622623 on Build GRCh38
rs756767880 Chr.15: 74622473-74622473 on Build GRCh38
rs145279134 Chr.15: 74622843-74622843 on Build GRCh38
rs577814747 Chr.15: 74622605-74622605 on Build GRCh38
rs748479659 Chr.15: 74622811-74622811 on Build GRCh38
rs1050303284 Chr.15: 74622684-74622684 on Build GRCh38
rs909108016 Chr.15: 74622470-74622470 on Build GRCh38
rs934830469 Chr.15: 74622819-74622819 on Build GRCh38
rs749245187 Chr.15: 74622443-74622443 on Build GRCh38
rs970250533 Chr.15: 74622679-74622679 on Build GRCh38
rs1011664468 Chr.15: 74622666-74622666 on Build GRCh38
rs753217814 Chr.15: 74622468-74622468 on Build GRCh38
rs921258213 Chr.15: 74622785-74622785 on Build GRCh38
rs777018491 Chr.15: 74622735-74622735 on Build GRCh38
rs141358186 Chr.15: 74622767-74622767 on Build GRCh38
rs946617425 Chr.15: 74622683-74622683 on Build GRCh38
rs962009942 Chr.15: 74622474-74622474 on Build GRCh38
rs147812882 Chr.15: 74622511-74622511 on Build GRCh38
rs982602578 Chr.15: 74622612-74622612 on Build GRCh38
rs113528781 Chr.15: 74622648-74622648 on Build GRCh38
rs755764514 Chr.15: 74622484-74622484 on Build GRCh38
rs769942112 Chr.15: 74622814-74622814 on Build GRCh38
rs780011029 Chr.15: 74622487-74622487 on Build GRCh38
rs181805358 Chr.15: 74622567-74622567 on Build GRCh38
rs778761547 Chr.15: 74622540-74622540 on Build GRCh38
rs376490476 Chr.15: 74622538-74622538 on Build GRCh38
rs561460200 Chr.15: 74622783-74622783 on Build GRCh38
rs760108594 Chr.15: 74622463-74622463 on Build GRCh38
rs760882563 Chr.15: 74622454-74622454 on Build GRCh38
rs897509428 Chr.15: 74622790-74622790 on Build GRCh38
rs772165651 Chr.15: 74622451-74622451 on Build GRCh38
rs776921104 Chr.15: 74622460-74622460 on Build GRCh38
rs139458591 Chr.15: 74622552-74622552 on Build GRCh38
rs200189676 Chr.15: 74622476-74622476 on Build GRCh38
rs773424090 Chr.15: 74622846-74622846 on Build GRCh38
rs1038339878 Chr.15: 74622502-74622502 on Build GRCh38
rs916724820 Chr.15: 74622583-74622583 on Build GRCh38
rs750170722 Chr.15: 74622480-74622480 on Build GRCh38
rs969922287 Chr.15: 74622873-74622873 on Build GRCh38
rs1008685233 Chr.15: 74622765-74622765 on Build GRCh38
rs747244121 Chr.15: 74622566-74622566 on Build GRCh38
rs749898606 Chr.15: 74622599-74622599 on Build GRCh38
rs774518985 Chr.15: 74622448-74622448 on Build GRCh38
rs777762585 Chr.15: 74622563-74622563 on Build GRCh38
rs751192539 Chr.15: 74622447-74622448 on Build GRCh38
rs1023845863 Chr.15: 74622867-74622867 on Build GRCh38
rs963284074 Chr.15: 74622762-74622762 on Build GRCh38
rs1035821497 Chr.15: 74622695-74622695 on Build GRCh38
rs950551372 Chr.15: 74622703-74622703 on Build GRCh38
rs766734003 Chr.15: 74622457-74622457 on Build GRCh38
rs560181108 Chr.15: 74622652-74622652 on Build GRCh38
rs893699574 Chr.15: 74622661-74622661 on Build GRCh38
rs527329430 Chr.15: 74622694-74622694 on Build GRCh38
rs983246017 Chr.15: 74622722-74622722 on Build GRCh38
rs77232571 Chr.15: 74622634-74622634 on Build GRCh38
rs763516693 Chr.15: 74622591-74622591 on Build GRCh38
rs777013007 Chr.15: 74622573-74622573 on Build GRCh38
rs765624153 Chr.15: 74622467-74622467 on Build GRCh38
rs1052314990 Chr.15: 74622643-74622643 on Build GRCh38
rs775920119 Chr.15: 74622584-74622584 on Build GRCh38
rs984364791 Chr.15: 74622866-74622866 on Build GRCh38
rs767143600 Chr.15: 74622592-74622592 on Build GRCh38
rs186921543 Chr.15: 74622574-74622574 on Build GRCh38
rs754768654 Chr.15: 74622519-74622519 on Build GRCh38
rs893347761 Chr.15: 74622656-74622657 on Build GRCh38
rs773477147 Chr.15: 74622452-74622452 on Build GRCh38
rs780851953 Chr.15: 74622475-74622475 on Build GRCh38
rs929238711 Chr.15: 74622465-74622465 on Build GRCh38
rs909137489 Chr.15: 74622746-74622746 on Build GRCh38

View Full Product Details