Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75338818-75339308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD4
Entrez Gene ID
54939
Gene Name
COMM domain containing 4
Gene Aliases
-
UniGene
Hs.351327
Gene Chromosome Location
Chr.15: 75335996-75341927 on Build GRCh38
Transcripts
NM_001284377.1 NM_001284378.1 NM_001284379.1 NM_001321844.1 NM_001321845.1 NM_001321846.1 NM_001321847.1 NM_001321848.1 NM_017828.4 NR_104312.1 NR_104313.1 XM_005254509.3 XM_005254510.3 XM_011521739.1 XM_011521740.1 XM_011521741.1 XM_017022385.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1323843 c.278T>A p.L93Q Chr.15: 75339081-75339081 on Build GRCh38
1374594 c.185C>T p.S62L Chr.15: 75338988-75338988 on Build GRCh38
964964 c.215T>C p.L72P Chr.15: 75339018-75339018 on Build GRCh38
3504064 c.346C>T p.P116S Chr.15: 75339308-75339308 on Build GRCh38
7280723 c.302-7delC p.? Chr.15: 75339257-75339257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747722544 Chr.15: 75339220-75339220 on Build GRCh38
rs764480025 Chr.15: 75339268-75339268 on Build GRCh38
rs1046689951 Chr.15: 75338879-75338879 on Build GRCh38
rs758558958 Chr.15: 75339266-75339266 on Build GRCh38
rs769321777 Chr.15: 75339029-75339029 on Build GRCh38
rs368696939 Chr.15: 75339062-75339062 on Build GRCh38
rs776942100 Chr.15: 75339243-75339243 on Build GRCh38
rs747316541 Chr.15: 75339127-75339127 on Build GRCh38
rs181538606 Chr.15: 75338951-75338951 on Build GRCh38
rs199692029 Chr.15: 75338982-75338982 on Build GRCh38
rs769281206 Chr.15: 75338920-75338920 on Build GRCh38
rs771652258 Chr.15: 75339283-75339283 on Build GRCh38
rs766114279 Chr.15: 75338842-75338842 on Build GRCh38
rs776303950 Chr.15: 75338946-75338946 on Build GRCh38
rs34786833 Chr.15: 75338882-75338882 on Build GRCh38
rs3901343 Chr.15: 75339009-75339009 on Build GRCh38
rs754127271 Chr.15: 75339065-75339065 on Build GRCh38
rs200129803 Chr.15: 75339257-75339257 on Build GRCh38
rs781363391 Chr.15: 75338900-75338900 on Build GRCh38
rs769115837 Chr.15: 75339226-75339226 on Build GRCh38
rs373808744 Chr.15: 75339226-75339226 on Build GRCh38
rs747415419 Chr.15: 75339220-75339220 on Build GRCh38
rs755155570 Chr.15: 75339002-75339002 on Build GRCh38
rs750601365 Chr.15: 75338980-75338980 on Build GRCh38
rs386785431 Chr.15: 75339219-75339220 on Build GRCh38
rs377257352 Chr.15: 75338969-75338969 on Build GRCh38
rs746986971 Chr.15: 75339285-75339285 on Build GRCh38
rs929896749 Chr.15: 75338886-75338886 on Build GRCh38
rs367657145 Chr.15: 75339229-75339229 on Build GRCh38
rs900812236 Chr.15: 75339211-75339211 on Build GRCh38
rs781632059 Chr.15: 75339271-75339271 on Build GRCh38
rs748498665 Chr.15: 75338934-75338934 on Build GRCh38
rs916693622 Chr.15: 75338857-75338857 on Build GRCh38
rs190581637 Chr.15: 75338904-75338904 on Build GRCh38
rs372350039 Chr.15: 75338944-75338944 on Build GRCh38
rs369005232 Chr.15: 75338974-75338974 on Build GRCh38
rs373227084 Chr.15: 75339117-75339117 on Build GRCh38
rs376251396 Chr.15: 75339213-75339213 on Build GRCh38
rs780416198 Chr.15: 75339138-75339139 on Build GRCh38
rs201559945 Chr.15: 75338970-75338970 on Build GRCh38
rs774862821 Chr.15: 75338968-75338968 on Build GRCh38
rs553067929 Chr.15: 75339014-75339014 on Build GRCh38
rs748692962 Chr.15: 75339008-75339008 on Build GRCh38
rs375046199 Chr.15: 75339109-75339109 on Build GRCh38
rs142923667 Chr.15: 75339270-75339270 on Build GRCh38
rs776469947 Chr.15: 75338843-75338843 on Build GRCh38
rs772877394 Chr.15: 75339059-75339059 on Build GRCh38
rs376664753 Chr.15: 75338973-75338973 on Build GRCh38
rs199562016 Chr.15: 75338964-75338964 on Build GRCh38
rs532367746 Chr.15: 75339219-75339219 on Build GRCh38
rs761753406 Chr.15: 75339055-75339055 on Build GRCh38
rs543707483 Chr.15: 75339148-75339148 on Build GRCh38
rs779180604 Chr.15: 75339004-75339004 on Build GRCh38
rs377428086 Chr.15: 75339284-75339284 on Build GRCh38
rs747109411 Chr.15: 75339179-75339179 on Build GRCh38
rs564984307 Chr.15: 75339162-75339162 on Build GRCh38
rs760574579 Chr.15: 75339061-75339061 on Build GRCh38
rs146627306 Chr.15: 75339278-75339278 on Build GRCh38
rs749736159 Chr.15: 75338917-75338917 on Build GRCh38
rs747633000 Chr.15: 75339280-75339280 on Build GRCh38
rs867068408 Chr.15: 75339028-75339028 on Build GRCh38
rs1039799302 Chr.15: 75339186-75339186 on Build GRCh38
rs771889232 Chr.15: 75339225-75339225 on Build GRCh38
rs752627029 Chr.15: 75338875-75338875 on Build GRCh38
rs575280335 Chr.15: 75339269-75339269 on Build GRCh38
rs764681794 Chr.15: 75338858-75338858 on Build GRCh38
rs200572633 Chr.15: 75339034-75339034 on Build GRCh38
rs751344951 Chr.15: 75338890-75338890 on Build GRCh38
rs779619886 Chr.15: 75338921-75338921 on Build GRCh38
rs942580501 Chr.15: 75339164-75339164 on Build GRCh38
rs753848560 Chr.15: 75338993-75338993 on Build GRCh38
rs762333352 Chr.15: 75338838-75338838 on Build GRCh38
rs1048255496 Chr.15: 75339160-75339160 on Build GRCh38
rs200660272 Chr.15: 75338992-75338992 on Build GRCh38
rs772512212 Chr.15: 75339012-75339012 on Build GRCh38
rs1030933430 Chr.15: 75339007-75339007 on Build GRCh38
rs535119936 Chr.15: 75338965-75338965 on Build GRCh38
rs929761118 Chr.15: 75339159-75339159 on Build GRCh38
rs753193874 Chr.15: 75339076-75339076 on Build GRCh38
rs144517593 Chr.15: 75339035-75339035 on Build GRCh38
rs750644153 Chr.15: 75338901-75338901 on Build GRCh38
rs748873494 Chr.15: 75339276-75339276 on Build GRCh38
rs148548797 Chr.15: 75339071-75339071 on Build GRCh38
rs139615169 Chr.15: 75339040-75339040 on Build GRCh38
rs767322876 Chr.15: 75338976-75338976 on Build GRCh38
rs111503569 Chr.15: 75339142-75339142 on Build GRCh38
rs1006600407 Chr.15: 75338924-75338924 on Build GRCh38
rs537743487 Chr.15: 75338913-75338913 on Build GRCh38
rs765426701 Chr.15: 75339074-75339074 on Build GRCh38
rs758973188 Chr.15: 75339108-75339108 on Build GRCh38
rs757683558 Chr.15: 75339144-75339144 on Build GRCh38
rs778010069 Chr.15: 75339013-75339013 on Build GRCh38
rs762807056 Chr.15: 75338878-75338878 on Build GRCh38
rs373430411 Chr.15: 75339279-75339279 on Build GRCh38
rs759051820 Chr.15: 75338856-75338856 on Build GRCh38
rs757095545 Chr.15: 75338895-75338895 on Build GRCh38
rs563697646 Chr.15: 75339056-75339056 on Build GRCh38
rs34234047 Chr.15: 75339246-75339246 on Build GRCh38

View Full Product Details