Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 79293318-79293830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD34C
Entrez Gene ID
390616
Gene Name
ankyrin repeat domain 34C
Gene Aliases
-
UniGene
Hs.531231
Gene Chromosome Location
Chr.15: 79282804-79298239 on Build GRCh38
Transcripts
NM_001146341.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6563148 c.529G>C p.E177Q Chr.15: 79293813-79293813 on Build GRCh38
4484096 c.276C>T p.A92A Chr.15: 79293560-79293560 on Build GRCh38
4486772 c.309C>T p.S103S Chr.15: 79293593-79293593 on Build GRCh38
6555125 c.296G>T p.G99V Chr.15: 79293580-79293580 on Build GRCh38
1374849 c.132C>T p.G44G Chr.15: 79293416-79293416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760069754 Chr.15: 79293643-79293643 on Build GRCh38
rs535985577 Chr.15: 79293371-79293371 on Build GRCh38
rs921447487 Chr.15: 79293494-79293494 on Build GRCh38
rs572751058 Chr.15: 79293670-79293670 on Build GRCh38
rs541933496 Chr.15: 79293692-79293692 on Build GRCh38
rs867774663 Chr.15: 79293681-79293681 on Build GRCh38
rs565005422 Chr.15: 79293761-79293761 on Build GRCh38
rs1041777319 Chr.15: 79293684-79293684 on Build GRCh38
rs267604339 Chr.15: 79293799-79293799 on Build GRCh38
rs773427605 Chr.15: 79293539-79293539 on Build GRCh38
rs778701368 Chr.15: 79293548-79293548 on Build GRCh38
rs868182521 Chr.15: 79293506-79293506 on Build GRCh38
rs949779751 Chr.15: 79293461-79293461 on Build GRCh38
rs1007003517 Chr.15: 79293682-79293682 on Build GRCh38
rs1019578812 Chr.15: 79293457-79293457 on Build GRCh38
rs370963639 Chr.15: 79293434-79293434 on Build GRCh38
rs748425838 Chr.15: 79293491-79293491 on Build GRCh38
rs555151880 Chr.15: 79293791-79293791 on Build GRCh38
rs749040437 Chr.15: 79293425-79293425 on Build GRCh38
rs914322897 Chr.15: 79293644-79293644 on Build GRCh38
rs989883681 Chr.15: 79293614-79293614 on Build GRCh38
rs938609003 Chr.15: 79293702-79293702 on Build GRCh38
rs754807675 Chr.15: 79293437-79293437 on Build GRCh38
rs553046789 Chr.15: 79293669-79293669 on Build GRCh38
rs549187926 Chr.15: 79293416-79293416 on Build GRCh38
rs941491229 Chr.15: 79293359-79293359 on Build GRCh38
rs554040436 Chr.15: 79293570-79293570 on Build GRCh38
rs894336238 Chr.15: 79293801-79293801 on Build GRCh38
rs558511797 Chr.15: 79293608-79293608 on Build GRCh38
rs779874764 Chr.15: 79293417-79293417 on Build GRCh38
rs76207286 Chr.15: 79293647-79293647 on Build GRCh38
rs377442575 Chr.15: 79293401-79293401 on Build GRCh38
rs755261869 Chr.15: 79293450-79293450 on Build GRCh38
rs974198742 Chr.15: 79293771-79293771 on Build GRCh38
rs918852567 Chr.15: 79293777-79293777 on Build GRCh38
rs750040034 Chr.15: 79293754-79293754 on Build GRCh38
rs905731678 Chr.15: 79293523-79293523 on Build GRCh38
rs955502762 Chr.15: 79293656-79293656 on Build GRCh38
rs369047222 Chr.15: 79293458-79293458 on Build GRCh38
rs578238681 Chr.15: 79293633-79293633 on Build GRCh38
rs772276700 Chr.15: 79293529-79293529 on Build GRCh38

View Full Product Details