Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 79294871-79295381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD34C
Entrez Gene ID
390616
Gene Name
ankyrin repeat domain 34C
Gene Aliases
-
UniGene
Hs.531231
Gene Chromosome Location
Chr.15: 79282804-79298239 on Build GRCh38
Transcripts
NM_001146341.1

SNP Detail

SNP ID Chromosome Location
rs953869311 Chr.15: 79295225-79295225 on Build GRCh38
rs764110477 Chr.15: 79294936-79294936 on Build GRCh38
rs946763601 Chr.15: 79295245-79295245 on Build GRCh38
rs967125395 Chr.15: 79294984-79294984 on Build GRCh38
rs1042659208 Chr.15: 79294960-79294960 on Build GRCh38
rs1012412006 Chr.15: 79295344-79295344 on Build GRCh38
rs923908925 Chr.15: 79295261-79295261 on Build GRCh38
rs1004169822 Chr.15: 79295115-79295115 on Build GRCh38
rs181824493 Chr.15: 79295232-79295232 on Build GRCh38
rs185415153 Chr.15: 79295237-79295237 on Build GRCh38
rs919836475 Chr.15: 79295108-79295108 on Build GRCh38
rs1022708046 Chr.15: 79295142-79295142 on Build GRCh38
rs879173523 Chr.15: 79295256-79295256 on Build GRCh38
rs777748932 Chr.15: 79295349-79295349 on Build GRCh38
rs1002320125 Chr.15: 79295187-79295187 on Build GRCh38
rs774980392 Chr.15: 79294906-79294906 on Build GRCh38
rs770213713 Chr.15: 79294954-79294954 on Build GRCh38
rs754817147 Chr.15: 79295129-79295129 on Build GRCh38
rs977053469 Chr.15: 79295172-79295172 on Build GRCh38
rs1042399891 Chr.15: 79295258-79295258 on Build GRCh38
rs559387171 Chr.15: 79295186-79295186 on Build GRCh38
rs1020460863 Chr.15: 79294952-79294952 on Build GRCh38
rs908634625 Chr.15: 79295148-79295148 on Build GRCh38
rs548081943 Chr.15: 79295324-79295324 on Build GRCh38
rs1019924454 Chr.15: 79295243-79295243 on Build GRCh38
rs1019364725 Chr.15: 79295355-79295355 on Build GRCh38
rs368501433 Chr.15: 79295278-79295278 on Build GRCh38
rs542610009 Chr.15: 79294951-79294951 on Build GRCh38
rs1012694008 Chr.15: 79295141-79295141 on Build GRCh38
rs1015634785 Chr.15: 79295152-79295152 on Build GRCh38
rs1047027103 Chr.15: 79295139-79295139 on Build GRCh38
rs753073165 Chr.15: 79295256-79295257 on Build GRCh38
rs866412290 Chr.15: 79295143-79295143 on Build GRCh38
rs531884055 Chr.15: 79295149-79295149 on Build GRCh38
rs1005217042 Chr.15: 79295151-79295151 on Build GRCh38
rs529310359 Chr.15: 79294907-79294907 on Build GRCh38
rs986649896 Chr.15: 79294997-79294997 on Build GRCh38
rs961333806 Chr.15: 79295156-79295156 on Build GRCh38
rs912422585 Chr.15: 79295013-79295013 on Build GRCh38
rs942495933 Chr.15: 79295022-79295022 on Build GRCh38
rs915272633 Chr.15: 79294937-79294937 on Build GRCh38
rs931311024 Chr.15: 79295138-79295138 on Build GRCh38
rs1054005425 Chr.15: 79295211-79295211 on Build GRCh38
rs975359626 Chr.15: 79295038-79295038 on Build GRCh38
rs895280593 Chr.15: 79295131-79295131 on Build GRCh38
rs150706029 Chr.15: 79295124-79295124 on Build GRCh38
rs939637088 Chr.15: 79295277-79295277 on Build GRCh38
rs902767751 Chr.15: 79295096-79295096 on Build GRCh38
rs762306097 Chr.15: 79294930-79294930 on Build GRCh38
rs895343748 Chr.15: 79295306-79295306 on Build GRCh38
rs1002436000 Chr.15: 79294920-79294920 on Build GRCh38

View Full Product Details