Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 82759032-82759557 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSD2
Entrez Gene ID
123722
Gene Name
fibronectin type III and SPRY domain containing 2
Gene Aliases
SPRYD1
UniGene
Hs.719372
Gene Chromosome Location
Chr.15: 82755365-82807698 on Build GRCh38
Transcripts
NM_001007122.3 NM_001281805.1 NM_001281806.1 XM_005272425.4 XM_011521235.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3754509 c.2158G>A p.E720K Chr.15: 82759440-82759440 on Build GRCh38
5012926 c.2093C>T p.S698L Chr.15: 82759505-82759505 on Build GRCh38
3706768 c.2235T>C p.H745H Chr.15: 82759363-82759363 on Build GRCh38
3420666 c.2066T>G p.I689S Chr.15: 82759532-82759532 on Build GRCh38
3420665 c.2169T>C p.H723H Chr.15: 82759429-82759429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939468554 Chr.15: 82759058-82759058 on Build GRCh38
rs767590780 Chr.15: 82759446-82759446 on Build GRCh38
rs371581822 Chr.15: 82759332-82759332 on Build GRCh38
rs750635394 Chr.15: 82759313-82759313 on Build GRCh38
rs1108134 Chr.15: 82759440-82759440 on Build GRCh38
rs563943895 Chr.15: 82759293-82759293 on Build GRCh38
rs755325923 Chr.15: 82759468-82759468 on Build GRCh38
rs777940223 Chr.15: 82759233-82759233 on Build GRCh38
rs759772436 Chr.15: 82759528-82759528 on Build GRCh38
rs751612652 Chr.15: 82759346-82759346 on Build GRCh38
rs148135723 Chr.15: 82759107-82759107 on Build GRCh38
rs756812579 Chr.15: 82759354-82759354 on Build GRCh38
rs988238962 Chr.15: 82759267-82759267 on Build GRCh38
rs1025743089 Chr.15: 82759136-82759136 on Build GRCh38
rs1007762558 Chr.15: 82759138-82759138 on Build GRCh38
rs749592336 Chr.15: 82759385-82759385 on Build GRCh38
rs964378735 Chr.15: 82759223-82759223 on Build GRCh38
rs202018585 Chr.15: 82759376-82759376 on Build GRCh38
rs926492079 Chr.15: 82759244-82759244 on Build GRCh38
rs368720586 Chr.15: 82759357-82759357 on Build GRCh38
rs755695967 Chr.15: 82759460-82759460 on Build GRCh38
rs1108135 Chr.15: 82759441-82759441 on Build GRCh38
rs758501349 Chr.15: 82759186-82759186 on Build GRCh38
rs375920867 Chr.15: 82759425-82759425 on Build GRCh38
rs976535077 Chr.15: 82759163-82759163 on Build GRCh38
rs879197813 Chr.15: 82759101-82759101 on Build GRCh38
rs532163313 Chr.15: 82759294-82759294 on Build GRCh38
rs936848172 Chr.15: 82759261-82759261 on Build GRCh38
rs958259485 Chr.15: 82759416-82759416 on Build GRCh38
rs942835982 Chr.15: 82759149-82759162 on Build GRCh38
rs368352558 Chr.15: 82759318-82759318 on Build GRCh38
rs779539234 Chr.15: 82759464-82759464 on Build GRCh38
rs117998650 Chr.15: 82759102-82759102 on Build GRCh38
rs778442048 Chr.15: 82759424-82759424 on Build GRCh38
rs748312699 Chr.15: 82759324-82759325 on Build GRCh38
rs772584765 Chr.15: 82759414-82759414 on Build GRCh38
rs987227050 Chr.15: 82759191-82759191 on Build GRCh38
rs765490463 Chr.15: 82759208-82759208 on Build GRCh38
rs939399315 Chr.15: 82759265-82759265 on Build GRCh38
rs1034257966 Chr.15: 82759098-82759098 on Build GRCh38
rs28685273 Chr.15: 82759284-82759284 on Build GRCh38
rs201393187 Chr.15: 82759374-82759374 on Build GRCh38
rs374397158 Chr.15: 82759302-82759302 on Build GRCh38
rs975672240 Chr.15: 82759231-82759231 on Build GRCh38
rs1045379935 Chr.15: 82759060-82759060 on Build GRCh38
rs945839620 Chr.15: 82759290-82759290 on Build GRCh38
rs1055721400 Chr.15: 82759307-82759307 on Build GRCh38
rs138702546 Chr.15: 82759339-82759339 on Build GRCh38
rs774837721 Chr.15: 82759401-82759401 on Build GRCh38
rs575070747 Chr.15: 82759140-82759140 on Build GRCh38
rs371725076 Chr.15: 82759363-82759363 on Build GRCh38
rs190410190 Chr.15: 82759097-82759097 on Build GRCh38
rs781283860 Chr.15: 82759303-82759304 on Build GRCh38
rs1019636102 Chr.15: 82759209-82759209 on Build GRCh38
rs1029701407 Chr.15: 82759165-82759165 on Build GRCh38
rs925537137 Chr.15: 82759243-82759243 on Build GRCh38
rs771084427 Chr.15: 82759494-82759494 on Build GRCh38
rs773791671 Chr.15: 82759487-82759487 on Build GRCh38
rs1001485341 Chr.15: 82759095-82759095 on Build GRCh38
rs1008340253 Chr.15: 82759366-82759366 on Build GRCh38
rs980547857 Chr.15: 82759238-82759238 on Build GRCh38
rs567939519 Chr.15: 82759406-82759406 on Build GRCh38
rs559405778 Chr.15: 82759303-82759303 on Build GRCh38
rs766629023 Chr.15: 82759322-82759322 on Build GRCh38
rs199848732 Chr.15: 82759493-82759493 on Build GRCh38
rs375842909 Chr.15: 82759429-82759429 on Build GRCh38
rs962573637 Chr.15: 82759420-82759420 on Build GRCh38
rs769051141 Chr.15: 82759388-82759388 on Build GRCh38
rs539026057 Chr.15: 82759486-82759486 on Build GRCh38
rs1022594022 Chr.15: 82759110-82759111 on Build GRCh38
rs74569644 Chr.15: 82759066-82759066 on Build GRCh38
rs758237661 Chr.15: 82759336-82759336 on Build GRCh38
rs182082697 Chr.15: 82759522-82759522 on Build GRCh38
rs913001352 Chr.15: 82759271-82759271 on Build GRCh38
rs941178585 Chr.15: 82759236-82759236 on Build GRCh38
rs376416198 Chr.15: 82759371-82759371 on Build GRCh38
rs1043956414 Chr.15: 82759150-82759163 on Build GRCh38
rs779194704 Chr.15: 82759483-82759483 on Build GRCh38
rs909831837 Chr.15: 82759230-82759230 on Build GRCh38
rs926410876 Chr.15: 82759425-82759426 on Build GRCh38
rs1056840488 Chr.15: 82759076-82759076 on Build GRCh38
rs150119830 Chr.15: 82759291-82759291 on Build GRCh38
rs748520224 Chr.15: 82759484-82759484 on Build GRCh38
rs201500466 Chr.15: 82759466-82759466 on Build GRCh38
rs752978619 Chr.15: 82759134-82759134 on Build GRCh38
rs751566454 Chr.15: 82759240-82759240 on Build GRCh38
rs375670260 Chr.15: 82759450-82759450 on Build GRCh38
rs879070469 Chr.15: 82759351-82759351 on Build GRCh38
rs142701820 Chr.15: 82759423-82759423 on Build GRCh38
rs919861254 Chr.15: 82759513-82759513 on Build GRCh38
rs780790409 Chr.15: 82759358-82759358 on Build GRCh38
rs767653784 Chr.15: 82759306-82759306 on Build GRCh38
rs760080782 Chr.15: 82759503-82759503 on Build GRCh38
rs892460412 Chr.15: 82759282-82759282 on Build GRCh38
rs995298647 Chr.15: 82759132-82759132 on Build GRCh38

View Full Product Details