Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 12661613-12662119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPPED1
Entrez Gene ID
55313
Gene Name
calcineurin like phosphoesterase domain containing 1
Gene Aliases
CSTP1
UniGene
Hs.460002
Gene Chromosome Location
Chr.16: 12659799-12803887 on Build GRCh38
Transcripts
NM_001099455.1 NM_018340.2

SNP Detail

SNP ID Chromosome Location
rs905006392 Chr.16: 12661749-12661749 on Build GRCh38
rs150572471 Chr.16: 12661986-12661986 on Build GRCh38
rs374383848 Chr.16: 12661862-12661862 on Build GRCh38
rs1044914369 Chr.16: 12661742-12661742 on Build GRCh38
rs1002998884 Chr.16: 12662062-12662062 on Build GRCh38
rs947374251 Chr.16: 12662040-12662040 on Build GRCh38
rs758366919 Chr.16: 12662060-12662060 on Build GRCh38
rs529475868 Chr.16: 12661761-12661761 on Build GRCh38
rs74011934 Chr.16: 12661814-12661814 on Build GRCh38
rs35114670 Chr.16: 12661681-12661682 on Build GRCh38
rs1037939989 Chr.16: 12661693-12661693 on Build GRCh38
rs746949735 Chr.16: 12662062-12662062 on Build GRCh38
rs932444905 Chr.16: 12661924-12661924 on Build GRCh38
rs1023026726 Chr.16: 12661769-12661769 on Build GRCh38
rs778842898 Chr.16: 12662049-12662049 on Build GRCh38
rs954561156 Chr.16: 12661812-12661812 on Build GRCh38
rs551097973 Chr.16: 12661828-12661828 on Build GRCh38
rs1047679488 Chr.16: 12661716-12661716 on Build GRCh38
rs905973730 Chr.16: 12661770-12661770 on Build GRCh38
rs752443685 Chr.16: 12661745-12661747 on Build GRCh38
rs905935906 Chr.16: 12662056-12662056 on Build GRCh38
rs983699603 Chr.16: 12661906-12661906 on Build GRCh38
rs567551795 Chr.16: 12661990-12661990 on Build GRCh38
rs929524228 Chr.16: 12661887-12661887 on Build GRCh38
rs1009491323 Chr.16: 12661779-12661779 on Build GRCh38
rs917243784 Chr.16: 12661908-12661908 on Build GRCh38
rs973795347 Chr.16: 12661875-12661875 on Build GRCh38
rs973825098 Chr.16: 12661636-12661636 on Build GRCh38
rs950117893 Chr.16: 12661931-12661931 on Build GRCh38
rs1044407228 Chr.16: 12662044-12662044 on Build GRCh38
rs536466301 Chr.16: 12662052-12662052 on Build GRCh38
rs537039197 Chr.16: 12661866-12661866 on Build GRCh38
rs777382886 Chr.16: 12661757-12661757 on Build GRCh38
rs868155582 Chr.16: 12661759-12661759 on Build GRCh38
rs371101905 Chr.16: 12661704-12661704 on Build GRCh38
rs770388172 Chr.16: 12661944-12661944 on Build GRCh38
rs1053043835 Chr.16: 12661744-12661744 on Build GRCh38
rs757683384 Chr.16: 12662090-12662090 on Build GRCh38
rs1057474176 Chr.16: 12662087-12662087 on Build GRCh38
rs577933366 Chr.16: 12661948-12661948 on Build GRCh38
rs549128430 Chr.16: 12661775-12661775 on Build GRCh38
rs914552995 Chr.16: 12662017-12662017 on Build GRCh38
rs76337490 Chr.16: 12661705-12661705 on Build GRCh38
rs905913064 Chr.16: 12662011-12662011 on Build GRCh38
rs753000701 Chr.16: 12661648-12661648 on Build GRCh38
rs921069233 Chr.16: 12661687-12661687 on Build GRCh38
rs973953031 Chr.16: 12661859-12661859 on Build GRCh38
rs377633010 Chr.16: 12661732-12661732 on Build GRCh38
rs188537154 Chr.16: 12661848-12661848 on Build GRCh38
rs753990861 Chr.16: 12662006-12662006 on Build GRCh38
rs551004053 Chr.16: 12661995-12661995 on Build GRCh38
rs932495532 Chr.16: 12661689-12661689 on Build GRCh38
rs1014330807 Chr.16: 12661737-12661737 on Build GRCh38
rs934531608 Chr.16: 12661979-12661979 on Build GRCh38
rs893705391 Chr.16: 12661776-12661776 on Build GRCh38
rs912435291 Chr.16: 12661942-12661942 on Build GRCh38
rs748836888 Chr.16: 12661884-12661884 on Build GRCh38
rs1015234428 Chr.16: 12661831-12661831 on Build GRCh38
rs764080899 Chr.16: 12661920-12661921 on Build GRCh38
rs565657941 Chr.16: 12661790-12661790 on Build GRCh38

View Full Product Details