Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 15502185-15502686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf45
Entrez Gene ID
89927
Gene Name
chromosome 16 open reading frame 45
Gene Aliases
MINP
UniGene
Hs.401798
Gene Chromosome Location
Chr.16: 15434468-15594521 on Build GRCh38
Transcripts
NM_001142469.1 NM_033201.2 XM_011522731.2 XM_017023824.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3817259 c.51G>T p.K17N Chr.16: 15502372-15502372 on Build GRCh38
5384269 c.35G>A p.G12E Chr.16: 15502356-15502356 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1033408261 Chr.16: 15502628-15502628 on Build GRCh38
rs950257611 Chr.16: 15502257-15502257 on Build GRCh38
rs368928707 Chr.16: 15502271-15502271 on Build GRCh38
rs571948191 Chr.16: 15502358-15502358 on Build GRCh38
rs991637604 Chr.16: 15502230-15502230 on Build GRCh38
rs927586027 Chr.16: 15502280-15502280 on Build GRCh38
rs12932382 Chr.16: 15502424-15502424 on Build GRCh38
rs946444005 Chr.16: 15502574-15502574 on Build GRCh38
rs916043133 Chr.16: 15502522-15502522 on Build GRCh38
rs868533817 Chr.16: 15502575-15502575 on Build GRCh38
rs561451988 Chr.16: 15502547-15502547 on Build GRCh38
rs753387379 Chr.16: 15502384-15502384 on Build GRCh38
rs987813860 Chr.16: 15502567-15502567 on Build GRCh38
rs12918592 Chr.16: 15502378-15502378 on Build GRCh38
rs966378835 Chr.16: 15502264-15502264 on Build GRCh38
rs527944708 Chr.16: 15502398-15502398 on Build GRCh38
rs145970306 Chr.16: 15502519-15502519 on Build GRCh38
rs930791366 Chr.16: 15502488-15502488 on Build GRCh38
rs1000626025 Chr.16: 15502627-15502627 on Build GRCh38
rs979594174 Chr.16: 15502330-15502330 on Build GRCh38
rs777299616 Chr.16: 15502382-15502382 on Build GRCh38
rs916179063 Chr.16: 15502249-15502249 on Build GRCh38
rs952350537 Chr.16: 15502226-15502226 on Build GRCh38
rs142309512 Chr.16: 15502352-15502352 on Build GRCh38
rs527823612 Chr.16: 15502291-15502291 on Build GRCh38
rs944534743 Chr.16: 15502647-15502647 on Build GRCh38
rs572586630 Chr.16: 15502505-15502505 on Build GRCh38
rs757769117 Chr.16: 15502278-15502278 on Build GRCh38
rs924888217 Chr.16: 15502541-15502541 on Build GRCh38
rs968294761 Chr.16: 15502604-15502605 on Build GRCh38
rs188565459 Chr.16: 15502331-15502331 on Build GRCh38
rs545210362 Chr.16: 15502630-15502630 on Build GRCh38
rs779540340 Chr.16: 15502417-15502417 on Build GRCh38
rs562629773 Chr.16: 15502579-15502579 on Build GRCh38
rs772368115 Chr.16: 15502527-15502527 on Build GRCh38
rs560406851 Chr.16: 15502659-15502659 on Build GRCh38
rs991858318 Chr.16: 15502656-15502656 on Build GRCh38
rs752460743 Chr.16: 15502350-15502350 on Build GRCh38
rs952149806 Chr.16: 15502459-15502459 on Build GRCh38
rs12448500 Chr.16: 15502285-15502285 on Build GRCh38
rs993510422 Chr.16: 15502439-15502439 on Build GRCh38
rs577913766 Chr.16: 15502626-15502626 on Build GRCh38
rs538856868 Chr.16: 15502415-15502415 on Build GRCh38
rs1007628600 Chr.16: 15502529-15502529 on Build GRCh38
rs767007607 Chr.16: 15502391-15502393 on Build GRCh38
rs759302527 Chr.16: 15502309-15502309 on Build GRCh38
rs756886109 Chr.16: 15502408-15502408 on Build GRCh38
rs549365045 Chr.16: 15502301-15502301 on Build GRCh38
rs573557873 Chr.16: 15502531-15502531 on Build GRCh38
rs1028176932 Chr.16: 15502458-15502458 on Build GRCh38
rs551640861 Chr.16: 15502281-15502281 on Build GRCh38
rs889049045 Chr.16: 15502379-15502379 on Build GRCh38
rs12932269 Chr.16: 15502420-15502420 on Build GRCh38
rs372704770 Chr.16: 15502436-15502436 on Build GRCh38
rs1040425470 Chr.16: 15502536-15502536 on Build GRCh38
rs533656702 Chr.16: 15502509-15502509 on Build GRCh38
rs72770177 Chr.16: 15502573-15502573 on Build GRCh38
rs901834528 Chr.16: 15502542-15502542 on Build GRCh38
rs1050564823 Chr.16: 15502214-15502214 on Build GRCh38

View Full Product Details