Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19714425-19714931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KNOP1
Entrez Gene ID
400506
Gene Name
lysine rich nucleolar protein 1
Gene Aliases
101F10.1, C16orf88, FAM191A, TSG118
UniGene
Hs.585209
Gene Chromosome Location
Chr.16: 19706347-19718240 on Build GRCh38
Transcripts
NM_001012991.2 XM_005255317.4 XM_006721046.3 XM_011545855.1 XM_017023231.1 XM_017023232.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4607297 c.108T>G p.D36E Chr.16: 19714928-19714928 on Build GRCh38
7198536 c.365C>G p.A122G Chr.16: 19714671-19714671 on Build GRCh38
4611411 c.472_473insG p.A158fs*16 Chr.16: 19714563-19714564 on Build GRCh38
6034095 c.207_209delGAA p.K72delK Chr.16: 19714827-19714829 on Build GRCh38
4854446 c.526G>T p.E176* Chr.16: 19714510-19714510 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746730360 Chr.16: 19714789-19714789 on Build GRCh38
rs368756116 Chr.16: 19714846-19714846 on Build GRCh38
rs763134157 Chr.16: 19714633-19714633 on Build GRCh38
rs766085786 Chr.16: 19714595-19714597 on Build GRCh38
rs746019580 Chr.16: 19714710-19714710 on Build GRCh38
rs901320066 Chr.16: 19714573-19714573 on Build GRCh38
rs756062811 Chr.16: 19714820-19714831 on Build GRCh38
rs765683713 Chr.16: 19714875-19714875 on Build GRCh38
rs756789363 Chr.16: 19714669-19714669 on Build GRCh38
rs767293563 Chr.16: 19714451-19714451 on Build GRCh38
rs767894674 Chr.16: 19714643-19714643 on Build GRCh38
rs924101396 Chr.16: 19714603-19714603 on Build GRCh38
rs747921062 Chr.16: 19714896-19714896 on Build GRCh38
rs766441939 Chr.16: 19714640-19714640 on Build GRCh38
rs532318324 Chr.16: 19714882-19714882 on Build GRCh38
rs751808453 Chr.16: 19714521-19714521 on Build GRCh38
rs778266449 Chr.16: 19714660-19714660 on Build GRCh38
rs781096824 Chr.16: 19714893-19714893 on Build GRCh38
rs765384296 Chr.16: 19714460-19714460 on Build GRCh38
rs376355827 Chr.16: 19714512-19714512 on Build GRCh38
rs761906213 Chr.16: 19714729-19714729 on Build GRCh38
rs768653754 Chr.16: 19714716-19714716 on Build GRCh38
rs754724879 Chr.16: 19714892-19714892 on Build GRCh38
rs748670501 Chr.16: 19714548-19714548 on Build GRCh38
rs548962099 Chr.16: 19714761-19714761 on Build GRCh38
rs757024607 Chr.16: 19714575-19714575 on Build GRCh38
rs764157990 Chr.16: 19714621-19714621 on Build GRCh38
rs370606353 Chr.16: 19714664-19714664 on Build GRCh38
rs745575760 Chr.16: 19714580-19714580 on Build GRCh38
rs373865859 Chr.16: 19714587-19714587 on Build GRCh38
rs990767824 Chr.16: 19714569-19714569 on Build GRCh38
rs200792496 Chr.16: 19714564-19714564 on Build GRCh38
rs756568608 Chr.16: 19714657-19714657 on Build GRCh38
rs775494770 Chr.16: 19714452-19714452 on Build GRCh38
rs367676691 Chr.16: 19714634-19714634 on Build GRCh38
rs201558781 Chr.16: 19714695-19714695 on Build GRCh38
rs558221133 Chr.16: 19714473-19714473 on Build GRCh38
rs751286698 Chr.16: 19714542-19714542 on Build GRCh38
rs201956943 Chr.16: 19714564-19714564 on Build GRCh38
rs550794692 Chr.16: 19714796-19714796 on Build GRCh38
rs754332903 Chr.16: 19714768-19714768 on Build GRCh38
rs775196338 Chr.16: 19714608-19714608 on Build GRCh38
rs146629491 Chr.16: 19714771-19714771 on Build GRCh38
rs755207803 Chr.16: 19714642-19714642 on Build GRCh38
rs754239198 Chr.16: 19714835-19714835 on Build GRCh38
rs751190284 Chr.16: 19714635-19714651 on Build GRCh38
rs745510667 Chr.16: 19714685-19714685 on Build GRCh38
rs777515175 Chr.16: 19714819-19714820 on Build GRCh38
rs780571889 Chr.16: 19714645-19714645 on Build GRCh38
rs200153333 Chr.16: 19714760-19714760 on Build GRCh38
rs11546731 Chr.16: 19714493-19714493 on Build GRCh38
rs370164619 Chr.16: 19714541-19714541 on Build GRCh38
rs34427159 Chr.16: 19714552-19714553 on Build GRCh38
rs960845807 Chr.16: 19714569-19714569 on Build GRCh38
rs778453570 Chr.16: 19714775-19714775 on Build GRCh38
rs1023554071 Chr.16: 19714806-19714806 on Build GRCh38
rs533912193 Chr.16: 19714550-19714550 on Build GRCh38
rs781187376 Chr.16: 19714801-19714801 on Build GRCh38
rs532893765 Chr.16: 19714445-19714447 on Build GRCh38
rs749890886 Chr.16: 19714878-19714878 on Build GRCh38
rs779861814 Chr.16: 19714785-19714785 on Build GRCh38
rs1035534530 Chr.16: 19714567-19714567 on Build GRCh38
rs368422228 Chr.16: 19714680-19714680 on Build GRCh38
rs373473505 Chr.16: 19714490-19714490 on Build GRCh38
rs555663877 Chr.16: 19714574-19714574 on Build GRCh38
rs200472251 Chr.16: 19714701-19714701 on Build GRCh38
rs202037988 Chr.16: 19714762-19714762 on Build GRCh38
rs770735825 Chr.16: 19714827-19714829 on Build GRCh38
rs1005437383 Chr.16: 19714757-19714757 on Build GRCh38
rs754452355 Chr.16: 19714636-19714638 on Build GRCh38
rs774444123 Chr.16: 19714807-19714807 on Build GRCh38
rs779773768 Chr.16: 19714883-19714883 on Build GRCh38
rs774058154 Chr.16: 19714577-19714577 on Build GRCh38
rs756656687 Chr.16: 19714561-19714561 on Build GRCh38
rs1043097572 Chr.16: 19714514-19714514 on Build GRCh38
rs1047263020 Chr.16: 19714583-19714583 on Build GRCh38
rs201093179 Chr.16: 19714818-19714818 on Build GRCh38
rs773373442 Chr.16: 19714624-19714624 on Build GRCh38
rs760751029 Chr.16: 19714458-19714458 on Build GRCh38
rs372324081 Chr.16: 19714756-19714756 on Build GRCh38
rs749043471 Chr.16: 19714823-19714824 on Build GRCh38
rs922690133 Chr.16: 19714902-19714902 on Build GRCh38
rs58543897 Chr.16: 19714802-19714802 on Build GRCh38
rs758018997 Chr.16: 19714784-19714784 on Build GRCh38
rs748730354 Chr.16: 19714661-19714661 on Build GRCh38
rs544818600 Chr.16: 19714751-19714751 on Build GRCh38
rs759389493 Chr.16: 19714448-19714448 on Build GRCh38
rs775686240 Chr.16: 19714822-19714822 on Build GRCh38
rs760947781 Chr.16: 19714828-19714828 on Build GRCh38
rs760658613 Chr.16: 19714612-19714612 on Build GRCh38
rs764421510 Chr.16: 19714833-19714833 on Build GRCh38
rs762340026 Chr.16: 19714873-19714873 on Build GRCh38
rs368403074 Chr.16: 19714776-19714776 on Build GRCh38
rs748110675 Chr.16: 19714704-19714704 on Build GRCh38
rs770063374 Chr.16: 19714718-19714718 on Build GRCh38
rs35760533 Chr.16: 19714546-19714547 on Build GRCh38
rs759553262 Chr.16: 19714809-19714809 on Build GRCh38
rs780876645 Chr.16: 19714653-19714653 on Build GRCh38
rs750865246 Chr.16: 19714475-19714475 on Build GRCh38
rs752903191 Chr.16: 19714500-19714508 on Build GRCh38
rs753095555 Chr.16: 19714652-19714652 on Build GRCh38
rs771889409 Chr.16: 19714584-19714584 on Build GRCh38
rs751240167 Chr.16: 19714890-19714890 on Build GRCh38

View Full Product Details