Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67289092-67289601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD19
Entrez Gene ID
146212
Gene Name
potassium channel tetramerization domain containing 19
Gene Aliases
-
UniGene
Hs.299127
Gene Chromosome Location
Chr.16: 67289490-67326758 on Build GRCh38
Transcripts
NM_001100915.1 NM_001129727.2 NM_001129728.1 NM_001129729.2 NM_001129731.2 XM_011522985.2 XM_011522986.2 XM_011522987.2 XM_011522988.2

Gene Symbol
PLEKHG4
Entrez Gene ID
25894
Gene Name
pleckstrin homology and RhoGEF domain containing G4
Gene Aliases
ARHGEF44, PRTPHN1, SCA4
UniGene
Hs.188781
Gene Chromosome Location
Chr.16: 67277820-67289500 on Build GRCh38
Transcripts
NM_001100915.1 NM_001129727.2 NM_001129728.1 NM_001129729.2 NM_001129731.2 XM_011522985.2 XM_011522986.2 XM_011522987.2 XM_011522988.2

SNP Detail

SNP ID Chromosome Location
rs1013767490 Chr.16: 67289336-67289336 on Build GRCh38
rs935823657 Chr.16: 67289226-67289226 on Build GRCh38
rs1006782079 Chr.16: 67289311-67289311 on Build GRCh38
rs998132675 Chr.16: 67289123-67289123 on Build GRCh38
rs991101651 Chr.16: 67289165-67289165 on Build GRCh38
rs924474769 Chr.16: 67289216-67289216 on Build GRCh38
rs887845042 Chr.16: 67289569-67289569 on Build GRCh38
rs1033889217 Chr.16: 67289142-67289142 on Build GRCh38
rs954059622 Chr.16: 67289182-67289182 on Build GRCh38
rs549053116 Chr.16: 67289468-67289468 on Build GRCh38
rs1028282262 Chr.16: 67289546-67289546 on Build GRCh38
rs1031834457 Chr.16: 67289413-67289413 on Build GRCh38
rs752161156 Chr.16: 67289555-67289555 on Build GRCh38
rs910365749 Chr.16: 67289457-67289457 on Build GRCh38
rs766298598 Chr.16: 67289538-67289538 on Build GRCh38
rs116003678 Chr.16: 67289282-67289282 on Build GRCh38
rs1006508794 Chr.16: 67289132-67289132 on Build GRCh38
rs927543058 Chr.16: 67289298-67289298 on Build GRCh38
rs35097456 Chr.16: 67289300-67289300 on Build GRCh38
rs763301444 Chr.16: 67289529-67289529 on Build GRCh38
rs777348066 Chr.16: 67289577-67289577 on Build GRCh38
rs1044634325 Chr.16: 67289293-67289293 on Build GRCh38
rs957257577 Chr.16: 67289438-67289438 on Build GRCh38
rs532927642 Chr.16: 67289191-67289191 on Build GRCh38
rs997986285 Chr.16: 67289510-67289510 on Build GRCh38
rs374179662 Chr.16: 67289545-67289545 on Build GRCh38
rs937719709 Chr.16: 67289322-67289322 on Build GRCh38
rs1048034129 Chr.16: 67289323-67289323 on Build GRCh38
rs978754972 Chr.16: 67289210-67289210 on Build GRCh38
rs959628433 Chr.16: 67289327-67289327 on Build GRCh38
rs971186289 Chr.16: 67289208-67289208 on Build GRCh38
rs1023925502 Chr.16: 67289356-67289356 on Build GRCh38
rs888223729 Chr.16: 67289354-67289354 on Build GRCh38
rs901009776 Chr.16: 67289456-67289456 on Build GRCh38
rs977230693 Chr.16: 67289395-67289395 on Build GRCh38
rs958387619 Chr.16: 67289143-67289143 on Build GRCh38
rs989996999 Chr.16: 67289443-67289443 on Build GRCh38
rs758048004 Chr.16: 67289558-67289558 on Build GRCh38
rs923331497 Chr.16: 67289495-67289495 on Build GRCh38
rs754491719 Chr.16: 67289543-67289543 on Build GRCh38
rs926050374 Chr.16: 67289240-67289240 on Build GRCh38
rs753786814 Chr.16: 67289539-67289539 on Build GRCh38
rs977534403 Chr.16: 67289213-67289213 on Build GRCh38
rs369315078 Chr.16: 67289535-67289535 on Build GRCh38
rs887991984 Chr.16: 67289299-67289299 on Build GRCh38
rs375692277 Chr.16: 67289378-67289378 on Build GRCh38
rs1037165825 Chr.16: 67289407-67289407 on Build GRCh38
rs1047830551 Chr.16: 67289567-67289567 on Build GRCh38
rs866404841 Chr.16: 67289483-67289483 on Build GRCh38
rs764346258 Chr.16: 67289534-67289534 on Build GRCh38
rs1028206662 Chr.16: 67289167-67289167 on Build GRCh38
rs975962885 Chr.16: 67289488-67289488 on Build GRCh38
rs192023672 Chr.16: 67289276-67289276 on Build GRCh38
rs56342614 Chr.16: 67289408-67289408 on Build GRCh38
rs950222659 Chr.16: 67289283-67289283 on Build GRCh38
rs559217874 Chr.16: 67289160-67289160 on Build GRCh38
rs775777593 Chr.16: 67289241-67289241 on Build GRCh38

View Full Product Details