Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67291760-67292264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD19
Entrez Gene ID
146212
Gene Name
potassium channel tetramerization domain containing 19
Gene Aliases
-
UniGene
Hs.299127
Gene Chromosome Location
Chr.16: 67289490-67326758 on Build GRCh38
Transcripts
NM_001100915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4261147 c.2293C>A p.H765N Chr.16: 67291763-67291763 on Build GRCh38
4694813 c.2219-1G>A p.? Chr.16: 67291838-67291838 on Build GRCh38
5497275 c.2283C>G p.L761L Chr.16: 67291773-67291773 on Build GRCh38
3511005 c.2262G>A p.V754V Chr.16: 67291794-67291794 on Build GRCh38
1600109 c.2248G>A p.E750K Chr.16: 67291808-67291808 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779644101 Chr.16: 67291871-67291871 on Build GRCh38
rs558865237 Chr.16: 67292205-67292205 on Build GRCh38
rs910665569 Chr.16: 67292046-67292046 on Build GRCh38
rs1026615854 Chr.16: 67292031-67292031 on Build GRCh38
rs1043764364 Chr.16: 67292190-67292190 on Build GRCh38
rs943558634 Chr.16: 67291872-67291872 on Build GRCh38
rs1042992304 Chr.16: 67291892-67291892 on Build GRCh38
rs570829896 Chr.16: 67291882-67291882 on Build GRCh38
rs1042676679 Chr.16: 67292119-67292119 on Build GRCh38
rs894115824 Chr.16: 67292155-67292155 on Build GRCh38
rs16957291 Chr.16: 67292199-67292199 on Build GRCh38
rs933270356 Chr.16: 67291787-67291787 on Build GRCh38
rs755827805 Chr.16: 67291873-67291873 on Build GRCh38
rs1013192902 Chr.16: 67291989-67291989 on Build GRCh38
rs964778376 Chr.16: 67292087-67292087 on Build GRCh38
rs763705956 Chr.16: 67291872-67291873 on Build GRCh38
rs554679215 Chr.16: 67292080-67292080 on Build GRCh38
rs946587150 Chr.16: 67292188-67292188 on Build GRCh38
rs928215231 Chr.16: 67292131-67292131 on Build GRCh38
rs777881660 Chr.16: 67292212-67292212 on Build GRCh38
rs779191551 Chr.16: 67292156-67292156 on Build GRCh38
rs765489440 Chr.16: 67291827-67291827 on Build GRCh38
rs778875473 Chr.16: 67291852-67291852 on Build GRCh38
rs755085886 Chr.16: 67292133-67292133 on Build GRCh38
rs558029716 Chr.16: 67292177-67292177 on Build GRCh38
rs566535932 Chr.16: 67292089-67292089 on Build GRCh38
rs987468541 Chr.16: 67292035-67292035 on Build GRCh38
rs191926403 Chr.16: 67291873-67291873 on Build GRCh38
rs761695767 Chr.16: 67291836-67291836 on Build GRCh38
rs902862230 Chr.16: 67291901-67291901 on Build GRCh38
rs373574510 Chr.16: 67291784-67291784 on Build GRCh38
rs530127622 Chr.16: 67291798-67291798 on Build GRCh38
rs16957289 Chr.16: 67291808-67291808 on Build GRCh38
rs373185706 Chr.16: 67291809-67291809 on Build GRCh38
rs772629445 Chr.16: 67291796-67291796 on Build GRCh38
rs1026276798 Chr.16: 67292233-67292233 on Build GRCh38
rs920240084 Chr.16: 67292041-67292041 on Build GRCh38
rs894210691 Chr.16: 67291976-67291976 on Build GRCh38
rs550168023 Chr.16: 67291963-67291963 on Build GRCh38
rs767393945 Chr.16: 67291859-67291859 on Build GRCh38
rs1056711975 Chr.16: 67292144-67292144 on Build GRCh38
rs1016111063 Chr.16: 67292020-67292020 on Build GRCh38
rs1056848952 Chr.16: 67291947-67291947 on Build GRCh38
rs751413879 Chr.16: 67291833-67291833 on Build GRCh38
rs1052384805 Chr.16: 67292154-67292154 on Build GRCh38
rs775871276 Chr.16: 67292112-67292112 on Build GRCh38
rs989944750 Chr.16: 67292110-67292110 on Build GRCh38
rs1012687678 Chr.16: 67292165-67292165 on Build GRCh38
rs535965671 Chr.16: 67292032-67292032 on Build GRCh38
rs185877737 Chr.16: 67292108-67292108 on Build GRCh38
rs555366722 Chr.16: 67292030-67292030 on Build GRCh38
rs1002483760 Chr.16: 67291909-67291909 on Build GRCh38
rs752350683 Chr.16: 67291868-67291869 on Build GRCh38
rs750350954 Chr.16: 67291860-67291860 on Build GRCh38
rs531932328 Chr.16: 67291962-67291962 on Build GRCh38
rs996909660 Chr.16: 67291884-67291884 on Build GRCh38
rs755954569 Chr.16: 67291867-67291867 on Build GRCh38
rs114108483 Chr.16: 67291785-67291785 on Build GRCh38
rs969114174 Chr.16: 67292066-67292067 on Build GRCh38
rs927749363 Chr.16: 67291834-67291834 on Build GRCh38
rs1004508228 Chr.16: 67291938-67291938 on Build GRCh38
rs980893769 Chr.16: 67292090-67292090 on Build GRCh38
rs1037818337 Chr.16: 67292184-67292184 on Build GRCh38
rs575216580 Chr.16: 67292061-67292061 on Build GRCh38
rs1016242316 Chr.16: 67292000-67292000 on Build GRCh38
rs963521416 Chr.16: 67292027-67292027 on Build GRCh38
rs560498763 Chr.16: 67291816-67291816 on Build GRCh38
rs377131607 Chr.16: 67291818-67291818 on Build GRCh38
rs181038940 Chr.16: 67291881-67291881 on Build GRCh38
rs544548917 Chr.16: 67292111-67292111 on Build GRCh38

View Full Product Details