Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67294496-67294998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD19
Entrez Gene ID
146212
Gene Name
potassium channel tetramerization domain containing 19
Gene Aliases
-
UniGene
Hs.299127
Gene Chromosome Location
Chr.16: 67289490-67326758 on Build GRCh38
Transcripts
NM_001100915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3511010 c.1507G>A p.E503K Chr.16: 67294663-67294663 on Build GRCh38
972405 c.1495G>T p.E499* Chr.16: 67294675-67294675 on Build GRCh38
1211689 c.1491G>T p.E497D Chr.16: 67294679-67294679 on Build GRCh38
5913770 c.1489G>A p.E497K Chr.16: 67294681-67294681 on Build GRCh38
312235 c.1561C>G p.L521V Chr.16: 67294609-67294609 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956574701 Chr.16: 67294802-67294802 on Build GRCh38
rs750634015 Chr.16: 67294606-67294606 on Build GRCh38
rs369454332 Chr.16: 67294728-67294728 on Build GRCh38
rs760756494 Chr.16: 67294607-67294607 on Build GRCh38
rs762300525 Chr.16: 67294819-67294819 on Build GRCh38
rs751492532 Chr.16: 67294934-67294934 on Build GRCh38
rs770236578 Chr.16: 67294968-67294971 on Build GRCh38
rs541694257 Chr.16: 67294928-67294928 on Build GRCh38
rs3868143 Chr.16: 67294550-67294550 on Build GRCh38
rs1055808218 Chr.16: 67294842-67294842 on Build GRCh38
rs537854501 Chr.16: 67294784-67294784 on Build GRCh38
rs763454281 Chr.16: 67294555-67294555 on Build GRCh38
rs769216696 Chr.16: 67294695-67294695 on Build GRCh38
rs139855834 Chr.16: 67294750-67294750 on Build GRCh38
rs752476629 Chr.16: 67294633-67294633 on Build GRCh38
rs749638733 Chr.16: 67294685-67294685 on Build GRCh38
rs775790651 Chr.16: 67294553-67294553 on Build GRCh38
rs200748914 Chr.16: 67294725-67294725 on Build GRCh38
rs1014381448 Chr.16: 67294874-67294874 on Build GRCh38
rs752601467 Chr.16: 67294745-67294745 on Build GRCh38
rs372562104 Chr.16: 67294781-67294781 on Build GRCh38
rs770968726 Chr.16: 67294549-67294551 on Build GRCh38
rs757095239 Chr.16: 67294935-67294935 on Build GRCh38
rs774804881 Chr.16: 67294698-67294698 on Build GRCh38
rs371235068 Chr.16: 67294559-67294559 on Build GRCh38
rs767669499 Chr.16: 67294601-67294601 on Build GRCh38
rs1048187615 Chr.16: 67294751-67294751 on Build GRCh38
rs760770773 Chr.16: 67294590-67294591 on Build GRCh38
rs374183561 Chr.16: 67294954-67294954 on Build GRCh38
rs1000625039 Chr.16: 67294753-67294753 on Build GRCh38
rs766653808 Chr.16: 67294732-67294732 on Build GRCh38
rs1041359417 Chr.16: 67294717-67294717 on Build GRCh38
rs760419648 Chr.16: 67294960-67294961 on Build GRCh38
rs775860778 Chr.16: 67294947-67294948 on Build GRCh38
rs749842965 Chr.16: 67294953-67294953 on Build GRCh38
rs780787043 Chr.16: 67294952-67294952 on Build GRCh38
rs368801418 Chr.16: 67294659-67294659 on Build GRCh38
rs774471557 Chr.16: 67294560-67294560 on Build GRCh38
rs531637330 Chr.16: 67294657-67294659 on Build GRCh38
rs777843756 Chr.16: 67294635-67294635 on Build GRCh38
rs895799118 Chr.16: 67294859-67294859 on Build GRCh38
rs765157829 Chr.16: 67294613-67294613 on Build GRCh38
rs769262453 Chr.16: 67294675-67294675 on Build GRCh38
rs1013352304 Chr.16: 67294831-67294831 on Build GRCh38
rs774771292 Chr.16: 67294622-67294622 on Build GRCh38
rs765737902 Chr.16: 67294658-67294658 on Build GRCh38
rs780407607 Chr.16: 67294679-67294679 on Build GRCh38
rs201529276 Chr.16: 67294527-67294527 on Build GRCh38
rs948071698 Chr.16: 67294520-67294520 on Build GRCh38
rs12935240 Chr.16: 67294827-67294827 on Build GRCh38
rs553717381 Chr.16: 67294932-67294932 on Build GRCh38
rs758427479 Chr.16: 67294634-67294634 on Build GRCh38
rs1028672225 Chr.16: 67294735-67294735 on Build GRCh38
rs772387836 Chr.16: 67294733-67294733 on Build GRCh38
rs756760897 Chr.16: 67294655-67294655 on Build GRCh38
rs557828022 Chr.16: 67294975-67294975 on Build GRCh38
rs761944170 Chr.16: 67294578-67294578 on Build GRCh38
rs761844144 Chr.16: 67294705-67294705 on Build GRCh38
rs539466701 Chr.16: 67294529-67294529 on Build GRCh38
rs183938011 Chr.16: 67294528-67294528 on Build GRCh38
rs111842396 Chr.16: 67294585-67294585 on Build GRCh38
rs371199457 Chr.16: 67294549-67294549 on Build GRCh38
rs555919595 Chr.16: 67294878-67294878 on Build GRCh38
rs751098440 Chr.16: 67294643-67294643 on Build GRCh38
rs374798005 Chr.16: 67294657-67294657 on Build GRCh38
rs1001819751 Chr.16: 67294791-67294791 on Build GRCh38
rs149806855 Chr.16: 67294903-67294903 on Build GRCh38
rs931431558 Chr.16: 67294535-67294535 on Build GRCh38

View Full Product Details