Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68678487-68678988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH3
Entrez Gene ID
1001
Gene Name
cadherin 3
Gene Aliases
CDHP, HJMD, PCAD
UniGene
Hs.191842
Gene Chromosome Location
Chr.16: 68644820-68727573 on Build GRCh38
Transcripts
NM_001317195.1 NM_001317196.1 NM_001793.5 XM_011522800.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4670954 c.543T>A p.Y181* Chr.16: 68678653-68678653 on Build GRCh38
6625064 c.558C>T p.H186H Chr.16: 68678773-68678773 on Build GRCh38
1200487 c.502T>A p.L168M Chr.16: 68678612-68678612 on Build GRCh38
3787203 c.518T>C p.L173P Chr.16: 68678628-68678628 on Build GRCh38
6625054 c.402T>C p.N134N Chr.16: 68678512-68678512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758051035 Chr.16: 68678907-68678907 on Build GRCh38
rs756715671 Chr.16: 68678931-68678931 on Build GRCh38
rs764034100 Chr.16: 68678777-68678777 on Build GRCh38
rs759791344 Chr.16: 68678611-68678611 on Build GRCh38
rs765171740 Chr.16: 68678901-68678901 on Build GRCh38
rs370978420 Chr.16: 68678750-68678750 on Build GRCh38
rs765259854 Chr.16: 68678955-68678955 on Build GRCh38
rs900672615 Chr.16: 68678583-68678583 on Build GRCh38
rs767499381 Chr.16: 68678689-68678689 on Build GRCh38
rs377177844 Chr.16: 68678945-68678945 on Build GRCh38
rs368291942 Chr.16: 68678834-68678834 on Build GRCh38
rs763136509 Chr.16: 68678869-68678869 on Build GRCh38
rs752829567 Chr.16: 68678633-68678633 on Build GRCh38
rs754145153 Chr.16: 68678833-68678833 on Build GRCh38
rs752739867 Chr.16: 68678553-68678553 on Build GRCh38
rs371682154 Chr.16: 68678877-68678877 on Build GRCh38
rs886052230 Chr.16: 68678906-68678906 on Build GRCh38
rs752489424 Chr.16: 68678958-68678958 on Build GRCh38
rs566610716 Chr.16: 68678643-68678643 on Build GRCh38
rs778699617 Chr.16: 68678608-68678610 on Build GRCh38
rs752509142 Chr.16: 68678903-68678903 on Build GRCh38
rs750898870 Chr.16: 68678747-68678747 on Build GRCh38
rs148063369 Chr.16: 68678797-68678797 on Build GRCh38
rs548340277 Chr.16: 68678822-68678822 on Build GRCh38
rs761941770 Chr.16: 68678876-68678876 on Build GRCh38
rs770108268 Chr.16: 68678857-68678857 on Build GRCh38
rs62057764 Chr.16: 68678630-68678630 on Build GRCh38
rs952095453 Chr.16: 68678767-68678767 on Build GRCh38
rs747666407 Chr.16: 68678731-68678731 on Build GRCh38
rs112542580 Chr.16: 68678759-68678759 on Build GRCh38
rs777186958 Chr.16: 68678828-68678828 on Build GRCh38
rs757142746 Chr.16: 68678599-68678600 on Build GRCh38
rs751041174 Chr.16: 68678916-68678916 on Build GRCh38
rs768894285 Chr.16: 68678871-68678871 on Build GRCh38
rs779790678 Chr.16: 68678567-68678567 on Build GRCh38
rs1022953871 Chr.16: 68678601-68678601 on Build GRCh38
rs779764715 Chr.16: 68678711-68678711 on Build GRCh38
rs773789550 Chr.16: 68678737-68678737 on Build GRCh38
rs756048455 Chr.16: 68678793-68678793 on Build GRCh38
rs761132411 Chr.16: 68678588-68678588 on Build GRCh38
rs754762082 Chr.16: 68678807-68678807 on Build GRCh38
rs769411955 Chr.16: 68678566-68678566 on Build GRCh38
rs373057862 Chr.16: 68678938-68678938 on Build GRCh38
rs8049247 Chr.16: 68678827-68678827 on Build GRCh38
rs780974578 Chr.16: 68678562-68678562 on Build GRCh38
rs746232602 Chr.16: 68678851-68678851 on Build GRCh38
rs777030574 Chr.16: 68678610-68678610 on Build GRCh38
rs1000998850 Chr.16: 68678641-68678641 on Build GRCh38
rs774363149 Chr.16: 68678872-68678872 on Build GRCh38
rs377449804 Chr.16: 68678675-68678675 on Build GRCh38
rs148231483 Chr.16: 68678558-68678558 on Build GRCh38
rs758410545 Chr.16: 68678648-68678648 on Build GRCh38
rs777466922 Chr.16: 68678913-68678913 on Build GRCh38
rs770438078 Chr.16: 68678568-68678568 on Build GRCh38
rs201590604 Chr.16: 68678721-68678721 on Build GRCh38
rs373992293 Chr.16: 68678949-68678949 on Build GRCh38
rs902743192 Chr.16: 68678617-68678617 on Build GRCh38
rs546779028 Chr.16: 68678572-68678572 on Build GRCh38
rs765454584 Chr.16: 68678762-68678762 on Build GRCh38
rs765438205 Chr.16: 68678613-68678613 on Build GRCh38
rs368814334 Chr.16: 68678742-68678742 on Build GRCh38
rs897013928 Chr.16: 68678952-68678952 on Build GRCh38
rs781586467 Chr.16: 68678936-68678936 on Build GRCh38
rs775627086 Chr.16: 68678773-68678773 on Build GRCh38
rs749189312 Chr.16: 68678572-68678572 on Build GRCh38
rs376349856 Chr.16: 68678698-68678698 on Build GRCh38
rs140049723 Chr.16: 68678584-68678584 on Build GRCh38
rs902688565 Chr.16: 68678910-68678910 on Build GRCh38
rs771435632 Chr.16: 68678746-68678746 on Build GRCh38
rs771441605 Chr.16: 68678732-68678732 on Build GRCh38
rs766601956 Chr.16: 68678589-68678589 on Build GRCh38
rs759427575 Chr.16: 68678885-68678885 on Build GRCh38
rs768402984 Chr.16: 68678570-68678570 on Build GRCh38
rs76528263 Chr.16: 68678691-68678691 on Build GRCh38
rs61730078 Chr.16: 68678761-68678761 on Build GRCh38
rs773525226 Chr.16: 68678585-68678585 on Build GRCh38
rs1000199470 Chr.16: 68678688-68678688 on Build GRCh38
rs200463078 Chr.16: 68678547-68678547 on Build GRCh38
rs532947095 Chr.16: 68678533-68678533 on Build GRCh38
rs1026712146 Chr.16: 68678755-68678755 on Build GRCh38
rs753785979 Chr.16: 68678883-68678883 on Build GRCh38
rs200737028 Chr.16: 68678520-68678520 on Build GRCh38
rs775715119 Chr.16: 68678864-68678864 on Build GRCh38
rs745692866 Chr.16: 68678563-68678563 on Build GRCh38
rs772229250 Chr.16: 68678946-68678946 on Build GRCh38
rs535792399 Chr.16: 68678754-68678754 on Build GRCh38
rs115879924 Chr.16: 68678788-68678788 on Build GRCh38
rs755378442 Chr.16: 68678935-68678935 on Build GRCh38
rs753744580 Chr.16: 68678798-68678798 on Build GRCh38
rs749062682 Chr.16: 68678712-68678712 on Build GRCh38
rs576611511 Chr.16: 68678717-68678717 on Build GRCh38
rs369822985 Chr.16: 68678934-68678934 on Build GRCh38
rs755070586 Chr.16: 68678527-68678527 on Build GRCh38
rs946811595 Chr.16: 68678894-68678894 on Build GRCh38

View Full Product Details