Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 69464272-69464804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5B
Entrez Gene ID
80777
Gene Name
cytochrome b5 type B
Gene Aliases
CYB5-M, CYPB5M, OMB5
UniGene
Hs.461131
Gene Chromosome Location
Chr.16: 69424595-69466264 on Build GRCh38
Transcripts
NM_030579.2

SNP Detail

SNP ID Chromosome Location
rs921963907 Chr.16: 69464486-69464486 on Build GRCh38
rs534941397 Chr.16: 69464598-69464598 on Build GRCh38
rs779001734 Chr.16: 69464425-69464425 on Build GRCh38
rs923735396 Chr.16: 69464527-69464527 on Build GRCh38
rs569862808 Chr.16: 69464352-69464352 on Build GRCh38
rs897432328 Chr.16: 69464353-69464353 on Build GRCh38
rs897379960 Chr.16: 69464557-69464557 on Build GRCh38
rs1011925953 Chr.16: 69464338-69464338 on Build GRCh38
rs1041249442 Chr.16: 69464549-69464549 on Build GRCh38
rs1019555305 Chr.16: 69464719-69464719 on Build GRCh38
rs748095531 Chr.16: 69464528-69464528 on Build GRCh38
rs992686109 Chr.16: 69464436-69464436 on Build GRCh38
rs966418035 Chr.16: 69464420-69464420 on Build GRCh38
rs185557458 Chr.16: 69464579-69464579 on Build GRCh38
rs551405437 Chr.16: 69464354-69464354 on Build GRCh38
rs911377723 Chr.16: 69464542-69464542 on Build GRCh38
rs569554617 Chr.16: 69464524-69464524 on Build GRCh38
rs995218543 Chr.16: 69464645-69464645 on Build GRCh38
rs556682342 Chr.16: 69464744-69464744 on Build GRCh38
rs1001614555 Chr.16: 69464594-69464594 on Build GRCh38
rs1005070962 Chr.16: 69464701-69464701 on Build GRCh38
rs1056754975 Chr.16: 69464544-69464544 on Build GRCh38
rs911618212 Chr.16: 69464746-69464746 on Build GRCh38
rs1028948214 Chr.16: 69464736-69464736 on Build GRCh38
rs118060426 Chr.16: 69464724-69464724 on Build GRCh38
rs913127323 Chr.16: 69464451-69464451 on Build GRCh38
rs898236550 Chr.16: 69464349-69464349 on Build GRCh38
rs777410273 Chr.16: 69464575-69464575 on Build GRCh38
rs888479921 Chr.16: 69464593-69464593 on Build GRCh38
rs1050203839 Chr.16: 69464527-69464528 on Build GRCh38
rs979525196 Chr.16: 69464505-69464505 on Build GRCh38
rs978417064 Chr.16: 69464460-69464460 on Build GRCh38
rs967712151 Chr.16: 69464454-69464454 on Build GRCh38
rs374289201 Chr.16: 69464756-69464756 on Build GRCh38
rs755055055 Chr.16: 69464381-69464381 on Build GRCh38
rs984076052 Chr.16: 69464386-69464386 on Build GRCh38
rs886578969 Chr.16: 69464654-69464654 on Build GRCh38
rs931745835 Chr.16: 69464492-69464492 on Build GRCh38
rs112630491 Chr.16: 69464650-69464650 on Build GRCh38
rs565965592 Chr.16: 69464384-69464385 on Build GRCh38
rs766063848 Chr.16: 69464620-69464620 on Build GRCh38
rs552669968 Chr.16: 69464619-69464619 on Build GRCh38
rs190324371 Chr.16: 69464580-69464580 on Build GRCh38
rs953190553 Chr.16: 69464742-69464742 on Build GRCh38
rs543877448 Chr.16: 69464385-69464385 on Build GRCh38
rs1014014117 Chr.16: 69464639-69464639 on Build GRCh38
rs1056385026 Chr.16: 69464317-69464317 on Build GRCh38
rs999845909 Chr.16: 69464310-69464310 on Build GRCh38
rs758272691 Chr.16: 69464537-69464537 on Build GRCh38
rs1023780188 Chr.16: 69464346-69464346 on Build GRCh38
rs979450156 Chr.16: 69464763-69464763 on Build GRCh38
rs946794586 Chr.16: 69464489-69464489 on Build GRCh38
rs979125563 Chr.16: 69464678-69464678 on Build GRCh38
rs12919617 Chr.16: 69464332-69464332 on Build GRCh38
rs35582283 Chr.16: 69464658-69464658 on Build GRCh38
rs574997908 Chr.16: 69464776-69464776 on Build GRCh38
rs975433596 Chr.16: 69464728-69464728 on Build GRCh38
rs1019206134 Chr.16: 69464397-69464397 on Build GRCh38
rs35256922 Chr.16: 69464772-69464772 on Build GRCh38
rs538492446 Chr.16: 69464568-69464568 on Build GRCh38
rs1027824141 Chr.16: 69464379-69464379 on Build GRCh38
rs34073253 Chr.16: 69464726-69464726 on Build GRCh38

View Full Product Details