Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70315712-70316211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX19B
Entrez Gene ID
11269
Gene Name
DEAD-box helicase 19B
Gene Aliases
DBP5, DDX19, RNAh
UniGene
Hs.221761
Gene Chromosome Location
Chr.16: 70289767-70335213 on Build GRCh38
Transcripts
NM_001014449.2 NM_001014451.2 NM_001257172.1 NM_001257173.1 NM_001257174.1 NM_001257175.1 NM_007242.5 XM_006721127.2 XM_011522832.2 XM_011522833.2 XM_011522834.2 XM_017022890.1 NR_039997.1

Gene Symbol
LOC100506083
Entrez Gene ID
100506083
Gene Name
uncharacterized LOC100506083
Gene Aliases
-
UniGene
Hs.635008
Gene Chromosome Location
Chr.16: 70315640-70346747 on Build GRCh38
Transcripts
NM_001014449.2 NM_001014451.2 NM_001257172.1 NM_001257173.1 NM_001257174.1 NM_001257175.1 NM_007242.5 XM_006721127.2 XM_011522832.2 XM_011522833.2 XM_011522834.2 XM_017022890.1 NR_039997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7400734 c.266A>G p.Y89C Chr.16: 70316074-70316074 on Build GRCh38
6641591 c.243G>T p.Q81H Chr.16: 70316051-70316051 on Build GRCh38
4062315 c.293G>A p.R98Q Chr.16: 70316101-70316101 on Build GRCh38
6641592 c.243G>T p.Q81H Chr.16: 70316051-70316051 on Build GRCh38
3272871 c.244C>T p.R82W Chr.16: 70316052-70316052 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565863200 Chr.16: 70315740-70315740 on Build GRCh38
rs758652714 Chr.16: 70316024-70316024 on Build GRCh38
rs971982769 Chr.16: 70315936-70315936 on Build GRCh38
rs564069580 Chr.16: 70316164-70316164 on Build GRCh38
rs748767458 Chr.16: 70315933-70315933 on Build GRCh38
rs768398748 Chr.16: 70316096-70316096 on Build GRCh38
rs1028610212 Chr.16: 70315914-70315914 on Build GRCh38
rs763071669 Chr.16: 70316134-70316134 on Build GRCh38
rs556520525 Chr.16: 70315872-70315872 on Build GRCh38
rs927211074 Chr.16: 70315752-70315752 on Build GRCh38
rs750390204 Chr.16: 70315940-70315940 on Build GRCh38
rs985021629 Chr.16: 70315873-70315873 on Build GRCh38
rs752901044 Chr.16: 70315988-70315988 on Build GRCh38
rs148811871 Chr.16: 70316102-70316102 on Build GRCh38
rs764396664 Chr.16: 70316053-70316053 on Build GRCh38
rs980502763 Chr.16: 70316148-70316148 on Build GRCh38
rs927224112 Chr.16: 70316169-70316169 on Build GRCh38
rs755584161 Chr.16: 70316073-70316073 on Build GRCh38
rs1050420827 Chr.16: 70316052-70316052 on Build GRCh38
rs779459626 Chr.16: 70316078-70316078 on Build GRCh38
rs545473837 Chr.16: 70316158-70316158 on Build GRCh38
rs542082886 Chr.16: 70315929-70315929 on Build GRCh38
rs764272089 Chr.16: 70316138-70316138 on Build GRCh38
rs971016593 Chr.16: 70315836-70315836 on Build GRCh38
rs57279408 Chr.16: 70315822-70315822 on Build GRCh38
rs370913054 Chr.16: 70315950-70315950 on Build GRCh38
rs765322820 Chr.16: 70316120-70316120 on Build GRCh38
rs775458252 Chr.16: 70316131-70316131 on Build GRCh38
rs16970609 Chr.16: 70315915-70315915 on Build GRCh38
rs538095598 Chr.16: 70315863-70315863 on Build GRCh38
rs777696853 Chr.16: 70316070-70316070 on Build GRCh38
rs766346087 Chr.16: 70315975-70315975 on Build GRCh38
rs754632716 Chr.16: 70315760-70315760 on Build GRCh38
rs760538686 Chr.16: 70316105-70316105 on Build GRCh38
rs1033139229 Chr.16: 70316020-70316020 on Build GRCh38
rs930515345 Chr.16: 70315754-70315754 on Build GRCh38
rs774184558 Chr.16: 70316097-70316097 on Build GRCh38
rs3207733 Chr.16: 70316118-70316118 on Build GRCh38
rs757604806 Chr.16: 70316155-70316155 on Build GRCh38
rs751901098 Chr.16: 70316055-70316055 on Build GRCh38
rs747948624 Chr.16: 70316100-70316100 on Build GRCh38
rs773147315 Chr.16: 70316103-70316103 on Build GRCh38
rs111644407 Chr.16: 70315738-70315738 on Build GRCh38
rs372853287 Chr.16: 70315847-70315847 on Build GRCh38
rs886193425 Chr.16: 70315791-70315791 on Build GRCh38
rs756944542 Chr.16: 70315966-70315967 on Build GRCh38
rs746374309 Chr.16: 70316067-70316067 on Build GRCh38
rs996224292 Chr.16: 70315893-70315893 on Build GRCh38
rs188647481 Chr.16: 70315846-70315846 on Build GRCh38
rs781720740 Chr.16: 70316063-70316063 on Build GRCh38
rs1055781 Chr.16: 70316051-70316051 on Build GRCh38
rs1053997387 Chr.16: 70315747-70315747 on Build GRCh38
rs772047383 Chr.16: 70316101-70316101 on Build GRCh38
rs757659949 Chr.16: 70316061-70316061 on Build GRCh38
rs748899169 Chr.16: 70316084-70316084 on Build GRCh38
rs892661226 Chr.16: 70316172-70316172 on Build GRCh38
rs751846263 Chr.16: 70316143-70316143 on Build GRCh38
rs1009851866 Chr.16: 70315768-70315769 on Build GRCh38
rs770625356 Chr.16: 70316081-70316081 on Build GRCh38
rs765248759 Chr.16: 70315983-70315983 on Build GRCh38

View Full Product Details