Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8228411-8228915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTC1
Entrez Gene ID
80169
Gene Name
CTS telomere maintenance complex component 1
Gene Aliases
AAF-132, AAF132, C17orf68, CRMCC, tmp494178
UniGene
Hs.156055
Gene Chromosome Location
Chr.17: 8224821-8248095 on Build GRCh38
Transcripts
NM_025099.5 NR_046431.1 XM_006721577.3 XM_006721578.3 XM_011524010.2 XM_011524011.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7140003 c.3504C>A p.I1168I Chr.17: 8228513-8228513 on Build GRCh38
7140004 c.3494C>A p.P1165Q Chr.17: 8228523-8228523 on Build GRCh38
4627568 c.3369G>A p.G1123G Chr.17: 8228745-8228745 on Build GRCh38
1246574 c.3458G>A p.R1153H Chr.17: 8228559-8228559 on Build GRCh38
5464241 c.3473C>A p.S1158Y Chr.17: 8228544-8228544 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752759103 Chr.17: 8228636-8228636 on Build GRCh38
rs901201927 Chr.17: 8228449-8228449 on Build GRCh38
rs945189616 Chr.17: 8228730-8228730 on Build GRCh38
rs745512137 Chr.17: 8228591-8228591 on Build GRCh38
rs372789589 Chr.17: 8228809-8228809 on Build GRCh38
rs754428780 Chr.17: 8228654-8228654 on Build GRCh38
rs755032241 Chr.17: 8228467-8228467 on Build GRCh38
rs773769861 Chr.17: 8228488-8228488 on Build GRCh38
rs373500129 Chr.17: 8228681-8228681 on Build GRCh38
rs201455840 Chr.17: 8228592-8228592 on Build GRCh38
rs758196202 Chr.17: 8228472-8228474 on Build GRCh38
rs774108596 Chr.17: 8228583-8228583 on Build GRCh38
rs780105987 Chr.17: 8228454-8228454 on Build GRCh38
rs374306002 Chr.17: 8228765-8228765 on Build GRCh38
rs375329347 Chr.17: 8228805-8228805 on Build GRCh38
rs376216450 Chr.17: 8228604-8228604 on Build GRCh38
rs766537691 Chr.17: 8228523-8228523 on Build GRCh38
rs202226502 Chr.17: 8228788-8228788 on Build GRCh38
rs536983035 Chr.17: 8228833-8228833 on Build GRCh38
rs751757398 Chr.17: 8228537-8228537 on Build GRCh38
rs748604135 Chr.17: 8228478-8228478 on Build GRCh38
rs770745414 Chr.17: 8228495-8228495 on Build GRCh38
rs769560758 Chr.17: 8228717-8228717 on Build GRCh38
rs779158760 Chr.17: 8228656-8228656 on Build GRCh38
rs368562418 Chr.17: 8228845-8228845 on Build GRCh38
rs201885165 Chr.17: 8228867-8228867 on Build GRCh38
rs551437595 Chr.17: 8228489-8228489 on Build GRCh38
rs543241073 Chr.17: 8228777-8228777 on Build GRCh38
rs755212243 Chr.17: 8228614-8228614 on Build GRCh38
rs766968528 Chr.17: 8228507-8228507 on Build GRCh38
rs370564569 Chr.17: 8228774-8228774 on Build GRCh38
rs555242943 Chr.17: 8228820-8228820 on Build GRCh38
rs767679831 Chr.17: 8228748-8228748 on Build GRCh38
rs755058193 Chr.17: 8228612-8228612 on Build GRCh38
rs373723138 Chr.17: 8228472-8228472 on Build GRCh38
rs755014847 Chr.17: 8228538-8228538 on Build GRCh38
rs746887979 Chr.17: 8228530-8228531 on Build GRCh38
rs745525096 Chr.17: 8228660-8228660 on Build GRCh38
rs769453932 Chr.17: 8228702-8228705 on Build GRCh38
rs760660240 Chr.17: 8228602-8228602 on Build GRCh38
rs760547081 Chr.17: 8228513-8228513 on Build GRCh38
rs759771248 Chr.17: 8228745-8228745 on Build GRCh38
rs370845759 Chr.17: 8228754-8228754 on Build GRCh38
rs779335414 Chr.17: 8228784-8228784 on Build GRCh38
rs773036519 Chr.17: 8228834-8228834 on Build GRCh38
rs756645315 Chr.17: 8228571-8228571 on Build GRCh38
rs772946559 Chr.17: 8228733-8228733 on Build GRCh38
rs199473681 Chr.17: 8228591-8228592 on Build GRCh38
rs952574848 Chr.17: 8228525-8228525 on Build GRCh38
rs776482338 Chr.17: 8228699-8228699 on Build GRCh38
rs748145891 Chr.17: 8228732-8228732 on Build GRCh38
rs780812864 Chr.17: 8228646-8228651 on Build GRCh38
rs201644249 Chr.17: 8228576-8228576 on Build GRCh38
rs778249594 Chr.17: 8228573-8228573 on Build GRCh38
rs1015803777 Chr.17: 8228499-8228499 on Build GRCh38
rs770137845 Chr.17: 8228485-8228485 on Build GRCh38
rs780029888 Chr.17: 8228482-8228483 on Build GRCh38
rs201124470 Chr.17: 8228868-8228868 on Build GRCh38
rs778378098 Chr.17: 8228643-8228643 on Build GRCh38
rs760797988 Chr.17: 8228760-8228760 on Build GRCh38
rs371452614 Chr.17: 8228667-8228667 on Build GRCh38
rs757422154 Chr.17: 8228459-8228460 on Build GRCh38
rs571554235 Chr.17: 8228554-8228554 on Build GRCh38
rs757629670 Chr.17: 8228655-8228655 on Build GRCh38
rs761553012 Chr.17: 8228611-8228611 on Build GRCh38
rs748072446 Chr.17: 8228828-8228828 on Build GRCh38
rs374284337 Chr.17: 8228783-8228783 on Build GRCh38
rs1015469098 Chr.17: 8228570-8228570 on Build GRCh38
rs1053378254 Chr.17: 8228479-8228479 on Build GRCh38
rs762606006 Chr.17: 8228838-8228838 on Build GRCh38
rs779530804 Chr.17: 8228799-8228799 on Build GRCh38
rs773978472 Chr.17: 8228496-8228496 on Build GRCh38
rs756665311 Chr.17: 8228637-8228637 on Build GRCh38
rs556097361 Chr.17: 8228522-8228522 on Build GRCh38
rs377491661 Chr.17: 8228512-8228512 on Build GRCh38
rs369209127 Chr.17: 8228519-8228519 on Build GRCh38
rs961502392 Chr.17: 8228504-8228504 on Build GRCh38
rs554183453 Chr.17: 8228597-8228597 on Build GRCh38
rs564631876 Chr.17: 8228841-8228841 on Build GRCh38
rs776568961 Chr.17: 8228607-8228607 on Build GRCh38
rs915672693 Chr.17: 8228438-8228438 on Build GRCh38
rs369845758 Chr.17: 8228487-8228487 on Build GRCh38
rs746666288 Chr.17: 8228817-8228817 on Build GRCh38
rs1049155611 Chr.17: 8228850-8228850 on Build GRCh38
rs368576599 Chr.17: 8228686-8228686 on Build GRCh38
rs747705151 Chr.17: 8228658-8228665 on Build GRCh38
rs761749257 Chr.17: 8228710-8228710 on Build GRCh38
rs768723270 Chr.17: 8228532-8228533 on Build GRCh38
rs747137988 Chr.17: 8228460-8228460 on Build GRCh38
rs3027243 Chr.17: 8228450-8228450 on Build GRCh38
rs367822614 Chr.17: 8228797-8228797 on Build GRCh38
rs1014401373 Chr.17: 8228891-8228891 on Build GRCh38
rs926281886 Chr.17: 8228620-8228620 on Build GRCh38
rs748674791 Chr.17: 8228559-8228559 on Build GRCh38
rs759039342 Chr.17: 8228500-8228500 on Build GRCh38
rs936469878 Chr.17: 8228673-8228673 on Build GRCh38
rs771446414 Chr.17: 8228577-8228577 on Build GRCh38
rs780734573 Chr.17: 8228822-8228822 on Build GRCh38
rs1042810195 Chr.17: 8228471-8228471 on Build GRCh38
rs373326655 Chr.17: 8228649-8228649 on Build GRCh38
rs376214091 Chr.17: 8228698-8228698 on Build GRCh38
rs779053803 Chr.17: 8228466-8228466 on Build GRCh38
rs767630257 Chr.17: 8228618-8228618 on Build GRCh38
rs993081062 Chr.17: 8228517-8228517 on Build GRCh38
rs770791985 Chr.17: 8228862-8228862 on Build GRCh38
rs745645742 Chr.17: 8228471-8228471 on Build GRCh38
rs764256437 Chr.17: 8228872-8228872 on Build GRCh38

View Full Product Details