Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 34256919-34257448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL2
Entrez Gene ID
6347
Gene Name
C-C motif chemokine ligand 2
Gene Aliases
GDCF-2, HC11, HSMCR30, MCAF, MCP-1, MCP1, SCYA2, SMC-CF
UniGene
Hs.303649
Gene Chromosome Location
Chr.17: 34255277-34257203 on Build GRCh38
Transcripts
NM_002982.3

SNP Detail

SNP ID Chromosome Location
rs1023463271 Chr.17: 34257330-34257330 on Build GRCh38
rs999503024 Chr.17: 34257248-34257248 on Build GRCh38
rs1021166438 Chr.17: 34257414-34257414 on Build GRCh38
rs1007705149 Chr.17: 34257048-34257048 on Build GRCh38
rs925682245 Chr.17: 34257195-34257195 on Build GRCh38
rs958462033 Chr.17: 34257206-34257206 on Build GRCh38
rs1023811146 Chr.17: 34256964-34256964 on Build GRCh38
rs932035113 Chr.17: 34257128-34257128 on Build GRCh38
rs1040896429 Chr.17: 34257394-34257394 on Build GRCh38
rs767794054 Chr.17: 34257365-34257365 on Build GRCh38
rs528328195 Chr.17: 34257096-34257096 on Build GRCh38
rs2215182 Chr.17: 34257052-34257052 on Build GRCh38
rs2190971 Chr.17: 34257416-34257416 on Build GRCh38
rs1036249658 Chr.17: 34257254-34257254 on Build GRCh38
rs1010155939 Chr.17: 34257411-34257411 on Build GRCh38
rs908097228 Chr.17: 34257074-34257074 on Build GRCh38
rs1009817530 Chr.17: 34257063-34257063 on Build GRCh38
rs1049107397 Chr.17: 34257368-34257368 on Build GRCh38
rs1007628411 Chr.17: 34257382-34257382 on Build GRCh38
rs917087227 Chr.17: 34257241-34257241 on Build GRCh38
rs919173085 Chr.17: 34257285-34257285 on Build GRCh38
rs893688829 Chr.17: 34257324-34257324 on Build GRCh38
rs902001140 Chr.17: 34257397-34257397 on Build GRCh38
rs191032356 Chr.17: 34257051-34257051 on Build GRCh38
rs967035668 Chr.17: 34257108-34257108 on Build GRCh38
rs75284178 Chr.17: 34257109-34257109 on Build GRCh38
rs1021657455 Chr.17: 34257101-34257101 on Build GRCh38
rs978398994 Chr.17: 34257182-34257182 on Build GRCh38
rs777894448 Chr.17: 34257119-34257119 on Build GRCh38
rs75433364 Chr.17: 34257111-34257111 on Build GRCh38
rs770114076 Chr.17: 34257110-34257111 on Build GRCh38
rs561227164 Chr.17: 34257329-34257329 on Build GRCh38
rs186178409 Chr.17: 34256999-34256999 on Build GRCh38
rs751829134 Chr.17: 34257251-34257251 on Build GRCh38
rs759734916 Chr.17: 34257350-34257350 on Build GRCh38
rs532809366 Chr.17: 34257116-34257116 on Build GRCh38
rs889152442 Chr.17: 34257380-34257380 on Build GRCh38
rs551665616 Chr.17: 34257262-34257262 on Build GRCh38
rs1003236569 Chr.17: 34257364-34257364 on Build GRCh38
rs1051793904 Chr.17: 34257196-34257196 on Build GRCh38
rs890472835 Chr.17: 34257198-34257198 on Build GRCh38
rs991222752 Chr.17: 34257208-34257208 on Build GRCh38
rs536851983 Chr.17: 34257111-34257111 on Build GRCh38
rs969570378 Chr.17: 34257352-34257352 on Build GRCh38
rs552570373 Chr.17: 34257176-34257176 on Build GRCh38

View Full Product Details