Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 34577389-34577898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf102
Entrez Gene ID
400591
Gene Name
chromosome 17 open reading frame 102
Gene Aliases
-
UniGene
Hs.514090
Gene Chromosome Location
Chr.17: 34574123-34579369 on Build GRCh38
Transcripts
NM_207454.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6146185 c.476C>A p.A159D Chr.17: 34577555-34577555 on Build GRCh38
5438078 c.464G>A p.R155K Chr.17: 34577567-34577567 on Build GRCh38
259746 c.420G>A p.V140V Chr.17: 34577611-34577611 on Build GRCh38
5440493 c.504G>C p.*168Y Chr.17: 34577527-34577527 on Build GRCh38
3516104 c.490G>A p.E164K Chr.17: 34577541-34577541 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148761253 Chr.17: 34577492-34577492 on Build GRCh38
rs887230 Chr.17: 34577567-34577567 on Build GRCh38
rs909662404 Chr.17: 34577664-34577664 on Build GRCh38
rs768336017 Chr.17: 34577610-34577610 on Build GRCh38
rs896943806 Chr.17: 34577822-34577822 on Build GRCh38
rs866394628 Chr.17: 34577717-34577717 on Build GRCh38
rs753869335 Chr.17: 34577543-34577543 on Build GRCh38
rs539191323 Chr.17: 34577603-34577603 on Build GRCh38
rs767985461 Chr.17: 34577504-34577504 on Build GRCh38
rs768895164 Chr.17: 34577558-34577558 on Build GRCh38
rs942344336 Chr.17: 34577685-34577685 on Build GRCh38
rs919425959 Chr.17: 34577576-34577576 on Build GRCh38
rs141588018 Chr.17: 34577591-34577591 on Build GRCh38
rs761105442 Chr.17: 34577661-34577661 on Build GRCh38
rs747527996 Chr.17: 34577507-34577508 on Build GRCh38
rs769069787 Chr.17: 34577783-34577783 on Build GRCh38
rs949038556 Chr.17: 34577758-34577760 on Build GRCh38
rs375653137 Chr.17: 34577670-34577670 on Build GRCh38
rs748773930 Chr.17: 34577609-34577609 on Build GRCh38
rs768226384 Chr.17: 34577478-34577478 on Build GRCh38
rs1000020337 Chr.17: 34577875-34577875 on Build GRCh38
rs769864942 Chr.17: 34577580-34577580 on Build GRCh38
rs183747472 Chr.17: 34577437-34577437 on Build GRCh38
rs781454619 Chr.17: 34577560-34577560 on Build GRCh38
rs549690989 Chr.17: 34577555-34577555 on Build GRCh38
rs745854872 Chr.17: 34577577-34577577 on Build GRCh38
rs879833758 Chr.17: 34577694-34577694 on Build GRCh38
rs773919687 Chr.17: 34577618-34577618 on Build GRCh38
rs755186406 Chr.17: 34577547-34577547 on Build GRCh38
rs192717592 Chr.17: 34577739-34577739 on Build GRCh38
rs780393150 Chr.17: 34577569-34577569 on Build GRCh38
rs774731796 Chr.17: 34577490-34577490 on Build GRCh38
rs772484256 Chr.17: 34577642-34577642 on Build GRCh38
rs185396582 Chr.17: 34577772-34577772 on Build GRCh38
rs773581453 Chr.17: 34577659-34577659 on Build GRCh38
rs534886793 Chr.17: 34577695-34577695 on Build GRCh38
rs903022622 Chr.17: 34577844-34577844 on Build GRCh38
rs370711928 Chr.17: 34577523-34577523 on Build GRCh38
rs550234036 Chr.17: 34577457-34577457 on Build GRCh38
rs781267063 Chr.17: 34577575-34577575 on Build GRCh38
rs930835922 Chr.17: 34577586-34577586 on Build GRCh38
rs187767784 Chr.17: 34577455-34577455 on Build GRCh38
rs978985441 Chr.17: 34577573-34577573 on Build GRCh38
rs751347186 Chr.17: 34577558-34577558 on Build GRCh38
rs1024954430 Chr.17: 34577522-34577522 on Build GRCh38
rs747277838 Chr.17: 34577606-34577606 on Build GRCh38
rs72817878 Chr.17: 34577465-34577465 on Build GRCh38
rs1020088810 Chr.17: 34577417-34577417 on Build GRCh38
rs937502815 Chr.17: 34577643-34577643 on Build GRCh38
rs377460489 Chr.17: 34577669-34577669 on Build GRCh38
rs750996161 Chr.17: 34577511-34577511 on Build GRCh38
rs766390051 Chr.17: 34577539-34577539 on Build GRCh38
rs887231 Chr.17: 34577461-34577461 on Build GRCh38
rs983862596 Chr.17: 34577588-34577588 on Build GRCh38
rs1039391769 Chr.17: 34577754-34577754 on Build GRCh38
rs1034891398 Chr.17: 34577714-34577714 on Build GRCh38
rs994310666 Chr.17: 34577826-34577826 on Build GRCh38
rs548506577 Chr.17: 34577468-34577468 on Build GRCh38
rs551981753 Chr.17: 34577827-34577827 on Build GRCh38
rs888293946 Chr.17: 34577877-34577877 on Build GRCh38
rs78865828 Chr.17: 34577699-34577699 on Build GRCh38
rs776981597 Chr.17: 34577786-34577786 on Build GRCh38

View Full Product Details