Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 34939162-34939667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT6B
Entrez Gene ID
10693
Gene Name
chaperonin containing TCP1 subunit 6B
Gene Aliases
CCT-zeta-2, CCTZ-2, Cctz2, TCP-1-zeta-2, TSA303
UniGene
Hs.73072
Gene Chromosome Location
Chr.17: 34927859-34981221 on Build GRCh38
Transcripts
NM_001193529.2 NM_001193530.1 NM_006584.3 XM_011524203.2 XM_017024023.1 XM_017024024.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
436371 c.1028G>A p.C343Y Chr.17: 34939654-34939654 on Build GRCh38
7127182 c.1090G>A p.E364K Chr.17: 34939306-34939306 on Build GRCh38
460369 c.1186C>T p.R396C Chr.17: 34939210-34939210 on Build GRCh38
3180979 c.1199delA p.N400fs*30 Chr.17: 34939197-34939197 on Build GRCh38
3180980 c.1187G>A p.R396H Chr.17: 34939209-34939209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540346323 Chr.17: 34939389-34939389 on Build GRCh38
rs773246465 Chr.17: 34939591-34939591 on Build GRCh38
rs1006628521 Chr.17: 34939277-34939277 on Build GRCh38
rs1024721604 Chr.17: 34939429-34939429 on Build GRCh38
rs760199064 Chr.17: 34939301-34939301 on Build GRCh38
rs770948415 Chr.17: 34939376-34939376 on Build GRCh38
rs772000270 Chr.17: 34939589-34939589 on Build GRCh38
rs748652578 Chr.17: 34939217-34939217 on Build GRCh38
rs200442034 Chr.17: 34939604-34939604 on Build GRCh38
rs761838921 Chr.17: 34939260-34939260 on Build GRCh38
rs376987977 Chr.17: 34939251-34939251 on Build GRCh38
rs189867157 Chr.17: 34939638-34939638 on Build GRCh38
rs760170731 Chr.17: 34939204-34939204 on Build GRCh38
rs376734116 Chr.17: 34939584-34939584 on Build GRCh38
rs560057504 Chr.17: 34939407-34939407 on Build GRCh38
rs755333614 Chr.17: 34939210-34939210 on Build GRCh38
rs760714484 Chr.17: 34939602-34939602 on Build GRCh38
rs779328984 Chr.17: 34939341-34939341 on Build GRCh38
rs777459383 Chr.17: 34939271-34939277 on Build GRCh38
rs17628745 Chr.17: 34939523-34939523 on Build GRCh38
rs752349208 Chr.17: 34939643-34939643 on Build GRCh38
rs151240877 Chr.17: 34939437-34939437 on Build GRCh38
rs753057753 Chr.17: 34939343-34939343 on Build GRCh38
rs111862987 Chr.17: 34939553-34939553 on Build GRCh38
rs868601988 Chr.17: 34939225-34939225 on Build GRCh38
rs375397470 Chr.17: 34939209-34939209 on Build GRCh38
rs558768678 Chr.17: 34939600-34939600 on Build GRCh38
rs536320955 Chr.17: 34939561-34939561 on Build GRCh38
rs937456072 Chr.17: 34939453-34939453 on Build GRCh38
rs771001368 Chr.17: 34939227-34939227 on Build GRCh38
rs550681681 Chr.17: 34939533-34939533 on Build GRCh38
rs1013377783 Chr.17: 34939427-34939429 on Build GRCh38
rs770708698 Chr.17: 34939353-34939354 on Build GRCh38
rs1020624501 Chr.17: 34939370-34939370 on Build GRCh38
rs373049805 Chr.17: 34939366-34939366 on Build GRCh38
rs761155021 Chr.17: 34939599-34939599 on Build GRCh38
rs778184505 Chr.17: 34939352-34939352 on Build GRCh38
rs184425043 Chr.17: 34939577-34939577 on Build GRCh38
rs139262312 Chr.17: 34939514-34939514 on Build GRCh38
rs896814993 Chr.17: 34939463-34939463 on Build GRCh38
rs202179345 Chr.17: 34939325-34939325 on Build GRCh38
rs12452225 Chr.17: 34939412-34939412 on Build GRCh38
rs750401688 Chr.17: 34939299-34939299 on Build GRCh38
rs925769794 Chr.17: 34939418-34939418 on Build GRCh38
rs764735174 Chr.17: 34939639-34939639 on Build GRCh38
rs748762061 Chr.17: 34939274-34939275 on Build GRCh38
rs772629699 Chr.17: 34939218-34939218 on Build GRCh38
rs1017984369 Chr.17: 34939285-34939285 on Build GRCh38
rs769473469 Chr.17: 34939243-34939244 on Build GRCh38
rs574427480 Chr.17: 34939319-34939319 on Build GRCh38
rs754622527 Chr.17: 34939336-34939336 on Build GRCh38
rs774054618 Chr.17: 34939253-34939253 on Build GRCh38
rs776871308 Chr.17: 34939232-34939232 on Build GRCh38
rs181826563 Chr.17: 34939569-34939569 on Build GRCh38
rs759698643 Chr.17: 34939240-34939240 on Build GRCh38
rs946029433 Chr.17: 34939301-34939303 on Build GRCh38
rs758773232 Chr.17: 34939347-34939347 on Build GRCh38
rs913257420 Chr.17: 34939292-34939292 on Build GRCh38
rs904746016 Chr.17: 34939340-34939340 on Build GRCh38
rs746072264 Chr.17: 34939573-34939573 on Build GRCh38
rs570217709 Chr.17: 34939229-34939229 on Build GRCh38
rs767623550 Chr.17: 34939289-34939289 on Build GRCh38
rs779798298 Chr.17: 34939198-34939198 on Build GRCh38
rs774139572 Chr.17: 34939575-34939575 on Build GRCh38
rs763580867 Chr.17: 34939222-34939222 on Build GRCh38
rs753351932 Chr.17: 34939627-34939627 on Build GRCh38
rs779171045 Chr.17: 34939213-34939213 on Build GRCh38
rs878889206 Chr.17: 34939339-34939339 on Build GRCh38
rs758194713 Chr.17: 34939526-34939526 on Build GRCh38

View Full Product Details