Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 37375958-37376468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACACA
Entrez Gene ID
31
Gene Name
acetyl-CoA carboxylase alpha
Gene Aliases
ACAC, ACACAD, ACC, ACC1, ACCA
UniGene
Hs.160556
Gene Chromosome Location
Chr.17: 37084992-37406822 on Build GRCh38
Transcripts
NM_198834.2 NM_198839.2 XM_006721853.1 XM_011524701.1 XM_017024553.1 XM_017024554.1 NM_001321399.1 NM_173625.4 XM_011524647.2 XM_011524648.2 XM_011524649.2

Gene Symbol
C17orf78
Entrez Gene ID
284099
Gene Name
chromosome 17 open reading frame 78
Gene Aliases
-
UniGene
Hs.439154
Gene Chromosome Location
Chr.17: 37375710-37392708 on Build GRCh38
Transcripts
NM_198834.2 NM_198839.2 XM_006721853.1 XM_011524701.1 XM_017024553.1 XM_017024554.1 NM_001321399.1 NM_173625.4 XM_011524647.2 XM_011524648.2 XM_011524649.2

SNP Detail

SNP ID Chromosome Location
rs765214527 Chr.17: 37376133-37376133 on Build GRCh38
rs116383361 Chr.17: 37376247-37376247 on Build GRCh38
rs937182110 Chr.17: 37376210-37376210 on Build GRCh38
rs752504175 Chr.17: 37376049-37376049 on Build GRCh38
rs922121789 Chr.17: 37376380-37376380 on Build GRCh38
rs1007056791 Chr.17: 37376337-37376337 on Build GRCh38
rs563108684 Chr.17: 37376106-37376106 on Build GRCh38
rs756927780 Chr.17: 37376171-37376171 on Build GRCh38
rs750011794 Chr.17: 37376003-37376003 on Build GRCh38
rs576299856 Chr.17: 37376422-37376422 on Build GRCh38
rs769203964 Chr.17: 37376101-37376101 on Build GRCh38
rs368122704 Chr.17: 37376081-37376081 on Build GRCh38
rs1048636501 Chr.17: 37376322-37376322 on Build GRCh38
rs897516933 Chr.17: 37376278-37376278 on Build GRCh38
rs942166115 Chr.17: 37376287-37376287 on Build GRCh38
rs879029218 Chr.17: 37376270-37376270 on Build GRCh38
rs780467886 Chr.17: 37376085-37376085 on Build GRCh38
rs556431292 Chr.17: 37376325-37376325 on Build GRCh38
rs751174928 Chr.17: 37376168-37376168 on Build GRCh38
rs527925148 Chr.17: 37376222-37376222 on Build GRCh38
rs368430689 Chr.17: 37376186-37376186 on Build GRCh38
rs545733175 Chr.17: 37376289-37376289 on Build GRCh38
rs763721888 Chr.17: 37376054-37376054 on Build GRCh38
rs888732711 Chr.17: 37376334-37376334 on Build GRCh38
rs902549352 Chr.17: 37376039-37376039 on Build GRCh38
rs751781871 Chr.17: 37376068-37376068 on Build GRCh38
rs1018882633 Chr.17: 37376338-37376338 on Build GRCh38
rs780786842 Chr.17: 37376075-37376075 on Build GRCh38
rs1035955482 Chr.17: 37376251-37376251 on Build GRCh38
rs183433667 Chr.17: 37376022-37376022 on Build GRCh38
rs756059476 Chr.17: 37376353-37376353 on Build GRCh38
rs749669560 Chr.17: 37376086-37376086 on Build GRCh38
rs374670469 Chr.17: 37376078-37376078 on Build GRCh38
rs1051889145 Chr.17: 37376405-37376405 on Build GRCh38
rs370967445 Chr.17: 37376153-37376153 on Build GRCh38
rs902473354 Chr.17: 37376431-37376431 on Build GRCh38
rs756689419 Chr.17: 37376064-37376064 on Build GRCh38
rs775347770 Chr.17: 37376149-37376149 on Build GRCh38
rs541206625 Chr.17: 37375983-37375983 on Build GRCh38
rs748408733 Chr.17: 37376121-37376121 on Build GRCh38
rs751226127 Chr.17: 37376057-37376057 on Build GRCh38
rs893595079 Chr.17: 37376151-37376151 on Build GRCh38
rs974919709 Chr.17: 37376345-37376345 on Build GRCh38
rs1039496014 Chr.17: 37376010-37376010 on Build GRCh38
rs199550394 Chr.17: 37376123-37376123 on Build GRCh38
rs117782053 Chr.17: 37376130-37376130 on Build GRCh38
rs770317044 Chr.17: 37376211-37376211 on Build GRCh38
rs1051753128 Chr.17: 37376033-37376033 on Build GRCh38
rs543408541 Chr.17: 37376031-37376031 on Build GRCh38
rs965885599 Chr.17: 37376021-37376021 on Build GRCh38
rs758254610 Chr.17: 37376050-37376050 on Build GRCh38
rs913265179 Chr.17: 37376024-37376024 on Build GRCh38
rs930191650 Chr.17: 37376290-37376290 on Build GRCh38
rs375673502 Chr.17: 37376164-37376164 on Build GRCh38
rs951090784 Chr.17: 37376246-37376246 on Build GRCh38
rs189618982 Chr.17: 37376011-37376011 on Build GRCh38
rs933383517 Chr.17: 37376401-37376401 on Build GRCh38
rs752564406 Chr.17: 37376190-37376190 on Build GRCh38
rs773317302 Chr.17: 37376126-37376126 on Build GRCh38

View Full Product Details