Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 37388817-37389352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACACA
Entrez Gene ID
31
Gene Name
acetyl-CoA carboxylase alpha
Gene Aliases
ACAC, ACACAD, ACC, ACC1, ACCA
UniGene
Hs.160556
Gene Chromosome Location
Chr.17: 37084992-37406822 on Build GRCh38
Transcripts
NM_198834.2 NM_198839.2 XM_006721853.1 XM_011524701.1 XM_017024553.1 XM_017024554.1 NM_001321399.1 NM_173625.4 XM_011524647.2 XM_011524648.2 XM_011524649.2

Gene Symbol
C17orf78
Entrez Gene ID
284099
Gene Name
chromosome 17 open reading frame 78
Gene Aliases
-
UniGene
Hs.439154
Gene Chromosome Location
Chr.17: 37375710-37392708 on Build GRCh38
Transcripts
NM_198834.2 NM_198839.2 XM_006721853.1 XM_011524701.1 XM_017024553.1 XM_017024554.1 NM_001321399.1 NM_173625.4 XM_011524647.2 XM_011524648.2 XM_011524649.2

SNP Detail

SNP ID Chromosome Location
rs192034327 Chr.17: 37388931-37388931 on Build GRCh38
rs142422952 Chr.17: 37389155-37389155 on Build GRCh38
rs1039620060 Chr.17: 37389023-37389023 on Build GRCh38
rs202068969 Chr.17: 37389316-37389316 on Build GRCh38
rs757857956 Chr.17: 37389219-37389219 on Build GRCh38
rs868186486 Chr.17: 37388861-37388861 on Build GRCh38
rs533343910 Chr.17: 37388962-37388962 on Build GRCh38
rs1013032592 Chr.17: 37389249-37389249 on Build GRCh38
rs918634615 Chr.17: 37388885-37388885 on Build GRCh38
rs749454954 Chr.17: 37389071-37389071 on Build GRCh38
rs901118087 Chr.17: 37389025-37389025 on Build GRCh38
rs764807537 Chr.17: 37388844-37388844 on Build GRCh38
rs777298912 Chr.17: 37389225-37389225 on Build GRCh38
rs749540176 Chr.17: 37389278-37389278 on Build GRCh38
rs373752146 Chr.17: 37388942-37388942 on Build GRCh38
rs1011278850 Chr.17: 37389292-37389292 on Build GRCh38
rs751798481 Chr.17: 37389302-37389302 on Build GRCh38
rs757761449 Chr.17: 37389238-37389238 on Build GRCh38
rs1052393684 Chr.17: 37389156-37389156 on Build GRCh38
rs752361855 Chr.17: 37389204-37389204 on Build GRCh38
rs940944240 Chr.17: 37388923-37388923 on Build GRCh38
rs775796113 Chr.17: 37389272-37389272 on Build GRCh38
rs951552971 Chr.17: 37389320-37389320 on Build GRCh38
rs372283806 Chr.17: 37389230-37389230 on Build GRCh38
rs970971760 Chr.17: 37389269-37389269 on Build GRCh38
rs930063563 Chr.17: 37388904-37388904 on Build GRCh38
rs528547782 Chr.17: 37389016-37389018 on Build GRCh38
rs776441260 Chr.17: 37389323-37389323 on Build GRCh38
rs761844357 Chr.17: 37389312-37389312 on Build GRCh38
rs200034389 Chr.17: 37389261-37389261 on Build GRCh38
rs773101578 Chr.17: 37389039-37389039 on Build GRCh38
rs774538336 Chr.17: 37389310-37389310 on Build GRCh38
rs908091838 Chr.17: 37388868-37388868 on Build GRCh38
rs781607152 Chr.17: 37389239-37389239 on Build GRCh38
rs982284448 Chr.17: 37388867-37388867 on Build GRCh38
rs1033200805 Chr.17: 37389223-37389223 on Build GRCh38
rs746238628 Chr.17: 37389252-37389252 on Build GRCh38
rs770354299 Chr.17: 37388980-37388980 on Build GRCh38
rs984202581 Chr.17: 37389321-37389321 on Build GRCh38
rs368731173 Chr.17: 37389279-37389279 on Build GRCh38
rs865991561 Chr.17: 37389299-37389299 on Build GRCh38
rs999890812 Chr.17: 37389177-37389177 on Build GRCh38
rs1048844557 Chr.17: 37388937-37388937 on Build GRCh38
rs997939396 Chr.17: 37389031-37389031 on Build GRCh38
rs1041552906 Chr.17: 37389109-37389109 on Build GRCh38

View Full Product Details