Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38674381-38674915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf96
Entrez Gene ID
100170841
Gene Name
chromosome 17 open reading frame 96
Gene Aliases
PRR28
UniGene
Hs.445574
Gene Chromosome Location
Chr.17: 38671703-38674934 on Build GRCh38
Transcripts
NM_001130677.1

SNP Detail

SNP ID Chromosome Location
rs1007163383 Chr.17: 38674556-38674556 on Build GRCh38
rs918679088 Chr.17: 38674470-38674470 on Build GRCh38
rs548343851 Chr.17: 38674429-38674429 on Build GRCh38
rs935636952 Chr.17: 38674449-38674449 on Build GRCh38
rs76756126 Chr.17: 38674474-38674474 on Build GRCh38
rs749244201 Chr.17: 38674438-38674438 on Build GRCh38
rs774071518 Chr.17: 38674471-38674471 on Build GRCh38
rs960516052 Chr.17: 38674767-38674767 on Build GRCh38
rs1016999883 Chr.17: 38674566-38674566 on Build GRCh38
rs867700438 Chr.17: 38674412-38674412 on Build GRCh38
rs1051006129 Chr.17: 38674835-38674835 on Build GRCh38
rs960905763 Chr.17: 38674633-38674633 on Build GRCh38
rs867319646 Chr.17: 38674408-38674408 on Build GRCh38
rs985421819 Chr.17: 38674425-38674425 on Build GRCh38
rs570756341 Chr.17: 38674595-38674595 on Build GRCh38
rs891567590 Chr.17: 38674617-38674617 on Build GRCh38
rs889617165 Chr.17: 38674521-38674521 on Build GRCh38
rs948042536 Chr.17: 38674484-38674484 on Build GRCh38
rs889493156 Chr.17: 38674644-38674644 on Build GRCh38
rs533991793 Chr.17: 38674477-38674477 on Build GRCh38
rs113327406 Chr.17: 38674851-38674851 on Build GRCh38
rs555554976 Chr.17: 38674815-38674815 on Build GRCh38
rs992421007 Chr.17: 38674634-38674634 on Build GRCh38
rs935751369 Chr.17: 38674695-38674695 on Build GRCh38
rs935496376 Chr.17: 38674580-38674580 on Build GRCh38
rs113176541 Chr.17: 38674850-38674850 on Build GRCh38
rs111725439 Chr.17: 38674427-38674427 on Build GRCh38
rs925703723 Chr.17: 38674679-38674679 on Build GRCh38
rs535669784 Chr.17: 38674777-38674777 on Build GRCh38
rs933475476 Chr.17: 38674867-38674867 on Build GRCh38
rs1017255175 Chr.17: 38674440-38674440 on Build GRCh38
rs1052789950 Chr.17: 38674462-38674462 on Build GRCh38
rs925541554 Chr.17: 38674838-38674838 on Build GRCh38
rs933675989 Chr.17: 38674802-38674802 on Build GRCh38
rs772891191 Chr.17: 38674481-38674481 on Build GRCh38
rs948084421 Chr.17: 38674799-38674799 on Build GRCh38
rs979809793 Chr.17: 38674821-38674821 on Build GRCh38
rs1006963911 Chr.17: 38674658-38674658 on Build GRCh38
rs953824250 Chr.17: 38674413-38674413 on Build GRCh38
rs113134517 Chr.17: 38674727-38674727 on Build GRCh38
rs572210701 Chr.17: 38674823-38674823 on Build GRCh38
rs980033188 Chr.17: 38674674-38674674 on Build GRCh38
rs539751313 Chr.17: 38674619-38674619 on Build GRCh38
rs898550218 Chr.17: 38674875-38674875 on Build GRCh38
rs1044023313 Chr.17: 38674552-38674552 on Build GRCh38
rs547813524 Chr.17: 38674589-38674589 on Build GRCh38
rs1052668156 Chr.17: 38674846-38674846 on Build GRCh38
rs1038623324 Chr.17: 38674853-38674853 on Build GRCh38
rs576371390 Chr.17: 38674751-38674751 on Build GRCh38
rs1052579863 Chr.17: 38674593-38674593 on Build GRCh38
rs768640657 Chr.17: 38674455-38674455 on Build GRCh38
rs962568359 Chr.17: 38674701-38674701 on Build GRCh38
rs962786359 Chr.17: 38674603-38674603 on Build GRCh38
rs912906907 Chr.17: 38674785-38674785 on Build GRCh38
rs1025456207 Chr.17: 38674755-38674755 on Build GRCh38
rs556170223 Chr.17: 38674725-38674725 on Build GRCh38
rs891691470 Chr.17: 38674464-38674464 on Build GRCh38
rs1006699823 Chr.17: 38674852-38674852 on Build GRCh38
rs889724142 Chr.17: 38674841-38674841 on Build GRCh38
rs1025608350 Chr.17: 38674622-38674622 on Build GRCh38
rs916651140 Chr.17: 38674789-38674789 on Build GRCh38
rs969571053 Chr.17: 38674660-38674660 on Build GRCh38
rs912961551 Chr.17: 38674730-38674730 on Build GRCh38
rs916852372 Chr.17: 38674641-38674641 on Build GRCh38

View Full Product Details