Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10551955-10552447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG4D
Entrez Gene ID
84971
Gene Name
autophagy related 4D cysteine peptidase
Gene Aliases
APG4-D, APG4D, AUTL4
UniGene
Hs.512799
Gene Chromosome Location
Chr.19: 10543894-10553423 on Build GRCh38
Transcripts
NM_001281504.1 NM_032885.5 NR_104024.1 NR_104025.1 XM_006722927.1 NR_031603.1

Gene Symbol
MIR1238
Entrez Gene ID
100302226
Gene Name
microRNA 1238
Gene Aliases
MIRN1238, hsa-mir-1238
UniGene
Gene Chromosome Location
Chr.19: 10552122-10552204 on Build GRCh38
Transcripts
NM_001281504.1 NM_032885.5 NR_104024.1 NR_104025.1 XM_006722927.1 NR_031603.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6606723 c.1122G>A p.E374E Chr.19: 10552121-10552121 on Build GRCh38
990619 c.1046-2A>G p.? Chr.19: 10552043-10552043 on Build GRCh38
5679549 c.1220C>T p.T407I Chr.19: 10552302-10552302 on Build GRCh38
4073883 c.1057C>T p.L353L Chr.19: 10552056-10552056 on Build GRCh38
7001598 c.1109_1110ins11 p.D370fs*47 Chr.19: 10552108-10552109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201291151 Chr.19: 10552265-10552265 on Build GRCh38
rs758790754 Chr.19: 10552218-10552218 on Build GRCh38
rs368969533 Chr.19: 10552146-10552146 on Build GRCh38
rs561166520 Chr.19: 10552362-10552362 on Build GRCh38
rs765558252 Chr.19: 10552160-10552160 on Build GRCh38
rs746490907 Chr.19: 10552154-10552154 on Build GRCh38
rs532075878 Chr.19: 10552031-10552031 on Build GRCh38
rs746282597 Chr.19: 10551993-10551997 on Build GRCh38
rs775774575 Chr.19: 10551993-10551993 on Build GRCh38
rs760450241 Chr.19: 10552192-10552192 on Build GRCh38
rs781550454 Chr.19: 10551990-10551991 on Build GRCh38
rs769534266 Chr.19: 10552106-10552106 on Build GRCh38
rs777803970 Chr.19: 10552344-10552344 on Build GRCh38
rs756224309 Chr.19: 10552318-10552318 on Build GRCh38
rs776358793 Chr.19: 10552290-10552290 on Build GRCh38
rs776239203 Chr.19: 10552354-10552354 on Build GRCh38
rs766736761 Chr.19: 10552221-10552221 on Build GRCh38
rs933816574 Chr.19: 10552392-10552392 on Build GRCh38
rs201531397 Chr.19: 10552168-10552168 on Build GRCh38
rs765893329 Chr.19: 10552132-10552132 on Build GRCh38
rs142513981 Chr.19: 10552264-10552264 on Build GRCh38
rs537782433 Chr.19: 10552183-10552183 on Build GRCh38
rs867610666 Chr.19: 10551981-10551981 on Build GRCh38
rs373647217 Chr.19: 10552351-10552351 on Build GRCh38
rs767215101 Chr.19: 10552154-10552154 on Build GRCh38
rs141346756 Chr.19: 10552121-10552121 on Build GRCh38
rs754151658 Chr.19: 10552348-10552348 on Build GRCh38
rs369078960 Chr.19: 10552303-10552303 on Build GRCh38
rs148200963 Chr.19: 10552103-10552103 on Build GRCh38
rs781014878 Chr.19: 10552023-10552023 on Build GRCh38
rs750709640 Chr.19: 10552211-10552211 on Build GRCh38
rs770777634 Chr.19: 10552248-10552248 on Build GRCh38
rs748970122 Chr.19: 10552247-10552247 on Build GRCh38
rs371715950 Chr.19: 10552158-10552158 on Build GRCh38
rs758778787 Chr.19: 10552162-10552162 on Build GRCh38
rs757567365 Chr.19: 10551990-10551990 on Build GRCh38
rs760259667 Chr.19: 10552286-10552286 on Build GRCh38
rs200136327 Chr.19: 10552190-10552190 on Build GRCh38
rs138092907 Chr.19: 10552242-10552242 on Build GRCh38
rs750985644 Chr.19: 10552137-10552137 on Build GRCh38
rs775280853 Chr.19: 10552191-10552191 on Build GRCh38
rs146007239 Chr.19: 10552289-10552289 on Build GRCh38
rs368082674 Chr.19: 10552018-10552018 on Build GRCh38
rs11882419 Chr.19: 10552030-10552030 on Build GRCh38
rs762841570 Chr.19: 10552125-10552125 on Build GRCh38
rs371466606 Chr.19: 10552227-10552227 on Build GRCh38
rs752118366 Chr.19: 10552222-10552222 on Build GRCh38
rs779322307 Chr.19: 10551984-10551984 on Build GRCh38
rs746423145 Chr.19: 10552058-10552058 on Build GRCh38
rs765873601 Chr.19: 10552016-10552016 on Build GRCh38
rs780044450 Chr.19: 10552067-10552067 on Build GRCh38
rs376594506 Chr.19: 10552014-10552014 on Build GRCh38
rs372408468 Chr.19: 10552364-10552364 on Build GRCh38
rs768822162 Chr.19: 10552094-10552094 on Build GRCh38
rs755043990 Chr.19: 10552224-10552224 on Build GRCh38
rs547538797 Chr.19: 10552033-10552033 on Build GRCh38
rs374913540 Chr.19: 10552188-10552188 on Build GRCh38
rs369002764 Chr.19: 10552219-10552219 on Build GRCh38
rs769601412 Chr.19: 10552174-10552174 on Build GRCh38
rs764246756 Chr.19: 10551990-10552004 on Build GRCh38
rs770621403 Chr.19: 10552331-10552333 on Build GRCh38
rs775088570 Chr.19: 10552285-10552285 on Build GRCh38
rs199912582 Chr.19: 10552159-10552159 on Build GRCh38
rs780524649 Chr.19: 10552001-10552007 on Build GRCh38
rs376707293 Chr.19: 10552189-10552189 on Build GRCh38
rs776143172 Chr.19: 10552375-10552375 on Build GRCh38
rs779420420 Chr.19: 10552037-10552037 on Build GRCh38
rs910022809 Chr.19: 10552385-10552385 on Build GRCh38
rs756516101 Chr.19: 10551998-10551999 on Build GRCh38
rs751529633 Chr.19: 10551990-10552000 on Build GRCh38
rs150808873 Chr.19: 10552226-10552226 on Build GRCh38
rs1055905727 Chr.19: 10552413-10552413 on Build GRCh38
rs768036629 Chr.19: 10552307-10552307 on Build GRCh38
rs923845236 Chr.19: 10552379-10552379 on Build GRCh38
rs755064220 Chr.19: 10552167-10552167 on Build GRCh38
rs745897034 Chr.19: 10551985-10551985 on Build GRCh38
rs772780311 Chr.19: 10552118-10552118 on Build GRCh38
rs761538562 Chr.19: 10552104-10552104 on Build GRCh38
rs964300302 Chr.19: 10552078-10552078 on Build GRCh38
rs763826655 Chr.19: 10552196-10552196 on Build GRCh38
rs766795163 Chr.19: 10552296-10552296 on Build GRCh38
rs746443658 Chr.19: 10552363-10552363 on Build GRCh38
rs752733894 Chr.19: 10552316-10552316 on Build GRCh38
rs772060737 Chr.19: 10551990-10551990 on Build GRCh38
rs576244185 Chr.19: 10552393-10552393 on Build GRCh38
rs780638134 Chr.19: 10552163-10552163 on Build GRCh38
rs760844215 Chr.19: 10551995-10551995 on Build GRCh38
rs765090878 Chr.19: 10552011-10552011 on Build GRCh38
rs139819593 Chr.19: 10552385-10552386 on Build GRCh38
rs371619415 Chr.19: 10552361-10552361 on Build GRCh38
rs759153195 Chr.19: 10552355-10552355 on Build GRCh38
rs768984488 Chr.19: 10552025-10552026 on Build GRCh38
rs756243375 Chr.19: 10552233-10552233 on Build GRCh38
rs1007279309 Chr.19: 10552329-10552329 on Build GRCh38
rs749820122 Chr.19: 10552022-10552023 on Build GRCh38
rs748434288 Chr.19: 10552171-10552171 on Build GRCh38
rs376871668 Chr.19: 10552270-10552270 on Build GRCh38
rs368055589 Chr.19: 10552356-10552356 on Build GRCh38
rs750917610 Chr.19: 10552161-10552161 on Build GRCh38
rs749131513 Chr.19: 10552184-10552184 on Build GRCh38
rs747111031 Chr.19: 10552092-10552092 on Build GRCh38
rs763214319 Chr.19: 10552294-10552294 on Build GRCh38
rs866498999 Chr.19: 10552000-10552000 on Build GRCh38
rs747023526 Chr.19: 10552166-10552166 on Build GRCh38
rs201971548 Chr.19: 10552155-10552155 on Build GRCh38
rs923787724 Chr.19: 10552142-10552142 on Build GRCh38
rs1032804091 Chr.19: 10552237-10552238 on Build GRCh38
rs34986913 Chr.19: 10552111-10552112 on Build GRCh38
rs201327730 Chr.19: 10552013-10552013 on Build GRCh38
rs759152766 Chr.19: 10552138-10552138 on Build GRCh38
rs201728867 Chr.19: 10552237-10552237 on Build GRCh38
rs761626734 Chr.19: 10552005-10552005 on Build GRCh38
rs776356423 Chr.19: 10552004-10552004 on Build GRCh38
rs778895215 Chr.19: 10552355-10552355 on Build GRCh38
rs768414611 Chr.19: 10551997-10551997 on Build GRCh38
rs772157379 Chr.19: 10552061-10552061 on Build GRCh38
rs760087282 Chr.19: 10552306-10552306 on Build GRCh38

View Full Product Details