Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35142366-35142861 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXYD1
Entrez Gene ID
5348
Gene Name
FXYD domain containing ion transport regulator 1
Gene Aliases
PLM
UniGene
Hs.442498
Gene Chromosome Location
Chr.19: 35137207-35143055 on Build GRCh38
Transcripts
NM_001278717.1 NM_001278718.1 NM_005031.4 NM_021902.3 XM_017026874.1 XM_017026875.1 XM_017026876.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6667976 c.259C>G p.L87V Chr.19: 35142722-35142722 on Build GRCh38
3532445 c.231G>A p.E77E Chr.19: 35142496-35142496 on Build GRCh38
6667975 c.254G>A p.R85H Chr.19: 35142519-35142519 on Build GRCh38
4649611 c.219C>T p.P73P Chr.19: 35142484-35142484 on Build GRCh38
6667974 c.257-8delC p.? Chr.19: 35142712-35142712 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs368962711 Chr.19: 35142678-35142678 on Build GRCh38
rs374848502 Chr.19: 35142545-35142545 on Build GRCh38
rs569083817 Chr.19: 35142442-35142442 on Build GRCh38
rs112541004 Chr.19: 35142719-35142719 on Build GRCh38
rs974371195 Chr.19: 35142839-35142839 on Build GRCh38
rs879208816 Chr.19: 35142654-35142654 on Build GRCh38
rs760787707 Chr.19: 35142692-35142692 on Build GRCh38
rs761938361 Chr.19: 35142675-35142675 on Build GRCh38
rs1022301037 Chr.19: 35142639-35142639 on Build GRCh38
rs377667040 Chr.19: 35142749-35142749 on Build GRCh38
rs199635474 Chr.19: 35142427-35142427 on Build GRCh38
rs572001504 Chr.19: 35142802-35142802 on Build GRCh38
rs1053332864 Chr.19: 35142636-35142636 on Build GRCh38
rs543842833 Chr.19: 35142628-35142643 on Build GRCh38
rs776905341 Chr.19: 35142789-35142789 on Build GRCh38
rs767711892 Chr.19: 35142677-35142677 on Build GRCh38
rs893373962 Chr.19: 35142646-35142646 on Build GRCh38
rs765503312 Chr.19: 35142701-35142701 on Build GRCh38
rs540738270 Chr.19: 35142806-35142806 on Build GRCh38
rs200871076 Chr.19: 35142712-35142712 on Build GRCh38
rs775415071 Chr.19: 35142569-35142569 on Build GRCh38
rs962566365 Chr.19: 35142458-35142458 on Build GRCh38
rs769554986 Chr.19: 35142556-35142556 on Build GRCh38
rs1035132533 Chr.19: 35142586-35142586 on Build GRCh38
rs934324366 Chr.19: 35142634-35142634 on Build GRCh38
rs758125236 Chr.19: 35142529-35142529 on Build GRCh38
rs779456486 Chr.19: 35142521-35142521 on Build GRCh38
rs115949809 Chr.19: 35142415-35142415 on Build GRCh38
rs570867145 Chr.19: 35142695-35142695 on Build GRCh38
rs751206657 Chr.19: 35142717-35142717 on Build GRCh38
rs568451022 Chr.19: 35142594-35142594 on Build GRCh38
rs988173148 Chr.19: 35142626-35142626 on Build GRCh38
rs745846966 Chr.19: 35142551-35142551 on Build GRCh38
rs61753924 Chr.19: 35142731-35142731 on Build GRCh38
rs113276819 Chr.19: 35142450-35142450 on Build GRCh38
rs765051658 Chr.19: 35142474-35142474 on Build GRCh38
rs962637716 Chr.19: 35142830-35142830 on Build GRCh38
rs770791151 Chr.19: 35142449-35142449 on Build GRCh38
rs767481794 Chr.19: 35142487-35142487 on Build GRCh38
rs759280268 Chr.19: 35142470-35142470 on Build GRCh38
rs745791850 Chr.19: 35142734-35142734 on Build GRCh38
rs762145950 Chr.19: 35142484-35142484 on Build GRCh38
rs777140104 Chr.19: 35142538-35142538 on Build GRCh38
rs867846506 Chr.19: 35142570-35142570 on Build GRCh38
rs1023225618 Chr.19: 35142571-35142571 on Build GRCh38
rs907004298 Chr.19: 35142578-35142578 on Build GRCh38
rs536603524 Chr.19: 35142785-35142785 on Build GRCh38
rs1012835828 Chr.19: 35142566-35142566 on Build GRCh38
rs756731949 Chr.19: 35142721-35142721 on Build GRCh38
rs755353547 Chr.19: 35142518-35142518 on Build GRCh38
rs770455512 Chr.19: 35142546-35142546 on Build GRCh38
rs772823247 Chr.19: 35142440-35142440 on Build GRCh38
rs912234334 Chr.19: 35142627-35142627 on Build GRCh38
rs1135396 Chr.19: 35142746-35142746 on Build GRCh38
rs753170408 Chr.19: 35142707-35142707 on Build GRCh38
rs756266698 Chr.19: 35142499-35142499 on Build GRCh38
rs1004874934 Chr.19: 35142753-35142753 on Build GRCh38
rs149407483 Chr.19: 35142697-35142697 on Build GRCh38
rs563744235 Chr.19: 35142421-35142421 on Build GRCh38
rs371469517 Chr.19: 35142575-35142575 on Build GRCh38
rs776566973 Chr.19: 35142460-35142460 on Build GRCh38
rs548795635 Chr.19: 35142584-35142584 on Build GRCh38
rs754736922 Chr.19: 35142767-35142767 on Build GRCh38
rs1673004 Chr.19: 35142599-35142599 on Build GRCh38
rs777617045 Chr.19: 35142712-35142712 on Build GRCh38
rs776514193 Chr.19: 35142549-35142549 on Build GRCh38
rs375111127 Chr.19: 35142431-35142431 on Build GRCh38
rs765705537 Chr.19: 35142791-35142791 on Build GRCh38
rs371429019 Chr.19: 35142433-35142433 on Build GRCh38
rs369430419 Chr.19: 35142750-35142750 on Build GRCh38
rs771818590 Chr.19: 35142674-35142674 on Build GRCh38
rs368655960 Chr.19: 35142601-35142601 on Build GRCh38
rs74693828 Chr.19: 35142434-35142434 on Build GRCh38
rs774406399 Chr.19: 35142478-35142478 on Build GRCh38
rs750584773 Chr.19: 35142488-35142488 on Build GRCh38
rs371865318 Chr.19: 35142652-35142652 on Build GRCh38
rs201764718 Chr.19: 35142558-35142558 on Build GRCh38
rs371689893 Chr.19: 35142445-35142445 on Build GRCh38
rs755587482 Chr.19: 35142699-35142699 on Build GRCh38
rs535283242 Chr.19: 35142743-35142743 on Build GRCh38
rs754418457 Chr.19: 35142508-35142508 on Build GRCh38
rs766934917 Chr.19: 35142507-35142507 on Build GRCh38
rs747729066 Chr.19: 35142524-35142524 on Build GRCh38
rs976638962 Chr.19: 35142715-35142715 on Build GRCh38

View Full Product Details