Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35142559-35143053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXYD1
Entrez Gene ID
5348
Gene Name
FXYD domain containing ion transport regulator 1
Gene Aliases
PLM
UniGene
Hs.442498
Gene Chromosome Location
Chr.19: 35137207-35143055 on Build GRCh38
Transcripts
NM_001278717.1 NM_001278718.1 NM_005031.4 NM_021902.3 XM_017026874.1 XM_017026875.1 XM_017026876.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6667976 c.259C>G p.L87V Chr.19: 35142722-35142722 on Build GRCh38
6667974 c.257-8delC p.? Chr.19: 35142712-35142712 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs368962711 Chr.19: 35142678-35142678 on Build GRCh38
rs1056528962 Chr.19: 35142962-35142962 on Build GRCh38
rs112541004 Chr.19: 35142719-35142719 on Build GRCh38
rs974371195 Chr.19: 35142839-35142839 on Build GRCh38
rs879208816 Chr.19: 35142654-35142654 on Build GRCh38
rs760787707 Chr.19: 35142692-35142692 on Build GRCh38
rs761938361 Chr.19: 35142675-35142675 on Build GRCh38
rs921117360 Chr.19: 35142852-35142852 on Build GRCh38
rs1022301037 Chr.19: 35142639-35142639 on Build GRCh38
rs377667040 Chr.19: 35142749-35142749 on Build GRCh38
rs984494727 Chr.19: 35143017-35143017 on Build GRCh38
rs572001504 Chr.19: 35142802-35142802 on Build GRCh38
rs1053332864 Chr.19: 35142636-35142636 on Build GRCh38
rs577243868 Chr.19: 35142871-35142871 on Build GRCh38
rs776905341 Chr.19: 35142789-35142789 on Build GRCh38
rs543842833 Chr.19: 35142628-35142643 on Build GRCh38
rs767711892 Chr.19: 35142677-35142677 on Build GRCh38
rs893373962 Chr.19: 35142646-35142646 on Build GRCh38
rs200871076 Chr.19: 35142712-35142712 on Build GRCh38
rs765503312 Chr.19: 35142701-35142701 on Build GRCh38
rs540738270 Chr.19: 35142806-35142806 on Build GRCh38
rs1052699628 Chr.19: 35142948-35142948 on Build GRCh38
rs1035132533 Chr.19: 35142586-35142586 on Build GRCh38
rs934324366 Chr.19: 35142634-35142634 on Build GRCh38
rs908908856 Chr.19: 35143020-35143020 on Build GRCh38
rs938499533 Chr.19: 35142870-35142870 on Build GRCh38
rs1045007892 Chr.19: 35142995-35142995 on Build GRCh38
rs1009653336 Chr.19: 35142903-35142903 on Build GRCh38
rs570867145 Chr.19: 35142695-35142695 on Build GRCh38
rs1022339971 Chr.19: 35142918-35142918 on Build GRCh38
rs751206657 Chr.19: 35142717-35142717 on Build GRCh38
rs568451022 Chr.19: 35142594-35142594 on Build GRCh38
rs878988440 Chr.19: 35142921-35142921 on Build GRCh38
rs988173148 Chr.19: 35142626-35142626 on Build GRCh38
rs61753924 Chr.19: 35142731-35142731 on Build GRCh38
rs953975450 Chr.19: 35142854-35142854 on Build GRCh38
rs914213135 Chr.19: 35142965-35142965 on Build GRCh38
rs962637716 Chr.19: 35142830-35142830 on Build GRCh38
rs1052930248 Chr.19: 35142869-35142869 on Build GRCh38
rs906034229 Chr.19: 35143007-35143007 on Build GRCh38
rs377132171 Chr.19: 35142866-35142866 on Build GRCh38
rs745791850 Chr.19: 35142734-35142734 on Build GRCh38
rs1017874061 Chr.19: 35142951-35142951 on Build GRCh38
rs968063094 Chr.19: 35142944-35142944 on Build GRCh38
rs536603524 Chr.19: 35142785-35142785 on Build GRCh38
rs756731949 Chr.19: 35142721-35142721 on Build GRCh38
rs543039998 Chr.19: 35142885-35142885 on Build GRCh38
rs563707390 Chr.19: 35142851-35142851 on Build GRCh38
rs912234334 Chr.19: 35142627-35142627 on Build GRCh38
rs549307609 Chr.19: 35142925-35142925 on Build GRCh38
rs1135396 Chr.19: 35142746-35142746 on Build GRCh38
rs753170408 Chr.19: 35142707-35142707 on Build GRCh38
rs919386077 Chr.19: 35142977-35142977 on Build GRCh38
rs1004874934 Chr.19: 35142753-35142753 on Build GRCh38
rs149407483 Chr.19: 35142697-35142697 on Build GRCh38
rs966665941 Chr.19: 35142953-35142953 on Build GRCh38
rs950508192 Chr.19: 35143016-35143016 on Build GRCh38
rs548795635 Chr.19: 35142584-35142584 on Build GRCh38
rs754736922 Chr.19: 35142767-35142767 on Build GRCh38
rs1673004 Chr.19: 35142599-35142599 on Build GRCh38
rs777617045 Chr.19: 35142712-35142712 on Build GRCh38
rs765705537 Chr.19: 35142791-35142791 on Build GRCh38
rs369430419 Chr.19: 35142750-35142750 on Build GRCh38
rs368655960 Chr.19: 35142601-35142601 on Build GRCh38
rs771818590 Chr.19: 35142674-35142674 on Build GRCh38
rs371865318 Chr.19: 35142652-35142652 on Build GRCh38
rs947652688 Chr.19: 35142974-35142974 on Build GRCh38
rs934355177 Chr.19: 35142913-35142913 on Build GRCh38
rs755587482 Chr.19: 35142699-35142699 on Build GRCh38
rs535283242 Chr.19: 35142743-35142743 on Build GRCh38
rs1057455 Chr.19: 35142955-35142955 on Build GRCh38
rs1002736826 Chr.19: 35142857-35142857 on Build GRCh38
rs976638962 Chr.19: 35142715-35142715 on Build GRCh38

View Full Product Details