Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38729823-38730312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTN4
Entrez Gene ID
81
Gene Name
actinin alpha 4
Gene Aliases
ACTININ-4, FSGS, FSGS1
UniGene
Hs.270291
Gene Chromosome Location
Chr.19: 38647616-38730532 on Build GRCh38
Transcripts
NM_001322033.1 NM_004924.5 XM_005259281.4 XM_006723406.2 XM_017027331.1 NM_144691.4

Gene Symbol
CAPN12
Entrez Gene ID
147968
Gene Name
calpain 12
Gene Aliases
-
UniGene
Hs.731775
Gene Chromosome Location
Chr.19: 38730124-38744474 on Build GRCh38
Transcripts
NM_001322033.1 NM_004924.5 XM_005259281.4 XM_006723406.2 XM_017027331.1 NM_144691.4

SNP Detail

SNP ID Chromosome Location
rs920474253 Chr.19: 38729968-38729968 on Build GRCh38
rs894409309 Chr.19: 38730282-38730282 on Build GRCh38
rs1010691720 Chr.19: 38730121-38730151 on Build GRCh38
rs908952655 Chr.19: 38730252-38730252 on Build GRCh38
rs191237993 Chr.19: 38730008-38730008 on Build GRCh38
rs916881931 Chr.19: 38730112-38730112 on Build GRCh38
rs935484310 Chr.19: 38730012-38730012 on Build GRCh38
rs372527421 Chr.19: 38729923-38729923 on Build GRCh38
rs1055713748 Chr.19: 38730281-38730281 on Build GRCh38
rs1049150087 Chr.19: 38730276-38730276 on Build GRCh38
rs1015160355 Chr.19: 38729954-38729955 on Build GRCh38
rs774242236 Chr.19: 38730220-38730220 on Build GRCh38
rs936969185 Chr.19: 38729856-38729856 on Build GRCh38
rs747938382 Chr.19: 38730238-38730238 on Build GRCh38
rs879463051 Chr.19: 38729920-38729926 on Build GRCh38
rs866599301 Chr.19: 38730104-38730104 on Build GRCh38
rs958633968 Chr.19: 38730176-38730177 on Build GRCh38
rs984444025 Chr.19: 38730196-38730196 on Build GRCh38
rs895526235 Chr.19: 38729877-38729878 on Build GRCh38
rs572186555 Chr.19: 38730045-38730045 on Build GRCh38
rs1027610760 Chr.19: 38730091-38730102 on Build GRCh38
rs201057110 Chr.19: 38730089-38730089 on Build GRCh38
rs915529630 Chr.19: 38730151-38730154 on Build GRCh38
rs570518830 Chr.19: 38730016-38730016 on Build GRCh38
rs1053879657 Chr.19: 38730147-38730152 on Build GRCh38
rs74176459 Chr.19: 38730130-38730131 on Build GRCh38
rs1042134274 Chr.19: 38730122-38730146 on Build GRCh38
rs971846787 Chr.19: 38730259-38730259 on Build GRCh38
rs912115569 Chr.19: 38730258-38730258 on Build GRCh38
rs935505298 Chr.19: 38730140-38730140 on Build GRCh38
rs917038052 Chr.19: 38729978-38729978 on Build GRCh38
rs548648036 Chr.19: 38729957-38729957 on Build GRCh38
rs971479178 Chr.19: 38730202-38730203 on Build GRCh38
rs761698176 Chr.19: 38730044-38730044 on Build GRCh38
rs541743616 Chr.19: 38730263-38730263 on Build GRCh38
rs924395714 Chr.19: 38730207-38730208 on Build GRCh38
rs949616203 Chr.19: 38729880-38729880 on Build GRCh38
rs1137116 Chr.19: 38730114-38730114 on Build GRCh38
rs10034 Chr.19: 38730068-38730068 on Build GRCh38
rs143818437 Chr.19: 38730145-38730148 on Build GRCh38
rs1032532128 Chr.19: 38729911-38729917 on Build GRCh38
rs377014257 Chr.19: 38730129-38730130 on Build GRCh38
rs878970245 Chr.19: 38730131-38730131 on Build GRCh38
rs1012856657 Chr.19: 38730177-38730180 on Build GRCh38
rs1053973072 Chr.19: 38730013-38730013 on Build GRCh38
rs1046581913 Chr.19: 38729988-38729990 on Build GRCh38
rs576029011 Chr.19: 38730111-38730111 on Build GRCh38
rs958602686 Chr.19: 38730097-38730103 on Build GRCh38
rs548118854 Chr.19: 38729953-38729953 on Build GRCh38
rs897938881 Chr.19: 38730266-38730266 on Build GRCh38
rs201758431 Chr.19: 38730155-38730155 on Build GRCh38
rs973192137 Chr.19: 38730076-38730076 on Build GRCh38
rs944741990 Chr.19: 38730262-38730262 on Build GRCh38
rs755979185 Chr.19: 38729894-38729894 on Build GRCh38
rs186792035 Chr.19: 38729983-38729983 on Build GRCh38
rs566253447 Chr.19: 38729954-38729954 on Build GRCh38
rs897905002 Chr.19: 38730173-38730174 on Build GRCh38
rs936990535 Chr.19: 38729970-38729970 on Build GRCh38
rs117199570 Chr.19: 38729897-38729897 on Build GRCh38
rs770253393 Chr.19: 38729994-38729995 on Build GRCh38
rs1036855812 Chr.19: 38730167-38730168 on Build GRCh38
rs936537796 Chr.19: 38730086-38730086 on Build GRCh38
rs868789894 Chr.19: 38730062-38730062 on Build GRCh38
rs74176458 Chr.19: 38730091-38730091 on Build GRCh38
rs1024196524 Chr.19: 38730195-38730196 on Build GRCh38
rs374190850 Chr.19: 38730060-38730060 on Build GRCh38
rs538534458 Chr.19: 38730023-38730023 on Build GRCh38
rs1014988450 Chr.19: 38730067-38730067 on Build GRCh38
rs890473478 Chr.19: 38730155-38730157 on Build GRCh38
rs989637033 Chr.19: 38730214-38730215 on Build GRCh38
rs532185985 Chr.19: 38730261-38730261 on Build GRCh38
rs949825195 Chr.19: 38729987-38729987 on Build GRCh38
rs948024889 Chr.19: 38730292-38730292 on Build GRCh38
rs1051135194 Chr.19: 38730257-38730258 on Build GRCh38
rs753606467 Chr.19: 38730175-38730176 on Build GRCh38
rs564893795 Chr.19: 38729921-38729921 on Build GRCh38
rs954957025 Chr.19: 38729937-38729937 on Build GRCh38
rs74584773 Chr.19: 38729976-38729976 on Build GRCh38
rs1028943442 Chr.19: 38730028-38730028 on Build GRCh38
rs139090612 Chr.19: 38730256-38730256 on Build GRCh38
rs768747977 Chr.19: 38730194-38730194 on Build GRCh38
rs971063253 Chr.19: 38730116-38730124 on Build GRCh38
rs866237477 Chr.19: 38729962-38729962 on Build GRCh38
rs996100633 Chr.19: 38729921-38729922 on Build GRCh38
rs1009380880 Chr.19: 38730047-38730047 on Build GRCh38
rs772909923 Chr.19: 38730267-38730267 on Build GRCh38
rs1046991196 Chr.19: 38729888-38729894 on Build GRCh38
rs932224337 Chr.19: 38730154-38730155 on Build GRCh38
rs560845751 Chr.19: 38729892-38729892 on Build GRCh38
rs553824510 Chr.19: 38730024-38730024 on Build GRCh38
rs14091 Chr.19: 38729941-38729941 on Build GRCh38
rs953044773 Chr.19: 38730191-38730191 on Build GRCh38
rs957111552 Chr.19: 38730209-38730211 on Build GRCh38
rs563858750 Chr.19: 38730162-38730162 on Build GRCh38
rs890576100 Chr.19: 38730041-38730041 on Build GRCh38
rs543546312 Chr.19: 38730158-38730158 on Build GRCh38
rs148908950 Chr.19: 38730169-38730172 on Build GRCh38
rs543406206 Chr.19: 38729904-38729904 on Build GRCh38
rs533178839 Chr.19: 38729858-38729858 on Build GRCh38
rs1027474961 Chr.19: 38730175-38730175 on Build GRCh38
rs530258779 Chr.19: 38729956-38729956 on Build GRCh38
rs759092967 Chr.19: 38730203-38730207 on Build GRCh38

View Full Product Details