Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54609185-54609715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT11
Entrez Gene ID
26027
Gene Name
acyl-CoA thioesterase 11
Gene Aliases
BFIT, STARD14, THEA, THEM1
UniGene
Hs.745173
Gene Chromosome Location
Chr.1: 54548134-54634744 on Build GRCh38
Transcripts
NM_015547.3 NM_147161.3 NM_176782.2 XM_006710599.3

Gene Symbol
FAM151A
Entrez Gene ID
338094
Gene Name
family with sequence similarity 151 member A
Gene Aliases
C1orf179
UniGene
Hs.234786
Gene Chromosome Location
Chr.1: 54609177-54623527 on Build GRCh38
Transcripts
NM_015547.3 NM_147161.3 NM_176782.2 XM_006710599.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2193852 c.1408G>T p.V470F Chr.1: 54609618-54609618 on Build GRCh38
7164648 c.1405G>A p.E469K Chr.1: 54609621-54609621 on Build GRCh38
3977764 c.1530C>A p.F510L Chr.1: 54609496-54609496 on Build GRCh38
4682613 c.1600delC p.R534fs*18 Chr.1: 54609426-54609426 on Build GRCh38
3997588 c.1577G>C p.G526A Chr.1: 54609449-54609449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747053558 Chr.1: 54609444-54609444 on Build GRCh38
rs530182458 Chr.1: 54609387-54609387 on Build GRCh38
rs11206394 Chr.1: 54609449-54609449 on Build GRCh38
rs773283147 Chr.1: 54609356-54609356 on Build GRCh38
rs778700519 Chr.1: 54609425-54609425 on Build GRCh38
rs768881428 Chr.1: 54609230-54609230 on Build GRCh38
rs745451468 Chr.1: 54609450-54609466 on Build GRCh38
rs373886982 Chr.1: 54609644-54609644 on Build GRCh38
rs768766573 Chr.1: 54609570-54609585 on Build GRCh38
rs2664060 Chr.1: 54609423-54609423 on Build GRCh38
rs145140282 Chr.1: 54609556-54609556 on Build GRCh38
rs751175546 Chr.1: 54609302-54609302 on Build GRCh38
rs749645766 Chr.1: 54609590-54609590 on Build GRCh38
rs370276349 Chr.1: 54609578-54609578 on Build GRCh38
rs935990821 Chr.1: 54609373-54609374 on Build GRCh38
rs1026394905 Chr.1: 54609464-54609464 on Build GRCh38
rs762970177 Chr.1: 54609613-54609613 on Build GRCh38
rs545983046 Chr.1: 54609220-54609220 on Build GRCh38
rs201295487 Chr.1: 54609347-54609347 on Build GRCh38
rs778540176 Chr.1: 54609446-54609447 on Build GRCh38
rs3179918 Chr.1: 54609441-54609441 on Build GRCh38
rs932048554 Chr.1: 54609403-54609418 on Build GRCh38
rs989748313 Chr.1: 54609405-54609405 on Build GRCh38
rs748601727 Chr.1: 54609481-54609482 on Build GRCh38
rs749916550 Chr.1: 54609313-54609313 on Build GRCh38
rs777318239 Chr.1: 54609385-54609385 on Build GRCh38
rs774404535 Chr.1: 54609232-54609232 on Build GRCh38
rs758167465 Chr.1: 54609531-54609531 on Build GRCh38
rs757537816 Chr.1: 54609642-54609642 on Build GRCh38
rs756840814 Chr.1: 54609254-54609254 on Build GRCh38
rs763297462 Chr.1: 54609485-54609485 on Build GRCh38
rs941343958 Chr.1: 54609471-54609471 on Build GRCh38
rs374539969 Chr.1: 54609424-54609424 on Build GRCh38
rs149944281 Chr.1: 54609446-54609446 on Build GRCh38
rs940614114 Chr.1: 54609297-54609297 on Build GRCh38
rs759408794 Chr.1: 54609519-54609519 on Build GRCh38
rs146657009 Chr.1: 54609536-54609536 on Build GRCh38
rs200946242 Chr.1: 54609258-54609258 on Build GRCh38
rs1047501126 Chr.1: 54609285-54609285 on Build GRCh38
rs754240542 Chr.1: 54609289-54609289 on Build GRCh38
rs543957068 Chr.1: 54609573-54609573 on Build GRCh38
rs200756038 Chr.1: 54609684-54609684 on Build GRCh38
rs2289015 Chr.1: 54609389-54609389 on Build GRCh38
rs747114381 Chr.1: 54609358-54609358 on Build GRCh38
rs368473935 Chr.1: 54609660-54609660 on Build GRCh38
rs768857868 Chr.1: 54609594-54609594 on Build GRCh38
rs368153467 Chr.1: 54609612-54609612 on Build GRCh38
rs551901825 Chr.1: 54609652-54609652 on Build GRCh38
rs765581239 Chr.1: 54609226-54609233 on Build GRCh38
rs771738348 Chr.1: 54609525-54609527 on Build GRCh38
rs200747808 Chr.1: 54609332-54609332 on Build GRCh38
rs750636549 Chr.1: 54609227-54609232 on Build GRCh38
rs748818498 Chr.1: 54609427-54609476 on Build GRCh38
rs113109924 Chr.1: 54609213-54609213 on Build GRCh38
rs376623047 Chr.1: 54609504-54609504 on Build GRCh38
rs199668243 Chr.1: 54609417-54609417 on Build GRCh38
rs764084146 Chr.1: 54609223-54609224 on Build GRCh38
rs781648110 Chr.1: 54609425-54609426 on Build GRCh38
rs578112279 Chr.1: 54609527-54609527 on Build GRCh38
rs753347392 Chr.1: 54609515-54609515 on Build GRCh38
rs368761147 Chr.1: 54609219-54609219 on Build GRCh38
rs201744943 Chr.1: 54609251-54609251 on Build GRCh38
rs780590432 Chr.1: 54609255-54609255 on Build GRCh38
rs199740522 Chr.1: 54609305-54609305 on Build GRCh38
rs759391330 Chr.1: 54609400-54609400 on Build GRCh38
rs751940506 Chr.1: 54609366-54609370 on Build GRCh38
rs201640566 Chr.1: 54609661-54609661 on Build GRCh38
rs146179147 Chr.1: 54609388-54609388 on Build GRCh38
rs1053179488 Chr.1: 54609222-54609222 on Build GRCh38
rs758796531 Chr.1: 54609313-54609332 on Build GRCh38
rs775911803 Chr.1: 54609480-54609480 on Build GRCh38
rs899023119 Chr.1: 54609309-54609309 on Build GRCh38
rs761175244 Chr.1: 54609282-54609282 on Build GRCh38
rs899076786 Chr.1: 54609526-54609526 on Build GRCh38
rs4356095 Chr.1: 54609514-54609514 on Build GRCh38
rs747906860 Chr.1: 54609431-54609431 on Build GRCh38
rs776664272 Chr.1: 54609607-54609608 on Build GRCh38
rs762023513 Chr.1: 54609233-54609233 on Build GRCh38
rs760347088 Chr.1: 54609507-54609507 on Build GRCh38
rs141833986 Chr.1: 54609418-54609418 on Build GRCh38
rs769599087 Chr.1: 54609427-54609427 on Build GRCh38
rs767884606 Chr.1: 54609235-54609235 on Build GRCh38
rs779194676 Chr.1: 54609678-54609678 on Build GRCh38
rs765831243 Chr.1: 54609508-54609508 on Build GRCh38
rs144626946 Chr.1: 54609333-54609333 on Build GRCh38
rs377007476 Chr.1: 54609242-54609242 on Build GRCh38
rs772185563 Chr.1: 54609348-54609348 on Build GRCh38
rs774725084 Chr.1: 54609596-54609596 on Build GRCh38
rs756713987 Chr.1: 54609438-54609473 on Build GRCh38
rs41297137 Chr.1: 54609668-54609668 on Build GRCh38
rs778591642 Chr.1: 54609338-54609338 on Build GRCh38
rs371266614 Chr.1: 54609606-54609606 on Build GRCh38
rs150959157 Chr.1: 54609316-54609316 on Build GRCh38
rs760043558 Chr.1: 54609292-54609292 on Build GRCh38
rs775212406 Chr.1: 54609534-54609534 on Build GRCh38
rs776895107 Chr.1: 54609225-54609226 on Build GRCh38
rs746792207 Chr.1: 54609558-54609558 on Build GRCh38
rs779769189 Chr.1: 54609259-54609259 on Build GRCh38
rs761473627 Chr.1: 54609401-54609401 on Build GRCh38
rs772138737 Chr.1: 54609269-54609269 on Build GRCh38
rs374971961 Chr.1: 54609263-54609263 on Build GRCh38
rs569444564 Chr.1: 54609696-54609696 on Build GRCh38
rs767136119 Chr.1: 54609408-54609408 on Build GRCh38
rs748358204 Chr.1: 54609345-54609345 on Build GRCh38
rs151326879 Chr.1: 54609618-54609618 on Build GRCh38
rs989681583 Chr.1: 54609245-54609245 on Build GRCh38
rs746311498 Chr.1: 54609460-54609460 on Build GRCh38
rs759114756 Chr.1: 54609607-54609607 on Build GRCh38
rs761106294 Chr.1: 54609241-54609241 on Build GRCh38
rs767004485 Chr.1: 54609288-54609288 on Build GRCh38
rs766986695 Chr.1: 54609307-54609307 on Build GRCh38
rs761897488 Chr.1: 54609237-54609237 on Build GRCh38
rs532834434 Chr.1: 54609689-54609689 on Build GRCh38
rs758065053 Chr.1: 54609246-54609246 on Build GRCh38
rs765191802 Chr.1: 54609609-54609609 on Build GRCh38
rs913221290 Chr.1: 54609656-54609656 on Build GRCh38
rs774600452 Chr.1: 54609264-54609264 on Build GRCh38
rs556361645 Chr.1: 54609597-54609597 on Build GRCh38
rs200568107 Chr.1: 54609294-54609294 on Build GRCh38
rs766597423 Chr.1: 54609320-54609329 on Build GRCh38
rs754257162 Chr.1: 54609244-54609244 on Build GRCh38
rs570204209 Chr.1: 54609337-54609337 on Build GRCh38
rs752530885 Chr.1: 54609528-54609528 on Build GRCh38
rs761772812 Chr.1: 54609265-54609265 on Build GRCh38
rs770438918 Chr.1: 54609463-54609463 on Build GRCh38
rs770332119 Chr.1: 54609574-54609574 on Build GRCh38
rs146631039 Chr.1: 54609363-54609363 on Build GRCh38
rs751792295 Chr.1: 54609543-54609543 on Build GRCh38
rs2289016 Chr.1: 54609249-54609249 on Build GRCh38
rs201630556 Chr.1: 54609605-54609605 on Build GRCh38
rs757558126 Chr.1: 54609555-54609555 on Build GRCh38
rs755504666 Chr.1: 54609389-54609389 on Build GRCh38
rs773193581 Chr.1: 54609272-54609272 on Build GRCh38
rs41297135 Chr.1: 54609426-54609426 on Build GRCh38
rs908542717 Chr.1: 54609468-54609468 on Build GRCh38
rs372058248 Chr.1: 54609632-54609632 on Build GRCh38
rs767170524 Chr.1: 54609491-54609491 on Build GRCh38
rs138994615 Chr.1: 54609692-54609692 on Build GRCh38
rs749948124 Chr.1: 54609416-54609416 on Build GRCh38
rs138880093 Chr.1: 54609679-54609679 on Build GRCh38
rs763096737 Chr.1: 54609224-54609224 on Build GRCh38
rs760141909 Chr.1: 54609386-54609386 on Build GRCh38
rs150466275 Chr.1: 54609667-54609667 on Build GRCh38
rs771184728 Chr.1: 54609445-54609445 on Build GRCh38

View Full Product Details