Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54609978-54610480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT11
Entrez Gene ID
26027
Gene Name
acyl-CoA thioesterase 11
Gene Aliases
BFIT, STARD14, THEA, THEM1
UniGene
Hs.745173
Gene Chromosome Location
Chr.1: 54548134-54634744 on Build GRCh38
Transcripts
NM_015547.3 NM_147161.3 NM_176782.2 XM_006710599.3

Gene Symbol
FAM151A
Entrez Gene ID
338094
Gene Name
family with sequence similarity 151 member A
Gene Aliases
C1orf179
UniGene
Hs.234786
Gene Chromosome Location
Chr.1: 54609177-54623527 on Build GRCh38
Transcripts
NM_015547.3 NM_147161.3 NM_176782.2 XM_006710599.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
910893 c.1045G>A p.V349I Chr.1: 54610451-54610451 on Build GRCh38
146518 c.1056C>T p.S352S Chr.1: 54610440-54610440 on Build GRCh38
5745641 c.1060A>G p.K354E Chr.1: 54610436-54610436 on Build GRCh38
3865806 c.1018C>G p.L340V Chr.1: 54610478-54610478 on Build GRCh38
5713031 c.1003_1031del29 p.P335fs*4 Chr.1: 54610465-54610493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767962722 Chr.1: 54610444-54610444 on Build GRCh38
rs964101784 Chr.1: 54610187-54610187 on Build GRCh38
rs1001851491 Chr.1: 54610079-54610079 on Build GRCh38
rs905488302 Chr.1: 54610327-54610327 on Build GRCh38
rs572219475 Chr.1: 54610340-54610340 on Build GRCh38
rs896719274 Chr.1: 54610160-54610160 on Build GRCh38
rs143545014 Chr.1: 54610048-54610048 on Build GRCh38
rs115869968 Chr.1: 54610135-54610135 on Build GRCh38
rs772244822 Chr.1: 54610049-54610049 on Build GRCh38
rs775735591 Chr.1: 54610427-54610427 on Build GRCh38
rs535219532 Chr.1: 54610104-54610104 on Build GRCh38
rs201337035 Chr.1: 54610405-54610405 on Build GRCh38
rs201589820 Chr.1: 54610448-54610448 on Build GRCh38
rs758034352 Chr.1: 54610367-54610367 on Build GRCh38
rs571999912 Chr.1: 54610244-54610244 on Build GRCh38
rs529021924 Chr.1: 54610086-54610086 on Build GRCh38
rs762319397 Chr.1: 54610443-54610443 on Build GRCh38
rs896766124 Chr.1: 54610389-54610389 on Build GRCh38
rs764963503 Chr.1: 54610457-54610457 on Build GRCh38
rs150412566 Chr.1: 54610438-54610438 on Build GRCh38
rs955609765 Chr.1: 54610115-54610115 on Build GRCh38
rs551035721 Chr.1: 54610296-54610296 on Build GRCh38
rs893258278 Chr.1: 54610028-54610029 on Build GRCh38
rs139191993 Chr.1: 54610416-54610416 on Build GRCh38
rs974205611 Chr.1: 54610407-54610407 on Build GRCh38
rs552089179 Chr.1: 54610239-54610239 on Build GRCh38
rs1056132817 Chr.1: 54610344-54610344 on Build GRCh38
rs1037206162 Chr.1: 54610393-54610393 on Build GRCh38
rs3820113 Chr.1: 54610320-54610320 on Build GRCh38
rs192037002 Chr.1: 54610439-54610439 on Build GRCh38
rs761166080 Chr.1: 54610452-54610452 on Build GRCh38
rs777171000 Chr.1: 54610385-54610385 on Build GRCh38
rs536753777 Chr.1: 54610337-54610337 on Build GRCh38
rs949938151 Chr.1: 54610269-54610269 on Build GRCh38
rs187518554 Chr.1: 54610180-54610180 on Build GRCh38
rs375570155 Chr.1: 54610224-54610224 on Build GRCh38
rs1043453980 Chr.1: 54610046-54610046 on Build GRCh38
rs757146308 Chr.1: 54610401-54610401 on Build GRCh38
rs141458641 Chr.1: 54610422-54610422 on Build GRCh38
rs747092521 Chr.1: 54610139-54610139 on Build GRCh38
rs1001282266 Chr.1: 54610005-54610005 on Build GRCh38
rs911836164 Chr.1: 54610311-54610311 on Build GRCh38
rs746927156 Chr.1: 54610390-54610390 on Build GRCh38
rs988427880 Chr.1: 54610332-54610332 on Build GRCh38
rs370504805 Chr.1: 54610341-54610341 on Build GRCh38
rs930447318 Chr.1: 54610241-54610241 on Build GRCh38
rs547175365 Chr.1: 54610212-54610212 on Build GRCh38
rs776735122 Chr.1: 54610181-54610181 on Build GRCh38
rs549265919 Chr.1: 54610088-54610088 on Build GRCh38
rs1033183959 Chr.1: 54610018-54610018 on Build GRCh38
rs373528506 Chr.1: 54610268-54610268 on Build GRCh38
rs35287092 Chr.1: 54610017-54610017 on Build GRCh38
rs1008678067 Chr.1: 54610451-54610451 on Build GRCh38
rs937699631 Chr.1: 54610343-54610343 on Build GRCh38
rs898687109 Chr.1: 54610398-54610398 on Build GRCh38
rs756770476 Chr.1: 54610390-54610390 on Build GRCh38
rs913685301 Chr.1: 54610037-54610037 on Build GRCh38
rs770560230 Chr.1: 54610197-54610197 on Build GRCh38
rs986799218 Chr.1: 54610310-54610310 on Build GRCh38
rs41297139 Chr.1: 54610041-54610041 on Build GRCh38
rs115202613 Chr.1: 54610338-54610338 on Build GRCh38
rs148333188 Chr.1: 54610226-54610226 on Build GRCh38
rs4926650 Chr.1: 54610440-54610440 on Build GRCh38
rs878886168 Chr.1: 54610276-54610276 on Build GRCh38

View Full Product Details