Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 54114981-54115472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACYP2
Entrez Gene ID
98
Gene Name
acylphosphatase 2
Gene Aliases
ACYM, ACYP
UniGene
Hs.516173
Gene Chromosome Location
Chr.2: 53970838-54305300 on Build GRCh38
Transcripts
NM_001320586.1 NM_001320587.1 NM_001320589.1 NM_001320590.1 NM_138448.3 XM_017005406.1 XM_017005407.1 XM_017005408.1 XM_017005409.1 XM_017005410.1 XM_017005411.1 XM_017005412.1 XM_017005413.1

SNP Detail

SNP ID Chromosome Location
rs998862066 Chr.2: 54115101-54115101 on Build GRCh38
rs999351820 Chr.2: 54115337-54115337 on Build GRCh38
rs1008875613 Chr.2: 54115387-54115387 on Build GRCh38
rs569062965 Chr.2: 54115367-54115367 on Build GRCh38
rs537707722 Chr.2: 54115390-54115390 on Build GRCh38
rs1053096826 Chr.2: 54115361-54115361 on Build GRCh38
rs777400501 Chr.2: 54115010-54115010 on Build GRCh38
rs916196935 Chr.2: 54115270-54115270 on Build GRCh38
rs1051285037 Chr.2: 54115204-54115204 on Build GRCh38
rs1017882978 Chr.2: 54115240-54115240 on Build GRCh38
rs956587578 Chr.2: 54115117-54115117 on Build GRCh38
rs555412923 Chr.2: 54115055-54115055 on Build GRCh38
rs770383621 Chr.2: 54115087-54115087 on Build GRCh38
rs941214394 Chr.2: 54115148-54115148 on Build GRCh38
rs978242269 Chr.2: 54115276-54115276 on Build GRCh38
rs890820032 Chr.2: 54115206-54115206 on Build GRCh38
rs1043135478 Chr.2: 54115032-54115032 on Build GRCh38
rs912286527 Chr.2: 54115428-54115428 on Build GRCh38
rs904637300 Chr.2: 54115063-54115063 on Build GRCh38
rs975681671 Chr.2: 54115150-54115150 on Build GRCh38
rs746417497 Chr.2: 54115064-54115064 on Build GRCh38
rs59235664 Chr.2: 54115384-54115384 on Build GRCh38
rs547544581 Chr.2: 54115217-54115217 on Build GRCh38
rs894666742 Chr.2: 54115017-54115017 on Build GRCh38
rs978507327 Chr.2: 54115045-54115045 on Build GRCh38
rs900733272 Chr.2: 54115241-54115241 on Build GRCh38
rs544469074 Chr.2: 54115247-54115247 on Build GRCh38
rs938892760 Chr.2: 54115218-54115218 on Build GRCh38
rs530142696 Chr.2: 54115296-54115297 on Build GRCh38
rs144662525 Chr.2: 54115273-54115273 on Build GRCh38
rs74817071 Chr.2: 54115228-54115228 on Build GRCh38
rs769735733 Chr.2: 54115255-54115256 on Build GRCh38
rs916499329 Chr.2: 54115348-54115348 on Build GRCh38
rs1035498823 Chr.2: 54115281-54115281 on Build GRCh38
rs929774782 Chr.2: 54115184-54115184 on Build GRCh38
rs1038165888 Chr.2: 54115151-54115151 on Build GRCh38
rs963827982 Chr.2: 54115132-54115132 on Build GRCh38
rs142322749 Chr.2: 54115114-54115114 on Build GRCh38
rs942363318 Chr.2: 54115442-54115442 on Build GRCh38
rs974389173 Chr.2: 54115380-54115380 on Build GRCh38
rs952466446 Chr.2: 54115440-54115440 on Build GRCh38
rs990756233 Chr.2: 54115342-54115342 on Build GRCh38
rs568268757 Chr.2: 54115168-54115168 on Build GRCh38
rs1027241042 Chr.2: 54115432-54115432 on Build GRCh38
rs1008111327 Chr.2: 54115118-54115118 on Build GRCh38
rs896905358 Chr.2: 54115169-54115173 on Build GRCh38
rs551800077 Chr.2: 54115100-54115100 on Build GRCh38
rs575616574 Chr.2: 54115195-54115195 on Build GRCh38
rs996652918 Chr.2: 54115423-54115423 on Build GRCh38
rs866700493 Chr.2: 54115172-54115172 on Build GRCh38
rs928022976 Chr.2: 54115094-54115094 on Build GRCh38
rs566694554 Chr.2: 54115258-54115258 on Build GRCh38
rs1013116540 Chr.2: 54115031-54115031 on Build GRCh38
rs1019477393 Chr.2: 54115396-54115396 on Build GRCh38
rs904565075 Chr.2: 54115291-54115291 on Build GRCh38
rs1019381581 Chr.2: 54115128-54115128 on Build GRCh38
rs757159892 Chr.2: 54115248-54115248 on Build GRCh38
rs549308996 Chr.2: 54115278-54115278 on Build GRCh38
rs952572949 Chr.2: 54115190-54115190 on Build GRCh38
rs768761841 Chr.2: 54115412-54115412 on Build GRCh38

View Full Product Details