Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 54254163-54254672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACYP2
Entrez Gene ID
98
Gene Name
acylphosphatase 2
Gene Aliases
ACYM, ACYP
UniGene
Hs.516173
Gene Chromosome Location
Chr.2: 53970838-54305300 on Build GRCh38
Transcripts
NM_001320586.1 NM_001320587.1 NM_001320588.1 NM_001320589.1 NM_138448.3 XM_017005411.1 XM_017005412.1 XM_017005413.1 NM_001003937.2

Gene Symbol
TSPYL6
Entrez Gene ID
388951
Gene Name
TSPY like 6
Gene Aliases
-
UniGene
Hs.620508
Gene Chromosome Location
Chr.2: 54253178-54256272 on Build GRCh38
Transcripts
NM_001320586.1 NM_001320587.1 NM_001320588.1 NM_001320589.1 NM_138448.3 XM_017005411.1 XM_017005412.1 XM_017005413.1 NM_001003937.2

SNP Detail

SNP ID Chromosome Location
rs976930813 Chr.2: 54254433-54254433 on Build GRCh38
rs1003866615 Chr.2: 54254529-54254529 on Build GRCh38
rs843705 Chr.2: 54254455-54254455 on Build GRCh38
rs1021211375 Chr.2: 54254412-54254412 on Build GRCh38
rs368927774 Chr.2: 54254618-54254618 on Build GRCh38
rs147238101 Chr.2: 54254238-54254238 on Build GRCh38
rs887707647 Chr.2: 54254269-54254269 on Build GRCh38
rs921357975 Chr.2: 54254229-54254229 on Build GRCh38
rs1002291143 Chr.2: 54254367-54254367 on Build GRCh38
rs866084988 Chr.2: 54254227-54254227 on Build GRCh38
rs1036158616 Chr.2: 54254315-54254315 on Build GRCh38
rs1032432064 Chr.2: 54254594-54254594 on Build GRCh38
rs767321698 Chr.2: 54254624-54254624 on Build GRCh38
rs548687428 Chr.2: 54254587-54254587 on Build GRCh38
rs140717255 Chr.2: 54254248-54254248 on Build GRCh38
rs1025167231 Chr.2: 54254347-54254347 on Build GRCh38
rs762893723 Chr.2: 54254369-54254369 on Build GRCh38
rs1013025174 Chr.2: 54254432-54254432 on Build GRCh38
rs143277831 Chr.2: 54254319-54254319 on Build GRCh38
rs893512876 Chr.2: 54254609-54254609 on Build GRCh38
rs1041186202 Chr.2: 54254224-54254224 on Build GRCh38
rs984529322 Chr.2: 54254488-54254488 on Build GRCh38
rs144863037 Chr.2: 54254405-54254405 on Build GRCh38
rs10165485 Chr.2: 54254499-54254499 on Build GRCh38
rs181890516 Chr.2: 54254447-54254447 on Build GRCh38
rs933871014 Chr.2: 54254338-54254338 on Build GRCh38
rs186263570 Chr.2: 54254548-54254548 on Build GRCh38
rs951068622 Chr.2: 54254521-54254521 on Build GRCh38
rs865862818 Chr.2: 54254616-54254616 on Build GRCh38
rs1001922085 Chr.2: 54254591-54254591 on Build GRCh38
rs571999793 Chr.2: 54254377-54254377 on Build GRCh38
rs940017037 Chr.2: 54254517-54254517 on Build GRCh38
rs1054436376 Chr.2: 54254642-54254642 on Build GRCh38
rs528545944 Chr.2: 54254582-54254582 on Build GRCh38
rs190882058 Chr.2: 54254326-54254326 on Build GRCh38
rs148559721 Chr.2: 54254536-54254536 on Build GRCh38
rs921360796 Chr.2: 54254438-54254438 on Build GRCh38
rs1036549921 Chr.2: 54254399-54254415 on Build GRCh38
rs151093608 Chr.2: 54254644-54254644 on Build GRCh38
rs534739502 Chr.2: 54254402-54254402 on Build GRCh38
rs907375606 Chr.2: 54254490-54254490 on Build GRCh38
rs180768415 Chr.2: 54254193-54254193 on Build GRCh38
rs1006052249 Chr.2: 54254270-54254270 on Build GRCh38
rs6735998 Chr.2: 54254362-54254362 on Build GRCh38
rs924564485 Chr.2: 54254384-54254384 on Build GRCh38
rs559898199 Chr.2: 54254549-54254549 on Build GRCh38
rs944114695 Chr.2: 54254218-54254218 on Build GRCh38
rs949248706 Chr.2: 54254560-54254560 on Build GRCh38
rs921113036 Chr.2: 54254336-54254336 on Build GRCh38
rs186455695 Chr.2: 54254245-54254245 on Build GRCh38
rs550079954 Chr.2: 54254290-54254290 on Build GRCh38
rs909627559 Chr.2: 54254288-54254288 on Build GRCh38
rs924541264 Chr.2: 54254622-54254622 on Build GRCh38
rs768411979 Chr.2: 54254424-54254424 on Build GRCh38
rs750137989 Chr.2: 54254628-54254628 on Build GRCh38
rs149522515 Chr.2: 54254197-54254197 on Build GRCh38
rs142939043 Chr.2: 54254620-54254620 on Build GRCh38

View Full Product Details