Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 54816570-54817069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EML6
Entrez Gene ID
400954
Gene Name
echinoderm microtubule associated protein like 6
Gene Aliases
-
UniGene
Hs.656692
Gene Chromosome Location
Chr.2: 54723467-54972908 on Build GRCh38
Transcripts
NM_001039753.2 XM_017004098.1 XM_017004099.1 XM_017004100.1 XM_017004102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7280299 c.375G>A p.G125G Chr.2: 54816804-54816804 on Build GRCh38
1205278 c.407A>T p.D136V Chr.2: 54816836-54816836 on Build GRCh38
6226580 c.432G>A p.A144A Chr.2: 54816861-54816861 on Build GRCh38
1408584 c.413G>T p.R138M Chr.2: 54816842-54816842 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775581352 Chr.2: 54816882-54816882 on Build GRCh38
rs972146332 Chr.2: 54817007-54817007 on Build GRCh38
rs750264327 Chr.2: 54816958-54816958 on Build GRCh38
rs965422561 Chr.2: 54816779-54816779 on Build GRCh38
rs765100524 Chr.2: 54816921-54816921 on Build GRCh38
rs755360982 Chr.2: 54816821-54816821 on Build GRCh38
rs970823693 Chr.2: 54816798-54816799 on Build GRCh38
rs533861152 Chr.2: 54816647-54816647 on Build GRCh38
rs942902529 Chr.2: 54816875-54816875 on Build GRCh38
rs1046660412 Chr.2: 54816869-54816869 on Build GRCh38
rs953989776 Chr.2: 54816804-54816804 on Build GRCh38
rs186281872 Chr.2: 54816870-54816870 on Build GRCh38
rs370159775 Chr.2: 54816628-54816628 on Build GRCh38
rs563871558 Chr.2: 54817018-54817018 on Build GRCh38
rs1027976598 Chr.2: 54816947-54816947 on Build GRCh38
rs766916949 Chr.2: 54816776-54816776 on Build GRCh38
rs2578711 Chr.2: 54816661-54816661 on Build GRCh38
rs749748928 Chr.2: 54816629-54816629 on Build GRCh38
rs552165813 Chr.2: 54816631-54816631 on Build GRCh38
rs952966555 Chr.2: 54816842-54816842 on Build GRCh38
rs749042204 Chr.2: 54816902-54816902 on Build GRCh38
rs900810780 Chr.2: 54816925-54816925 on Build GRCh38
rs763718947 Chr.2: 54816846-54816846 on Build GRCh38
rs370983931 Chr.2: 54816914-54816914 on Build GRCh38
rs1012522810 Chr.2: 54816985-54816985 on Build GRCh38
rs756771011 Chr.2: 54816847-54816847 on Build GRCh38
rs919231664 Chr.2: 54816856-54816856 on Build GRCh38
rs889695116 Chr.2: 54816953-54816953 on Build GRCh38
rs902144692 Chr.2: 54816944-54816944 on Build GRCh38
rs369858624 Chr.2: 54816822-54816822 on Build GRCh38
rs1042065084 Chr.2: 54816733-54816736 on Build GRCh38
rs751597001 Chr.2: 54817028-54817028 on Build GRCh38
rs144662245 Chr.2: 54816600-54816600 on Build GRCh38
rs370463321 Chr.2: 54816873-54816873 on Build GRCh38
rs545074589 Chr.2: 54816767-54816767 on Build GRCh38
rs556298540 Chr.2: 54816683-54816683 on Build GRCh38
rs373616236 Chr.2: 54816893-54816893 on Build GRCh38
rs902445601 Chr.2: 54816735-54816735 on Build GRCh38
rs368055266 Chr.2: 54816945-54816945 on Build GRCh38
rs769549311 Chr.2: 54816876-54816876 on Build GRCh38
rs561575160 Chr.2: 54816946-54816946 on Build GRCh38
rs113913794 Chr.2: 54816666-54816666 on Build GRCh38
rs1028212277 Chr.2: 54816788-54816788 on Build GRCh38
rs760117560 Chr.2: 54816730-54816730 on Build GRCh38
rs993803235 Chr.2: 54816754-54816767 on Build GRCh38
rs570054729 Chr.2: 54816595-54816595 on Build GRCh38
rs1032183231 Chr.2: 54816746-54816746 on Build GRCh38
rs1000762491 Chr.2: 54816736-54816736 on Build GRCh38
rs1042096029 Chr.2: 54816936-54816936 on Build GRCh38
rs754147048 Chr.2: 54816784-54816784 on Build GRCh38
rs889914324 Chr.2: 54816620-54816620 on Build GRCh38
rs761940801 Chr.2: 54816751-54816751 on Build GRCh38
rs536063460 Chr.2: 54816682-54816682 on Build GRCh38
rs576071812 Chr.2: 54816734-54816734 on Build GRCh38
rs542568677 Chr.2: 54816703-54816703 on Build GRCh38
rs543673834 Chr.2: 54816994-54816994 on Build GRCh38
rs541784728 Chr.2: 54816871-54816871 on Build GRCh38
rs767530733 Chr.2: 54816756-54816756 on Build GRCh38
rs879012483 Chr.2: 54816613-54816613 on Build GRCh38
rs949624308 Chr.2: 54816609-54816609 on Build GRCh38
rs575974835 Chr.2: 54816991-54816992 on Build GRCh38
rs750917481 Chr.2: 54816760-54816760 on Build GRCh38
rs971237607 Chr.2: 54817016-54817016 on Build GRCh38
rs1026833990 Chr.2: 54817011-54817011 on Build GRCh38
rs1009769191 Chr.2: 54816714-54816714 on Build GRCh38
rs377645256 Chr.2: 54816837-54816837 on Build GRCh38
rs961511678 Chr.2: 54816989-54816989 on Build GRCh38
rs997617994 Chr.2: 54816706-54816706 on Build GRCh38
rs190459032 Chr.2: 54816971-54816971 on Build GRCh38
rs952749701 Chr.2: 54817029-54817029 on Build GRCh38
rs1025608537 Chr.2: 54817022-54817022 on Build GRCh38
rs374066595 Chr.2: 54816834-54816834 on Build GRCh38
rs933029981 Chr.2: 54816933-54816933 on Build GRCh38
rs565719278 Chr.2: 54816646-54816646 on Build GRCh38
rs911470163 Chr.2: 54816866-54816866 on Build GRCh38
rs768602897 Chr.2: 54816909-54816909 on Build GRCh38
rs949427800 Chr.2: 54816858-54816858 on Build GRCh38
rs780413903 Chr.2: 54816893-54816894 on Build GRCh38

View Full Product Details