Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 110732682-110733187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOXL
Entrez Gene ID
55289
Gene Name
acyl-CoA oxidase-like
Gene Aliases
-
UniGene
Hs.253320
Gene Chromosome Location
Chr.2: 110732539-111118571 on Build GRCh38
Transcripts
NM_001142807.1 XM_011511404.2 XM_011511405.2 XM_011511409.1 XM_011511411.2 XM_011511413.1 XM_011511414.2 XM_011511415.2 XM_011511416.2 XM_011511419.2 XM_011511420.2 XM_011511421.2 XM_011511422.1 XM_011511423.1 XM_011511427.2 XM_011511428.2 XM_011511429.2 XM_011511432.2 XM_017004431.1 XM_017004432.1

SNP Detail

SNP ID Chromosome Location
rs114086993 Chr.2: 110732979-110732979 on Build GRCh38
rs1018331084 Chr.2: 110732823-110732823 on Build GRCh38
rs556162525 Chr.2: 110732864-110732864 on Build GRCh38
rs1029818757 Chr.2: 110733066-110733066 on Build GRCh38
rs911244814 Chr.2: 110732797-110732797 on Build GRCh38
rs754916686 Chr.2: 110732910-110732910 on Build GRCh38
rs967156902 Chr.2: 110732987-110732987 on Build GRCh38
rs752209655 Chr.2: 110733150-110733150 on Build GRCh38
rs371728493 Chr.2: 110733074-110733074 on Build GRCh38
rs572597967 Chr.2: 110732997-110732997 on Build GRCh38
rs551519491 Chr.2: 110732759-110732759 on Build GRCh38
rs868805305 Chr.2: 110732802-110732802 on Build GRCh38
rs571386071 Chr.2: 110732760-110732760 on Build GRCh38
rs1056051898 Chr.2: 110733165-110733165 on Build GRCh38
rs1034400545 Chr.2: 110732934-110732934 on Build GRCh38
rs573686001 Chr.2: 110732793-110732793 on Build GRCh38
rs554017555 Chr.2: 110732781-110732781 on Build GRCh38
rs898623473 Chr.2: 110732859-110732859 on Build GRCh38
rs534274088 Chr.2: 110732761-110732761 on Build GRCh38
rs560250257 Chr.2: 110733010-110733010 on Build GRCh38
rs766229503 Chr.2: 110732716-110732716 on Build GRCh38
rs1047247179 Chr.2: 110732876-110732876 on Build GRCh38
rs942759700 Chr.2: 110732835-110732835 on Build GRCh38
rs933284283 Chr.2: 110732791-110732791 on Build GRCh38
rs548970211 Chr.2: 110733139-110733139 on Build GRCh38
rs938896559 Chr.2: 110733161-110733161 on Build GRCh38
rs967639774 Chr.2: 110732895-110732895 on Build GRCh38
rs188180860 Chr.2: 110733005-110733005 on Build GRCh38
rs754079132 Chr.2: 110732820-110732820 on Build GRCh38
rs75508216 Chr.2: 110732860-110732860 on Build GRCh38
rs7572507 Chr.2: 110733154-110733154 on Build GRCh38
rs977241030 Chr.2: 110732990-110732990 on Build GRCh38
rs545414315 Chr.2: 110732937-110732937 on Build GRCh38
rs1047648995 Chr.2: 110733151-110733151 on Build GRCh38
rs200020093 Chr.2: 110732721-110732721 on Build GRCh38
rs576296466 Chr.2: 110732931-110732931 on Build GRCh38
rs1005508973 Chr.2: 110733033-110733033 on Build GRCh38
rs1051518029 Chr.2: 110732795-110732795 on Build GRCh38
rs998702327 Chr.2: 110733063-110733069 on Build GRCh38
rs1018278753 Chr.2: 110733035-110733035 on Build GRCh38
rs976952312 Chr.2: 110732866-110732867 on Build GRCh38
rs989909955 Chr.2: 110732740-110732740 on Build GRCh38
rs925582560 Chr.2: 110732936-110732936 on Build GRCh38
rs999095783 Chr.2: 110733102-110733102 on Build GRCh38
rs34061237 Chr.2: 110732984-110732984 on Build GRCh38
rs915671718 Chr.2: 110732944-110732944 on Build GRCh38
rs919998096 Chr.2: 110733082-110733082 on Build GRCh38
rs562481854 Chr.2: 110733141-110733141 on Build GRCh38
rs1011289088 Chr.2: 110732939-110732939 on Build GRCh38
rs1034040111 Chr.2: 110732727-110732727 on Build GRCh38
rs899918166 Chr.2: 110733051-110733051 on Build GRCh38
rs1005561495 Chr.2: 110732799-110732799 on Build GRCh38
rs938995311 Chr.2: 110732909-110732909 on Build GRCh38
rs1038426716 Chr.2: 110732841-110732841 on Build GRCh38
rs958675421 Chr.2: 110732731-110732731 on Build GRCh38
rs79918262 Chr.2: 110732702-110732702 on Build GRCh38
rs942700470 Chr.2: 110733067-110733067 on Build GRCh38
rs954764537 Chr.2: 110732999-110732999 on Build GRCh38
rs538159881 Chr.2: 110732750-110732750 on Build GRCh38
rs529002150 Chr.2: 110733119-110733119 on Build GRCh38
rs907429559 Chr.2: 110732877-110732877 on Build GRCh38
rs897676329 Chr.2: 110732998-110732998 on Build GRCh38

View Full Product Details