Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 110784524-110784959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOXL
Entrez Gene ID
55289
Gene Name
acyl-CoA oxidase-like
Gene Aliases
-
UniGene
Hs.253320
Gene Chromosome Location
Chr.2: 110732539-111118571 on Build GRCh38
Transcripts
NM_001142807.1 XM_011511404.2 XM_011511405.2 XM_011511406.2 XM_011511407.2 XM_011511409.1 XM_011511411.2 XM_011511413.1 XM_011511414.2 XM_011511415.2 XM_011511416.2 XM_011511419.2 XM_011511420.2 XM_011511421.2 XM_011511422.1 XM_011511423.1 XM_011511427.2 XM_011511428.2 XM_011511429.2 XM_011511432.2 XM_017004431.1 XM_017004432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5505740 c.95T>C p.F32S Chr.2: 110784751-110784751 on Build GRCh38
3565212 c.126C>T p.V42V Chr.2: 110784782-110784782 on Build GRCh38
6847574 c.76G>A p.A26T Chr.2: 110784732-110784732 on Build GRCh38
3565210 c.104G>A p.R35Q Chr.2: 110784760-110784760 on Build GRCh38
4301950 c.139G>A p.D47N Chr.2: 110784795-110784795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936099727 Chr.2: 110784639-110784639 on Build GRCh38
rs948691550 Chr.2: 110784568-110784568 on Build GRCh38
rs764041951 Chr.2: 110784711-110784711 on Build GRCh38
rs953214113 Chr.2: 110784607-110784607 on Build GRCh38
rs533769226 Chr.2: 110784907-110784907 on Build GRCh38
rs375666693 Chr.2: 110784794-110784794 on Build GRCh38
rs911688773 Chr.2: 110784630-110784630 on Build GRCh38
rs111623296 Chr.2: 110784856-110784856 on Build GRCh38
rs1052294150 Chr.2: 110784681-110784681 on Build GRCh38
rs758824889 Chr.2: 110784764-110784764 on Build GRCh38
rs944915037 Chr.2: 110784715-110784715 on Build GRCh38
rs200338206 Chr.2: 110784806-110784806 on Build GRCh38
rs540285573 Chr.2: 110784559-110784559 on Build GRCh38
rs780616662 Chr.2: 110784771-110784771 on Build GRCh38
rs377423131 Chr.2: 110784778-110784778 on Build GRCh38
rs757414173 Chr.2: 110784835-110784835 on Build GRCh38
rs755541055 Chr.2: 110784785-110784785 on Build GRCh38
rs775292039 Chr.2: 110784824-110784824 on Build GRCh38
rs973039287 Chr.2: 110784584-110784584 on Build GRCh38
rs921615736 Chr.2: 110784588-110784588 on Build GRCh38
rs913357488 Chr.2: 110784891-110784891 on Build GRCh38
rs761952302 Chr.2: 110784847-110784847 on Build GRCh38
rs771820899 Chr.2: 110784811-110784811 on Build GRCh38
rs887505142 Chr.2: 110784804-110784804 on Build GRCh38
rs750745087 Chr.2: 110784759-110784759 on Build GRCh38
rs891944404 Chr.2: 110784708-110784708 on Build GRCh38
rs775383528 Chr.2: 110784683-110784683 on Build GRCh38
rs750217081 Chr.2: 110784886-110784886 on Build GRCh38
rs763434995 Chr.2: 110784743-110784743 on Build GRCh38
rs989281930 Chr.2: 110784888-110784888 on Build GRCh38
rs995766650 Chr.2: 110784775-110784775 on Build GRCh38
rs745568724 Chr.2: 110784808-110784808 on Build GRCh38
rs200208829 Chr.2: 110784827-110784827 on Build GRCh38
rs1029047319 Chr.2: 110784908-110784908 on Build GRCh38
rs980106097 Chr.2: 110784617-110784617 on Build GRCh38
rs925986833 Chr.2: 110784623-110784623 on Build GRCh38
rs117512281 Chr.2: 110784724-110784724 on Build GRCh38
rs567969774 Chr.2: 110784669-110784669 on Build GRCh38
rs760643331 Chr.2: 110784707-110784707 on Build GRCh38
rs764037962 Chr.2: 110784719-110784719 on Build GRCh38
rs758735090 Chr.2: 110784857-110784857 on Build GRCh38
rs560011004 Chr.2: 110784577-110784577 on Build GRCh38
rs528834094 Chr.2: 110784598-110784598 on Build GRCh38
rs915151949 Chr.2: 110784698-110784698 on Build GRCh38
rs35792628 Chr.2: 110784588-110784589 on Build GRCh38
rs765504663 Chr.2: 110784758-110784758 on Build GRCh38
rs925954044 Chr.2: 110784870-110784870 on Build GRCh38
rs201574685 Chr.2: 110784793-110784793 on Build GRCh38
rs935130834 Chr.2: 110784634-110784634 on Build GRCh38
rs987250522 Chr.2: 110784614-110784614 on Build GRCh38
rs776369007 Chr.2: 110784773-110784775 on Build GRCh38
rs373206795 Chr.2: 110784722-110784722 on Build GRCh38
rs367701569 Chr.2: 110784797-110784797 on Build GRCh38
rs764102517 Chr.2: 110784828-110784828 on Build GRCh38
rs761208453 Chr.2: 110784852-110784853 on Build GRCh38
rs963633082 Chr.2: 110784579-110784579 on Build GRCh38

View Full Product Details