Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112783639-112784137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1A
Entrez Gene ID
3552
Gene Name
interleukin 1 alpha
Gene Aliases
IL-1A, IL1, IL1-ALPHA, IL1F1
UniGene
Hs.1722
Gene Chromosome Location
Chr.2: 112773915-112785398 on Build GRCh38
Transcripts
NM_000575.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3565380 c.36G>A p.K12K Chr.2: 112783735-112783735 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs576941251 Chr.2: 112783664-112783664 on Build GRCh38
rs1013973341 Chr.2: 112783908-112783908 on Build GRCh38
rs911915752 Chr.2: 112784034-112784034 on Build GRCh38
rs200686721 Chr.2: 112783724-112783724 on Build GRCh38
rs370724677 Chr.2: 112783709-112783709 on Build GRCh38
rs55926957 Chr.2: 112783938-112783938 on Build GRCh38
rs149164954 Chr.2: 112783769-112783769 on Build GRCh38
rs878881595 Chr.2: 112783831-112783831 on Build GRCh38
rs963647127 Chr.2: 112783944-112783944 on Build GRCh38
rs986047491 Chr.2: 112784033-112784033 on Build GRCh38
rs902259491 Chr.2: 112783740-112783740 on Build GRCh38
rs990854962 Chr.2: 112784049-112784049 on Build GRCh38
rs781201225 Chr.2: 112783812-112783812 on Build GRCh38
rs1029414701 Chr.2: 112783768-112783768 on Build GRCh38
rs966005240 Chr.2: 112784035-112784035 on Build GRCh38
rs1055417794 Chr.2: 112783669-112783669 on Build GRCh38
rs193027324 Chr.2: 112784003-112784003 on Build GRCh38
rs772492962 Chr.2: 112783719-112783719 on Build GRCh38
rs950774971 Chr.2: 112783965-112783965 on Build GRCh38
rs753418375 Chr.2: 112783731-112783731 on Build GRCh38
rs973555231 Chr.2: 112783959-112783959 on Build GRCh38
rs150033245 Chr.2: 112783730-112783730 on Build GRCh38
rs3783572 Chr.2: 112783805-112783805 on Build GRCh38
rs189583529 Chr.2: 112783915-112783915 on Build GRCh38
rs997926639 Chr.2: 112783758-112783758 on Build GRCh38
rs371037111 Chr.2: 112783761-112783761 on Build GRCh38
rs928080190 Chr.2: 112784008-112784008 on Build GRCh38
rs750126688 Chr.2: 112783764-112783764 on Build GRCh38
rs756928845 Chr.2: 112783749-112783749 on Build GRCh38
rs377394295 Chr.2: 112783728-112783728 on Build GRCh38
rs915143203 Chr.2: 112784074-112784074 on Build GRCh38
rs766861513 Chr.2: 112784085-112784085 on Build GRCh38
rs1026516963 Chr.2: 112783963-112783963 on Build GRCh38
rs771267948 Chr.2: 112784064-112784064 on Build GRCh38
rs766742636 Chr.2: 112783723-112783723 on Build GRCh38
rs937981110 Chr.2: 112784038-112784038 on Build GRCh38
rs878984761 Chr.2: 112783978-112783978 on Build GRCh38
rs748260513 Chr.2: 112783815-112783815 on Build GRCh38
rs758270558 Chr.2: 112783779-112783779 on Build GRCh38
rs761036723 Chr.2: 112783818-112783818 on Build GRCh38
rs1027481993 Chr.2: 112783969-112783969 on Build GRCh38
rs1014595523 Chr.2: 112783939-112783939 on Build GRCh38
rs779852325 Chr.2: 112783793-112783793 on Build GRCh38
rs747007652 Chr.2: 112783794-112783794 on Build GRCh38
rs773528399 Chr.2: 112783690-112783690 on Build GRCh38
rs946530818 Chr.2: 112783703-112783703 on Build GRCh38
rs973686262 Chr.2: 112783964-112783964 on Build GRCh38
rs763383903 Chr.2: 112783712-112783712 on Build GRCh38
rs1018248374 Chr.2: 112783941-112783941 on Build GRCh38

View Full Product Details