Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17104454-17104948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL17RA
Entrez Gene ID
23765
Gene Name
interleukin 17 receptor A
Gene Aliases
CANDF5, CD217, CDw217, IL-17RA, IL17R, hIL-17R
UniGene
Hs.48353
Gene Chromosome Location
Chr.22: 17084959-17115694 on Build GRCh38
Transcripts
NM_001289905.1 NM_014339.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4925871 c.930A>G p.P310P Chr.22: 17104809-17104809 on Build GRCh38
3424021 c.895G>A p.A299T Chr.22: 17104774-17104774 on Build GRCh38
1254742 c.903T>A p.V301V Chr.22: 17104782-17104782 on Build GRCh38
6684969 c.863G>A p.S288N Chr.22: 17104742-17104742 on Build GRCh38
5889419 c.931+3A>G p.? Chr.22: 17104813-17104813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201410617 Chr.22: 17104712-17104712 on Build GRCh38
rs779171527 Chr.22: 17104850-17104850 on Build GRCh38
rs531944007 Chr.22: 17104814-17104814 on Build GRCh38
rs2302519 Chr.22: 17104551-17104551 on Build GRCh38
rs377707677 Chr.22: 17104823-17104823 on Build GRCh38
rs753793215 Chr.22: 17104738-17104738 on Build GRCh38
rs778788017 Chr.22: 17104843-17104845 on Build GRCh38
rs911225585 Chr.22: 17104871-17104871 on Build GRCh38
rs1054369086 Chr.22: 17104901-17104902 on Build GRCh38
rs759393806 Chr.22: 17104807-17104807 on Build GRCh38
rs777335889 Chr.22: 17104766-17104766 on Build GRCh38
rs868241754 Chr.22: 17104784-17104784 on Build GRCh38
rs746356951 Chr.22: 17104775-17104775 on Build GRCh38
rs763154741 Chr.22: 17104732-17104732 on Build GRCh38
rs2228077 Chr.22: 17104752-17104752 on Build GRCh38
rs970625814 Chr.22: 17104611-17104611 on Build GRCh38
rs919985232 Chr.22: 17104817-17104817 on Build GRCh38
rs200422686 Chr.22: 17104778-17104778 on Build GRCh38
rs751204457 Chr.22: 17104821-17104821 on Build GRCh38
rs750705596 Chr.22: 17104827-17104827 on Build GRCh38
rs775751135 Chr.22: 17104730-17104730 on Build GRCh38
rs761989355 Chr.22: 17104797-17104797 on Build GRCh38
rs375514449 Chr.22: 17104875-17104875 on Build GRCh38
rs781718594 Chr.22: 17104721-17104721 on Build GRCh38
rs780702228 Chr.22: 17104838-17104838 on Build GRCh38
rs931308887 Chr.22: 17104654-17104654 on Build GRCh38
rs902781334 Chr.22: 17104880-17104880 on Build GRCh38
rs149220878 Chr.22: 17104780-17104780 on Build GRCh38
rs752631425 Chr.22: 17104813-17104813 on Build GRCh38
rs767379269 Chr.22: 17104916-17104916 on Build GRCh38
rs374537654 Chr.22: 17104773-17104773 on Build GRCh38
rs765097002 Chr.22: 17104808-17104808 on Build GRCh38
rs721930 Chr.22: 17104918-17104918 on Build GRCh38
rs770726543 Chr.22: 17104801-17104801 on Build GRCh38
rs972827656 Chr.22: 17104644-17104644 on Build GRCh38
rs568686550 Chr.22: 17104560-17104560 on Build GRCh38
rs539945067 Chr.22: 17104583-17104583 on Build GRCh38
rs902665119 Chr.22: 17104711-17104711 on Build GRCh38
rs558258253 Chr.22: 17104619-17104619 on Build GRCh38
rs369019806 Chr.22: 17104754-17104754 on Build GRCh38
rs139950741 Chr.22: 17104507-17104507 on Build GRCh38
rs990906378 Chr.22: 17104596-17104596 on Build GRCh38
rs369285103 Chr.22: 17104722-17104722 on Build GRCh38
rs766145662 Chr.22: 17104737-17104737 on Build GRCh38
rs367558296 Chr.22: 17104824-17104824 on Build GRCh38
rs920016888 Chr.22: 17104647-17104647 on Build GRCh38
rs886057202 Chr.22: 17104726-17104726 on Build GRCh38
rs774700286 Chr.22: 17104789-17104789 on Build GRCh38
rs148571704 Chr.22: 17104913-17104913 on Build GRCh38
rs760515134 Chr.22: 17104736-17104736 on Build GRCh38
rs387906913 Chr.22: 17104729-17104729 on Build GRCh38
rs756514784 Chr.22: 17104828-17104828 on Build GRCh38
rs768923702 Chr.22: 17104783-17104783 on Build GRCh38
rs776247658 Chr.22: 17104805-17104805 on Build GRCh38
rs762449094 Chr.22: 17104816-17104816 on Build GRCh38
rs940549275 Chr.22: 17104643-17104643 on Build GRCh38
rs781461932 Chr.22: 17104774-17104774 on Build GRCh38
rs890054699 Chr.22: 17104490-17104490 on Build GRCh38
rs759434781 Chr.22: 17104740-17104740 on Build GRCh38
rs755085731 Chr.22: 17104844-17104844 on Build GRCh38
rs780730644 Chr.22: 17104779-17104779 on Build GRCh38
rs552901103 Chr.22: 17104897-17104897 on Build GRCh38
rs765192966 Chr.22: 17104745-17104745 on Build GRCh38
rs982028746 Chr.22: 17104620-17104620 on Build GRCh38
rs1050195149 Chr.22: 17104674-17104674 on Build GRCh38
rs935490930 Chr.22: 17104891-17104891 on Build GRCh38
rs766332170 Chr.22: 17104877-17104877 on Build GRCh38
rs368399060 Chr.22: 17104681-17104681 on Build GRCh38
rs749457584 Chr.22: 17104676-17104677 on Build GRCh38
rs2302520 Chr.22: 17104575-17104575 on Build GRCh38
rs1041360876 Chr.22: 17104686-17104686 on Build GRCh38
rs41339945 Chr.22: 17104734-17104734 on Build GRCh38
rs145788381 Chr.22: 17104904-17104904 on Build GRCh38
rs770161898 Chr.22: 17104776-17104776 on Build GRCh38
rs777430120 Chr.22: 17104682-17104682 on Build GRCh38
rs966903982 Chr.22: 17104550-17104550 on Build GRCh38
rs566234483 Chr.22: 17104582-17104582 on Build GRCh38
rs371864908 Chr.22: 17104709-17104709 on Build GRCh38
rs1019804434 Chr.22: 17104511-17104511 on Build GRCh38
rs374467653 Chr.22: 17104676-17104676 on Build GRCh38
rs184139865 Chr.22: 17104849-17104849 on Build GRCh38
rs180787596 Chr.22: 17104768-17104768 on Build GRCh38
rs907711602 Chr.22: 17104624-17104624 on Build GRCh38
rs1023512300 Chr.22: 17104599-17104599 on Build GRCh38
rs749867898 Chr.22: 17104840-17104840 on Build GRCh38
rs981973256 Chr.22: 17104770-17104770 on Build GRCh38
rs748237942 Chr.22: 17104850-17104850 on Build GRCh38
rs185454826 Chr.22: 17104565-17104565 on Build GRCh38
rs748288454 Chr.22: 17104855-17104855 on Build GRCh38
rs769892740 Chr.22: 17104728-17104728 on Build GRCh38

View Full Product Details