Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17105507-17105946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL17RA
Entrez Gene ID
23765
Gene Name
interleukin 17 receptor A
Gene Aliases
CANDF5, CD217, CDw217, IL-17RA, IL17R, hIL-17R
UniGene
Hs.48353
Gene Chromosome Location
Chr.22: 17084959-17115694 on Build GRCh38
Transcripts
NM_001289905.1 NM_014339.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6579993 c.932-1G>T p.? Chr.22: 17105590-17105590 on Build GRCh38
4513515 c.941C>A p.P314Q Chr.22: 17105600-17105600 on Build GRCh38
4692194 c.971T>C p.F324S Chr.22: 17105880-17105880 on Build GRCh38
1162502 c.942G>A p.P314P Chr.22: 17105601-17105601 on Build GRCh38
311960 c.959G>T p.W320L Chr.22: 17105868-17105868 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777960930 Chr.22: 17105871-17105871 on Build GRCh38
rs769648701 Chr.22: 17105583-17105583 on Build GRCh38
rs374594183 Chr.22: 17105834-17105834 on Build GRCh38
rs201148897 Chr.22: 17105820-17105820 on Build GRCh38
rs766898199 Chr.22: 17105622-17105622 on Build GRCh38
rs386819495 Chr.22: 17105720-17105721 on Build GRCh38
rs759512999 Chr.22: 17105581-17105581 on Build GRCh38
rs772716073 Chr.22: 17105840-17105840 on Build GRCh38
rs984986020 Chr.22: 17105777-17105777 on Build GRCh38
rs144979983 Chr.22: 17105720-17105720 on Build GRCh38
rs191466693 Chr.22: 17105794-17105794 on Build GRCh38
rs775396300 Chr.22: 17105587-17105587 on Build GRCh38
rs1052965477 Chr.22: 17105836-17105836 on Build GRCh38
rs773943636 Chr.22: 17105829-17105829 on Build GRCh38
rs758399114 Chr.22: 17105869-17105869 on Build GRCh38
rs377254160 Chr.22: 17105605-17105605 on Build GRCh38
rs771451546 Chr.22: 17105919-17105919 on Build GRCh38
rs749026738 Chr.22: 17105892-17105892 on Build GRCh38
rs536619616 Chr.22: 17105715-17105716 on Build GRCh38
rs796678951 Chr.22: 17105719-17105720 on Build GRCh38
rs201595468 Chr.22: 17105837-17105837 on Build GRCh38
rs901663806 Chr.22: 17105718-17105718 on Build GRCh38
rs775810928 Chr.22: 17105717-17105718 on Build GRCh38
rs563873123 Chr.22: 17105809-17105809 on Build GRCh38
rs56736183 Chr.22: 17105826-17105829 on Build GRCh38
rs767174187 Chr.22: 17105831-17105831 on Build GRCh38
rs370875549 Chr.22: 17105848-17105848 on Build GRCh38
rs761432304 Chr.22: 17105829-17105830 on Build GRCh38
rs930419733 Chr.22: 17105776-17105776 on Build GRCh38
rs4819959 Chr.22: 17105741-17105741 on Build GRCh38
rs35078252 Chr.22: 17105724-17105724 on Build GRCh38
rs765592908 Chr.22: 17105863-17105863 on Build GRCh38
rs774360152 Chr.22: 17105833-17105833 on Build GRCh38
rs928135311 Chr.22: 17105656-17105656 on Build GRCh38
rs147505812 Chr.22: 17105914-17105914 on Build GRCh38
rs775307984 Chr.22: 17105819-17105819 on Build GRCh38
rs371742584 Chr.22: 17105857-17105857 on Build GRCh38
rs750095319 Chr.22: 17105618-17105618 on Build GRCh38
rs397780227 Chr.22: 17105830-17105833 on Build GRCh38
rs542664747 Chr.22: 17105678-17105678 on Build GRCh38
rs542177624 Chr.22: 17105720-17105721 on Build GRCh38
rs748115770 Chr.22: 17105633-17105633 on Build GRCh38
rs187020487 Chr.22: 17105623-17105623 on Build GRCh38
rs41321447 Chr.22: 17105601-17105601 on Build GRCh38
rs563863782 Chr.22: 17105721-17105721 on Build GRCh38
rs756532707 Chr.22: 17105621-17105621 on Build GRCh38
rs200724343 Chr.22: 17105861-17105861 on Build GRCh38
rs969401408 Chr.22: 17105721-17105722 on Build GRCh38
rs558196217 Chr.22: 17105699-17105699 on Build GRCh38
rs369108162 Chr.22: 17105725-17105726 on Build GRCh38
rs112474270 Chr.22: 17105731-17105731 on Build GRCh38
rs201611352 Chr.22: 17105719-17105720 on Build GRCh38
rs867675646 Chr.22: 17105713-17105713 on Build GRCh38
rs376227410 Chr.22: 17105647-17105647 on Build GRCh38
rs34484815 Chr.22: 17105705-17105705 on Build GRCh38
rs769458645 Chr.22: 17105641-17105642 on Build GRCh38
rs934442557 Chr.22: 17105828-17105828 on Build GRCh38
rs371093629 Chr.22: 17105726-17105727 on Build GRCh38
rs776585031 Chr.22: 17105576-17105577 on Build GRCh38
rs943043121 Chr.22: 17105804-17105804 on Build GRCh38
rs768533929 Chr.22: 17105823-17105823 on Build GRCh38
rs1035266246 Chr.22: 17105722-17105723 on Build GRCh38
rs138404135 Chr.22: 17105915-17105915 on Build GRCh38
rs575282226 Chr.22: 17105663-17105663 on Build GRCh38
rs768445401 Chr.22: 17105899-17105899 on Build GRCh38
rs143239201 Chr.22: 17105887-17105887 on Build GRCh38
rs762930594 Chr.22: 17105600-17105600 on Build GRCh38
rs773493228 Chr.22: 17105568-17105568 on Build GRCh38
rs2241047 Chr.22: 17105693-17105693 on Build GRCh38
rs371945170 Chr.22: 17105686-17105686 on Build GRCh38
rs11089412 Chr.22: 17105720-17105720 on Build GRCh38
rs2241046 Chr.22: 17105581-17105581 on Build GRCh38
rs750942534 Chr.22: 17105833-17105834 on Build GRCh38
rs998701838 Chr.22: 17105718-17105719 on Build GRCh38
rs533648027 Chr.22: 17105716-17105716 on Build GRCh38
rs370520914 Chr.22: 17105611-17105611 on Build GRCh38
rs769813318 Chr.22: 17105541-17105541 on Build GRCh38
rs775417077 Chr.22: 17105823-17105825 on Build GRCh38
rs763812239 Chr.22: 17105827-17105829 on Build GRCh38
rs751752224 Chr.22: 17105875-17105875 on Build GRCh38
rs560126106 Chr.22: 17105715-17105715 on Build GRCh38
rs796829911 Chr.22: 17105719-17105720 on Build GRCh38
rs140221307 Chr.22: 17105867-17105867 on Build GRCh38
rs200937197 Chr.22: 17105724-17105725 on Build GRCh38
rs747318139 Chr.22: 17105817-17105817 on Build GRCh38
rs369705848 Chr.22: 17105562-17105562 on Build GRCh38
rs770803207 Chr.22: 17105574-17105575 on Build GRCh38
rs918964572 Chr.22: 17105746-17105746 on Build GRCh38

View Full Product Details