Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17538853-17539351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CECR2
Entrez Gene ID
27443
Gene Name
CECR2, histone acetyl-lysine reader
Gene Aliases
-
UniGene
Hs.231895
Gene Chromosome Location
Chr.22: 17359949-17558155 on Build GRCh38
Transcripts
NM_001290046.1 NM_001290047.1 XM_006724079.3 XM_011546128.2 XM_011546129.2 XM_011546132.2 XM_017028785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6257692 c.1262G>T p.G421V Chr.22: 17539036-17539036 on Build GRCh38
338280 c.1326_1327GG>TT p.M442>? Chr.22: 17539016-17539017 on Build GRCh38
1616213 c.1424A>C p.E475A Chr.22: 17539111-17539111 on Build GRCh38
6257691 c.1346G>T p.G449V Chr.22: 17539036-17539036 on Build GRCh38
332378 c.1419G>T p.G473G Chr.22: 17539109-17539109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755888801 Chr.22: 17539179-17539179 on Build GRCh38
rs768057622 Chr.22: 17538951-17538951 on Build GRCh38
rs564460568 Chr.22: 17538919-17538919 on Build GRCh38
rs557578192 Chr.22: 17538927-17538927 on Build GRCh38
rs895855020 Chr.22: 17538912-17538914 on Build GRCh38
rs5747223 Chr.22: 17539271-17539271 on Build GRCh38
rs995207135 Chr.22: 17538882-17538882 on Build GRCh38
rs768298929 Chr.22: 17539036-17539036 on Build GRCh38
rs116033717 Chr.22: 17539132-17539132 on Build GRCh38
rs946188987 Chr.22: 17539150-17539150 on Build GRCh38
rs781006457 Chr.22: 17538999-17538999 on Build GRCh38
rs778337412 Chr.22: 17539152-17539153 on Build GRCh38
rs1025899256 Chr.22: 17538924-17538927 on Build GRCh38
rs376846244 Chr.22: 17539097-17539097 on Build GRCh38
rs62241413 Chr.22: 17539224-17539224 on Build GRCh38
rs764794287 Chr.22: 17539260-17539260 on Build GRCh38
rs747233058 Chr.22: 17539189-17539190 on Build GRCh38
rs775323660 Chr.22: 17539031-17539031 on Build GRCh38
rs774139283 Chr.22: 17539042-17539042 on Build GRCh38
rs937603346 Chr.22: 17539256-17539256 on Build GRCh38
rs139779789 Chr.22: 17539301-17539301 on Build GRCh38
rs773574847 Chr.22: 17539096-17539096 on Build GRCh38
rs768280456 Chr.22: 17539192-17539192 on Build GRCh38
rs1055193730 Chr.22: 17539154-17539154 on Build GRCh38
rs35746048 Chr.22: 17538986-17538986 on Build GRCh38
rs1037196017 Chr.22: 17539212-17539212 on Build GRCh38
rs762251481 Chr.22: 17538948-17538948 on Build GRCh38
rs2300687 Chr.22: 17538915-17538915 on Build GRCh38
rs1010107920 Chr.22: 17539324-17539324 on Build GRCh38
rs765340117 Chr.22: 17539151-17539151 on Build GRCh38
rs774065875 Chr.22: 17539193-17539193 on Build GRCh38
rs779988871 Chr.22: 17539180-17539180 on Build GRCh38
rs777981856 Chr.22: 17539156-17539156 on Build GRCh38
rs369929274 Chr.22: 17539114-17539114 on Build GRCh38
rs750926674 Chr.22: 17538953-17538953 on Build GRCh38
rs745773635 Chr.22: 17539023-17539023 on Build GRCh38
rs375202734 Chr.22: 17538965-17538965 on Build GRCh38
rs570422201 Chr.22: 17539292-17539292 on Build GRCh38
rs769725088 Chr.22: 17539030-17539030 on Build GRCh38
rs539100239 Chr.22: 17539295-17539295 on Build GRCh38
rs758675309 Chr.22: 17539153-17539153 on Build GRCh38
rs1007053584 Chr.22: 17539293-17539293 on Build GRCh38
rs749227836 Chr.22: 17539183-17539183 on Build GRCh38
rs900910734 Chr.22: 17539309-17539309 on Build GRCh38
rs533219201 Chr.22: 17538937-17538937 on Build GRCh38
rs374316521 Chr.22: 17539125-17539125 on Build GRCh38
rs918082735 Chr.22: 17539186-17539186 on Build GRCh38
rs766815433 Chr.22: 17538975-17538975 on Build GRCh38
rs1006610096 Chr.22: 17538926-17538926 on Build GRCh38
rs551789314 Chr.22: 17539287-17539287 on Build GRCh38
rs371376833 Chr.22: 17539078-17539078 on Build GRCh38
rs933732047 Chr.22: 17539014-17539014 on Build GRCh38
rs965201442 Chr.22: 17538934-17538934 on Build GRCh38
rs949547280 Chr.22: 17539197-17539197 on Build GRCh38
rs963957315 Chr.22: 17538978-17538978 on Build GRCh38
rs771285250 Chr.22: 17539193-17539194 on Build GRCh38
rs745606826 Chr.22: 17539165-17539165 on Build GRCh38
rs968604295 Chr.22: 17538968-17538968 on Build GRCh38
rs267606173 Chr.22: 17539107-17539107 on Build GRCh38
rs1296793 Chr.22: 17539251-17539251 on Build GRCh38
rs755016088 Chr.22: 17538979-17538979 on Build GRCh38
rs772299708 Chr.22: 17538987-17538988 on Build GRCh38
rs748427962 Chr.22: 17538987-17538987 on Build GRCh38
rs753913562 Chr.22: 17538976-17538976 on Build GRCh38
rs748769002 Chr.22: 17539035-17539035 on Build GRCh38

View Full Product Details