Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19447821-19448347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C22orf39
Entrez Gene ID
128977
Gene Name
chromosome 22 open reading frame 39
Gene Aliases
-
UniGene
Hs.659565
Gene Chromosome Location
Chr.22: 19440886-19448232 on Build GRCh38
Transcripts
NM_001166242.1 NM_173793.4

SNP Detail

SNP ID Chromosome Location
rs546305014 Chr.22: 19448109-19448109 on Build GRCh38
rs892168304 Chr.22: 19448150-19448150 on Build GRCh38
rs748404555 Chr.22: 19447929-19447929 on Build GRCh38
rs771033143 Chr.22: 19448165-19448165 on Build GRCh38
rs917343318 Chr.22: 19448284-19448284 on Build GRCh38
rs902547421 Chr.22: 19448133-19448133 on Build GRCh38
rs1028817360 Chr.22: 19447896-19447896 on Build GRCh38
rs768594156 Chr.22: 19447978-19447978 on Build GRCh38
rs568610935 Chr.22: 19447888-19447888 on Build GRCh38
rs1017090429 Chr.22: 19447935-19447935 on Build GRCh38
rs1050286569 Chr.22: 19448057-19448057 on Build GRCh38
rs993372984 Chr.22: 19448257-19448257 on Build GRCh38
rs779419669 Chr.22: 19447883-19447883 on Build GRCh38
rs962478018 Chr.22: 19448308-19448308 on Build GRCh38
rs988177500 Chr.22: 19447928-19447928 on Build GRCh38
rs1011381690 Chr.22: 19447901-19447901 on Build GRCh38
rs750971115 Chr.22: 19447905-19447905 on Build GRCh38
rs141397463 Chr.22: 19448033-19448033 on Build GRCh38
rs928960153 Chr.22: 19447956-19447956 on Build GRCh38
rs187278910 Chr.22: 19448155-19448155 on Build GRCh38
rs150350888 Chr.22: 19448136-19448136 on Build GRCh38
rs937909310 Chr.22: 19447977-19447977 on Build GRCh38
rs184743645 Chr.22: 19447904-19447904 on Build GRCh38
rs554851084 Chr.22: 19447945-19447945 on Build GRCh38
rs527891528 Chr.22: 19447899-19447899 on Build GRCh38
rs369407335 Chr.22: 19447936-19447936 on Build GRCh38
rs558162482 Chr.22: 19447930-19447930 on Build GRCh38
rs112682505 Chr.22: 19447932-19447932 on Build GRCh38
rs548892662 Chr.22: 19447881-19447881 on Build GRCh38
rs1029912681 Chr.22: 19447916-19447916 on Build GRCh38
rs527894580 Chr.22: 19448039-19448039 on Build GRCh38
rs547637463 Chr.22: 19447900-19447900 on Build GRCh38
rs1036733227 Chr.22: 19447980-19447980 on Build GRCh38
rs547994899 Chr.22: 19448190-19448190 on Build GRCh38
rs542201581 Chr.22: 19448156-19448156 on Build GRCh38
rs34515388 Chr.22: 19447870-19447870 on Build GRCh38
rs562417708 Chr.22: 19448160-19448160 on Build GRCh38
rs538876779 Chr.22: 19447966-19447966 on Build GRCh38
rs1007119504 Chr.22: 19448205-19448205 on Build GRCh38
rs1036295663 Chr.22: 19447878-19447878 on Build GRCh38
rs553658558 Chr.22: 19448019-19448019 on Build GRCh38
rs950460916 Chr.22: 19448317-19448317 on Build GRCh38
rs1046724664 Chr.22: 19447855-19447855 on Build GRCh38
rs1009313968 Chr.22: 19448152-19448152 on Build GRCh38
rs973546920 Chr.22: 19447941-19447941 on Build GRCh38
rs574602781 Chr.22: 19447948-19447948 on Build GRCh38
rs113953722 Chr.22: 19447969-19447969 on Build GRCh38
rs1040130089 Chr.22: 19448142-19448142 on Build GRCh38
rs906413845 Chr.22: 19447866-19447866 on Build GRCh38
rs183651653 Chr.22: 19448222-19448222 on Build GRCh38
rs191364874 Chr.22: 19448180-19448180 on Build GRCh38
rs780021951 Chr.22: 19447921-19447921 on Build GRCh38
rs992932295 Chr.22: 19447961-19447961 on Build GRCh38
rs981742532 Chr.22: 19448325-19448325 on Build GRCh38
rs895841032 Chr.22: 19447864-19447864 on Build GRCh38
rs963184733 Chr.22: 19448232-19448232 on Build GRCh38
rs973215911 Chr.22: 19448235-19448235 on Build GRCh38
rs1057226387 Chr.22: 19447850-19447850 on Build GRCh38
rs893629148 Chr.22: 19448102-19448102 on Build GRCh38
rs920603965 Chr.22: 19447994-19447994 on Build GRCh38
rs917293554 Chr.22: 19447967-19447967 on Build GRCh38
rs754813713 Chr.22: 19447844-19447844 on Build GRCh38
rs940664597 Chr.22: 19447867-19447867 on Build GRCh38
rs117016465 Chr.22: 19448122-19448122 on Build GRCh38
rs147861375 Chr.22: 19447848-19447848 on Build GRCh38
rs1026473555 Chr.22: 19448276-19448276 on Build GRCh38
rs538322767 Chr.22: 19447914-19447914 on Build GRCh38

View Full Product Details