Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19968449-19968955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARVCF
Entrez Gene ID
421
Gene Name
armadillo repeat gene deleted in velocardiofacial syndrome
Gene Aliases
-
UniGene
Hs.713616
Gene Chromosome Location
Chr.22: 19966491-20016802 on Build GRCh38
Transcripts
XM_005261242.2 XM_006724243.2 XM_006724246.3 XM_011530179.2 XM_011530182.2 NM_000754.3 NM_001135161.1 NM_001135162.1 NM_007310.2 XM_011529886.1 XM_017028594.1 XM_017028595.1

Gene Symbol
COMT
Entrez Gene ID
1312
Gene Name
catechol-O-methyltransferase
Gene Aliases
HEL-S-98n
UniGene
Hs.370408
Gene Chromosome Location
Chr.22: 19941740-19969975 on Build GRCh38
Transcripts
XM_005261242.2 XM_006724243.2 XM_006724246.3 XM_011530179.2 XM_011530182.2 NM_000754.3 NM_001135161.1 NM_001135162.1 NM_007310.2 XM_011529886.1 XM_017028594.1 XM_017028595.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3390096 c.616-5T>C p.? Chr.22: 19968531-19968531 on Build GRCh38
6413423 c.802G>A p.E268K Chr.22: 19968722-19968722 on Build GRCh38
6419885 c.632G>A p.R211Q Chr.22: 19968552-19968552 on Build GRCh38
4102632 c.752G>T p.R251M Chr.22: 19968672-19968672 on Build GRCh38
2892560 c.696C>T p.H232H Chr.22: 19968616-19968616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752035611 Chr.22: 19968647-19968647 on Build GRCh38
rs762226319 Chr.22: 19968739-19968739 on Build GRCh38
rs138327309 Chr.22: 19968747-19968747 on Build GRCh38
rs774375300 Chr.22: 19968517-19968517 on Build GRCh38
rs760480469 Chr.22: 19968609-19968609 on Build GRCh38
rs529947354 Chr.22: 19968928-19968928 on Build GRCh38
rs773725345 Chr.22: 19968773-19968773 on Build GRCh38
rs755852074 Chr.22: 19968747-19968747 on Build GRCh38
rs752735911 Chr.22: 19968744-19968745 on Build GRCh38
rs893987628 Chr.22: 19968616-19968617 on Build GRCh38
rs536538848 Chr.22: 19968598-19968598 on Build GRCh38
rs928195361 Chr.22: 19968818-19968818 on Build GRCh38
rs899829083 Chr.22: 19968583-19968583 on Build GRCh38
rs1030230368 Chr.22: 19968711-19968711 on Build GRCh38
rs201656482 Chr.22: 19968555-19968555 on Build GRCh38
rs9332380 Chr.22: 19968894-19968894 on Build GRCh38
rs748900622 Chr.22: 19968762-19968762 on Build GRCh38
rs973872583 Chr.22: 19968875-19968875 on Build GRCh38
rs555126080 Chr.22: 19968722-19968722 on Build GRCh38
rs939191728 Chr.22: 19968475-19968475 on Build GRCh38
rs139449932 Chr.22: 19968620-19968620 on Build GRCh38
rs750159038 Chr.22: 19968558-19968558 on Build GRCh38
rs745628165 Chr.22: 19968755-19968755 on Build GRCh38
rs201407028 Chr.22: 19968686-19968686 on Build GRCh38
rs768425882 Chr.22: 19968763-19968763 on Build GRCh38
rs775160442 Chr.22: 19968716-19968716 on Build GRCh38
rs941816813 Chr.22: 19968503-19968503 on Build GRCh38
rs772665216 Chr.22: 19968704-19968705 on Build GRCh38
rs201476400 Chr.22: 19968629-19968629 on Build GRCh38
rs1002298934 Chr.22: 19968847-19968847 on Build GRCh38
rs962542027 Chr.22: 19968867-19968867 on Build GRCh38
rs754435205 Chr.22: 19968535-19968535 on Build GRCh38
rs397758621 Chr.22: 19968746-19968747 on Build GRCh38
rs369908847 Chr.22: 19968712-19968712 on Build GRCh38
rs891228883 Chr.22: 19968764-19968764 on Build GRCh38
rs889790482 Chr.22: 19968882-19968882 on Build GRCh38
rs987233753 Chr.22: 19968925-19968925 on Build GRCh38
rs761850766 Chr.22: 19968519-19968519 on Build GRCh38
rs776763589 Chr.22: 19968504-19968504 on Build GRCh38
rs960769182 Chr.22: 19968858-19968858 on Build GRCh38
rs767422215 Chr.22: 19968521-19968521 on Build GRCh38
rs550339713 Chr.22: 19968538-19968538 on Build GRCh38
rs202111640 Chr.22: 19968691-19968691 on Build GRCh38
rs747375116 Chr.22: 19968557-19968557 on Build GRCh38
rs368314788 Chr.22: 19968753-19968753 on Build GRCh38
rs768651434 Chr.22: 19968677-19968677 on Build GRCh38
rs769605719 Chr.22: 19968756-19968756 on Build GRCh38
rs1035197522 Chr.22: 19968674-19968674 on Build GRCh38
rs550545168 Chr.22: 19968903-19968903 on Build GRCh38
rs181896876 Chr.22: 19968838-19968838 on Build GRCh38
rs973669692 Chr.22: 19968905-19968905 on Build GRCh38
rs779954250 Chr.22: 19968754-19968755 on Build GRCh38
rs749349777 Chr.22: 19968670-19968670 on Build GRCh38
rs12814 Chr.22: 19968769-19968769 on Build GRCh38
rs1053252285 Chr.22: 19968796-19968796 on Build GRCh38
rs770652365 Chr.22: 19968615-19968615 on Build GRCh38
rs540916953 Chr.22: 19968823-19968823 on Build GRCh38
rs758187540 Chr.22: 19968593-19968593 on Build GRCh38
rs200150695 Chr.22: 19968621-19968621 on Build GRCh38
rs753519008 Chr.22: 19968532-19968532 on Build GRCh38
rs778615118 Chr.22: 19968517-19968517 on Build GRCh38
rs999446749 Chr.22: 19968617-19968617 on Build GRCh38
rs758703641 Chr.22: 19968487-19968487 on Build GRCh38
rs752040355 Chr.22: 19968731-19968731 on Build GRCh38
rs1034674434 Chr.22: 19968853-19968853 on Build GRCh38
rs778861844 Chr.22: 19968751-19968751 on Build GRCh38
rs750361935 Chr.22: 19968745-19968746 on Build GRCh38
rs757585867 Chr.22: 19968648-19968648 on Build GRCh38
rs199744942 Chr.22: 19968698-19968698 on Build GRCh38
rs376273380 Chr.22: 19968573-19968573 on Build GRCh38
rs761439220 Chr.22: 19968785-19968785 on Build GRCh38
rs117222687 Chr.22: 19968491-19968491 on Build GRCh38
rs1011662017 Chr.22: 19968817-19968817 on Build GRCh38
rs560296967 Chr.22: 19968824-19968824 on Build GRCh38
rs752185888 Chr.22: 19968623-19968623 on Build GRCh38
rs567830018 Chr.22: 19968616-19968616 on Build GRCh38
rs531680429 Chr.22: 19968902-19968902 on Build GRCh38
rs780473420 Chr.22: 19968658-19968658 on Build GRCh38
rs1055337641 Chr.22: 19968608-19968608 on Build GRCh38
rs781428653 Chr.22: 19968559-19968559 on Build GRCh38
rs75081694 Chr.22: 19968748-19968748 on Build GRCh38
rs545915950 Chr.22: 19968857-19968857 on Build GRCh38
rs762414852 Chr.22: 19968634-19968634 on Build GRCh38
rs781417317 Chr.22: 19968652-19968652 on Build GRCh38
rs371134244 Chr.22: 19968497-19968497 on Build GRCh38
rs765203563 Chr.22: 19968742-19968743 on Build GRCh38
rs372535915 Chr.22: 19968657-19968657 on Build GRCh38
rs76902350 Chr.22: 19968746-19968746 on Build GRCh38
rs199743418 Chr.22: 19968754-19968754 on Build GRCh38
rs879060530 Chr.22: 19968897-19968897 on Build GRCh38
rs373882300 Chr.22: 19968522-19968522 on Build GRCh38
rs775072525 Chr.22: 19968759-19968759 on Build GRCh38
rs759852947 Chr.22: 19968741-19968742 on Build GRCh38
rs571126788 Chr.22: 19968515-19968515 on Build GRCh38
rs775712233 Chr.22: 19968581-19968581 on Build GRCh38
rs764628606 Chr.22: 19968643-19968643 on Build GRCh38
rs754031677 Chr.22: 19968749-19968749 on Build GRCh38
rs561135670 Chr.22: 19968927-19968927 on Build GRCh38
rs773064787 Chr.22: 19968602-19968602 on Build GRCh38
rs771394687 Chr.22: 19968740-19968740 on Build GRCh38
rs745909655 Chr.22: 19968512-19968512 on Build GRCh38
rs567483468 Chr.22: 19968549-19968549 on Build GRCh38
rs934748900 Chr.22: 19968788-19968788 on Build GRCh38
rs749437638 Chr.22: 19968597-19968597 on Build GRCh38
rs769932706 Chr.22: 19968580-19968580 on Build GRCh38
rs750888068 Chr.22: 19968655-19968655 on Build GRCh38
rs749822167 Chr.22: 19968866-19968866 on Build GRCh38
rs566414524 Chr.22: 19968680-19968680 on Build GRCh38
rs1003960733 Chr.22: 19968864-19968864 on Build GRCh38
rs768200645 Chr.22: 19968744-19968744 on Build GRCh38
rs14968 Chr.22: 19968910-19968911 on Build GRCh38
rs777714694 Chr.22: 19968679-19968679 on Build GRCh38
rs893303657 Chr.22: 19968797-19968797 on Build GRCh38
rs1007390072 Chr.22: 19968890-19968890 on Build GRCh38
rs766025497 Chr.22: 19968524-19968524 on Build GRCh38
rs201893998 Chr.22: 19968685-19968685 on Build GRCh38
rs748633677 Chr.22: 19968662-19968663 on Build GRCh38
rs369589068 Chr.22: 19968622-19968622 on Build GRCh38
rs546326145 Chr.22: 19968661-19968661 on Build GRCh38
rs760438359 Chr.22: 19968523-19968523 on Build GRCh38
rs144463570 Chr.22: 19968551-19968551 on Build GRCh38
rs879893042 Chr.22: 19968911-19968911 on Build GRCh38
rs754537148 Chr.22: 19968692-19968692 on Build GRCh38

View Full Product Details